Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92937435-92937954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs560889080 Chr.14: 92937476-92937476 on Build GRCh38
rs900928076 Chr.14: 92937887-92937887 on Build GRCh38
rs116839957 Chr.14: 92937908-92937908 on Build GRCh38
rs1008859514 Chr.14: 92937772-92937772 on Build GRCh38
rs953931795 Chr.14: 92937736-92937736 on Build GRCh38
rs1007034253 Chr.14: 92937745-92937745 on Build GRCh38
rs540919471 Chr.14: 92937795-92937795 on Build GRCh38
rs976310085 Chr.14: 92937555-92937555 on Build GRCh38
rs995759736 Chr.14: 92937497-92937497 on Build GRCh38
rs924955989 Chr.14: 92937895-92937895 on Build GRCh38
rs995312044 Chr.14: 92937720-92937720 on Build GRCh38
rs751749172 Chr.14: 92937807-92937807 on Build GRCh38
rs1020360979 Chr.14: 92937907-92937907 on Build GRCh38
rs753721071 Chr.14: 92937686-92937686 on Build GRCh38
rs186143018 Chr.14: 92937854-92937854 on Build GRCh38
rs576385755 Chr.14: 92937718-92937718 on Build GRCh38
rs1026700436 Chr.14: 92937510-92937510 on Build GRCh38
rs945054395 Chr.14: 92937680-92937680 on Build GRCh38
rs567481776 Chr.14: 92937662-92937662 on Build GRCh38
rs1044288365 Chr.14: 92937613-92937613 on Build GRCh38
rs896523983 Chr.14: 92937484-92937484 on Build GRCh38
rs977774243 Chr.14: 92937891-92937891 on Build GRCh38
rs536438267 Chr.14: 92937667-92937667 on Build GRCh38
rs1036995548 Chr.14: 92937839-92937839 on Build GRCh38
rs556285518 Chr.14: 92937690-92937690 on Build GRCh38
rs1027022832 Chr.14: 92937565-92937565 on Build GRCh38
rs959259054 Chr.14: 92937602-92937602 on Build GRCh38
rs1006743281 Chr.14: 92937882-92937882 on Build GRCh38
rs897189288 Chr.14: 92937840-92937840 on Build GRCh38
rs551866479 Chr.14: 92937528-92937528 on Build GRCh38
rs111541366 Chr.14: 92937796-92937796 on Build GRCh38
rs527943694 Chr.14: 92937621-92937621 on Build GRCh38
rs1041631113 Chr.14: 92937926-92937926 on Build GRCh38
rs558647933 Chr.14: 92937758-92937758 on Build GRCh38
rs961407285 Chr.14: 92937522-92937524 on Build GRCh38
rs931211482 Chr.14: 92937844-92937844 on Build GRCh38
rs952821385 Chr.14: 92937566-92937566 on Build GRCh38
rs984574821 Chr.14: 92937567-92937567 on Build GRCh38
rs992092422 Chr.14: 92937643-92937643 on Build GRCh38
rs1036879661 Chr.14: 92937698-92937698 on Build GRCh38
rs559057096 Chr.14: 92937927-92937927 on Build GRCh38
rs190464815 Chr.14: 92937861-92937861 on Build GRCh38
rs760385623 Chr.14: 92937461-92937461 on Build GRCh38
rs552715768 Chr.14: 92937624-92937624 on Build GRCh38
rs925511836 Chr.14: 92937622-92937622 on Build GRCh38
rs60511400 Chr.14: 92937490-92937490 on Build GRCh38
rs1015486961 Chr.14: 92937491-92937491 on Build GRCh38
rs1019394392 Chr.14: 92937762-92937762 on Build GRCh38
rs80252630 Chr.14: 92937782-92937782 on Build GRCh38
rs1049895473 Chr.14: 92937740-92937740 on Build GRCh38
rs538519007 Chr.14: 92937794-92937794 on Build GRCh38
rs1042487732 Chr.14: 92937689-92937689 on Build GRCh38
rs1032611857 Chr.14: 92937928-92937928 on Build GRCh38
rs2281512 Chr.14: 92937803-92937803 on Build GRCh38
rs1013558012 Chr.14: 92937677-92937677 on Build GRCh38
rs1055164825 Chr.14: 92937665-92937665 on Build GRCh38
rs1001591916 Chr.14: 92937825-92937825 on Build GRCh38
rs945532150 Chr.14: 92937608-92937608 on Build GRCh38
rs146992679 Chr.14: 92937495-92937495 on Build GRCh38
rs994607953 Chr.14: 92937524-92937524 on Build GRCh38
rs571941678 Chr.14: 92937553-92937553 on Build GRCh38
rs925427891 Chr.14: 92937818-92937818 on Build GRCh38
rs543637279 Chr.14: 92937819-92937819 on Build GRCh38
rs574723673 Chr.14: 92937805-92937805 on Build GRCh38
rs2281513 Chr.14: 92937521-92937521 on Build GRCh38
rs918013274 Chr.14: 92937832-92937832 on Build GRCh38
rs547974263 Chr.14: 92937653-92937653 on Build GRCh38
rs779002399 Chr.14: 92937724-92937724 on Build GRCh38
rs569494603 Chr.14: 92937631-92937631 on Build GRCh38
rs1010030570 Chr.14: 92937902-92937902 on Build GRCh38
rs949352486 Chr.14: 92937836-92937836 on Build GRCh38
rs1019341820 Chr.14: 92937884-92937884 on Build GRCh38
rs914173333 Chr.14: 92937557-92937557 on Build GRCh38
rs539060878 Chr.14: 92937747-92937747 on Build GRCh38
rs1048331934 Chr.14: 92937849-92937849 on Build GRCh38
rs117559055 Chr.14: 92937511-92937511 on Build GRCh38
rs145601942 Chr.14: 92937576-92937577 on Build GRCh38
rs936436634 Chr.14: 92937739-92937739 on Build GRCh38
rs573302978 Chr.14: 92937822-92937822 on Build GRCh38

View Full Product Details