Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93720879-93721388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIMA1
Entrez Gene ID
145270
Gene Name
proline rich membrane anchor 1
Gene Aliases
PRIMA
UniGene
Hs.432401
Gene Chromosome Location
Chr.14: 93718298-93789029 on Build GRCh38
Transcripts
NM_178013.3 XM_011536456.2

SNP Detail

SNP ID Chromosome Location
rs964571400 Chr.14: 93721025-93721025 on Build GRCh38
rs527552442 Chr.14: 93721133-93721133 on Build GRCh38
rs910408607 Chr.14: 93721009-93721009 on Build GRCh38
rs981674631 Chr.14: 93721073-93721073 on Build GRCh38
rs745482818 Chr.14: 93720981-93720981 on Build GRCh38
rs181714306 Chr.14: 93721145-93721145 on Build GRCh38
rs962765804 Chr.14: 93721054-93721054 on Build GRCh38
rs564407443 Chr.14: 93721285-93721285 on Build GRCh38
rs916047213 Chr.14: 93721056-93721056 on Build GRCh38
rs994400355 Chr.14: 93721161-93721161 on Build GRCh38
rs759255492 Chr.14: 93721055-93721055 on Build GRCh38
rs923071915 Chr.14: 93721253-93721253 on Build GRCh38
rs1025999999 Chr.14: 93721162-93721162 on Build GRCh38
rs916678774 Chr.14: 93721313-93721313 on Build GRCh38
rs951681793 Chr.14: 93721165-93721165 on Build GRCh38
rs370158053 Chr.14: 93721125-93721125 on Build GRCh38
rs975942273 Chr.14: 93721234-93721234 on Build GRCh38
rs555191959 Chr.14: 93721120-93721120 on Build GRCh38
rs142213106 Chr.14: 93721357-93721357 on Build GRCh38
rs562976248 Chr.14: 93721252-93721252 on Build GRCh38
rs936673048 Chr.14: 93721301-93721301 on Build GRCh38
rs936786108 Chr.14: 93721124-93721124 on Build GRCh38
rs897288033 Chr.14: 93721328-93721328 on Build GRCh38
rs544653715 Chr.14: 93721248-93721248 on Build GRCh38
rs1006000362 Chr.14: 93721224-93721224 on Build GRCh38
rs185525697 Chr.14: 93720987-93720987 on Build GRCh38
rs966610399 Chr.14: 93721255-93721255 on Build GRCh38
rs930243406 Chr.14: 93721347-93721347 on Build GRCh38
rs1025223108 Chr.14: 93720914-93720914 on Build GRCh38
rs569352381 Chr.14: 93721034-93721034 on Build GRCh38
rs1042388013 Chr.14: 93721067-93721067 on Build GRCh38
rs1014277928 Chr.14: 93720913-93720913 on Build GRCh38
rs928824023 Chr.14: 93721288-93721288 on Build GRCh38
rs925256673 Chr.14: 93721075-93721075 on Build GRCh38
rs1017345216 Chr.14: 93721231-93721231 on Build GRCh38
rs536777588 Chr.14: 93721077-93721077 on Build GRCh38
rs977965606 Chr.14: 93721267-93721267 on Build GRCh38
rs186727272 Chr.14: 93721314-93721314 on Build GRCh38
rs577484404 Chr.14: 93721146-93721146 on Build GRCh38
rs934944862 Chr.14: 93721105-93721108 on Build GRCh38
rs180733875 Chr.14: 93720952-93720952 on Build GRCh38
rs976437104 Chr.14: 93721037-93721037 on Build GRCh38
rs1013746965 Chr.14: 93721136-93721136 on Build GRCh38
rs1038730526 Chr.14: 93721339-93721339 on Build GRCh38
rs546530059 Chr.14: 93720946-93720946 on Build GRCh38
rs145017673 Chr.14: 93721132-93721132 on Build GRCh38
rs923220031 Chr.14: 93721038-93721038 on Build GRCh38
rs964779597 Chr.14: 93721232-93721232 on Build GRCh38
rs528231784 Chr.14: 93721326-93721326 on Build GRCh38
rs995567820 Chr.14: 93721258-93721258 on Build GRCh38
rs574983590 Chr.14: 93721268-93721268 on Build GRCh38
rs776238968 Chr.14: 93720951-93720951 on Build GRCh38
rs75767365 Chr.14: 93721302-93721302 on Build GRCh38
rs760582945 Chr.14: 93721160-93721160 on Build GRCh38
rs1039197324 Chr.14: 93721135-93721135 on Build GRCh38
rs189930415 Chr.14: 93721004-93721004 on Build GRCh38
rs762882344 Chr.14: 93721278-93721278 on Build GRCh38
rs895302455 Chr.14: 93720907-93720907 on Build GRCh38
rs886339744 Chr.14: 93721163-93721163 on Build GRCh38

View Full Product Details