Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96284633-96285182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2B
Entrez Gene ID
55102
Gene Name
autophagy related 2B
Gene Aliases
C14orf103
UniGene
Hs.168241
Gene Chromosome Location
Chr.14: 96281258-96363341 on Build GRCh38
Transcripts
XM_006720187.2

SNP Detail

SNP ID Chromosome Location
rs8019013 Chr.14: 96284673-96284673 on Build GRCh38
rs924226824 Chr.14: 96284743-96284743 on Build GRCh38
rs374900061 Chr.14: 96284737-96284737 on Build GRCh38
rs386780406 Chr.14: 96284672-96284673 on Build GRCh38
rs867099237 Chr.14: 96285119-96285119 on Build GRCh38
rs1043235764 Chr.14: 96285018-96285018 on Build GRCh38
rs138766942 Chr.14: 96284701-96284701 on Build GRCh38
rs1047664756 Chr.14: 96284909-96284909 on Build GRCh38
rs892063867 Chr.14: 96284954-96284954 on Build GRCh38
rs558848061 Chr.14: 96284915-96284915 on Build GRCh38
rs911606612 Chr.14: 96284899-96284899 on Build GRCh38
rs576790235 Chr.14: 96284724-96284724 on Build GRCh38
rs549106046 Chr.14: 96284858-96284858 on Build GRCh38
rs1014903197 Chr.14: 96285101-96285101 on Build GRCh38
rs920406595 Chr.14: 96284788-96284788 on Build GRCh38
rs977743544 Chr.14: 96285093-96285093 on Build GRCh38
rs960027649 Chr.14: 96284741-96284741 on Build GRCh38
rs144584949 Chr.14: 96284672-96284672 on Build GRCh38
rs1014683309 Chr.14: 96285097-96285097 on Build GRCh38
rs973659854 Chr.14: 96284773-96284773 on Build GRCh38
rs751842759 Chr.14: 96284979-96284979 on Build GRCh38
rs767661152 Chr.14: 96285150-96285150 on Build GRCh38
rs1052080630 Chr.14: 96284936-96284936 on Build GRCh38
rs916726351 Chr.14: 96285152-96285152 on Build GRCh38
rs183862169 Chr.14: 96284714-96284714 on Build GRCh38
rs142354246 Chr.14: 96284670-96284670 on Build GRCh38
rs763430781 Chr.14: 96284791-96284791 on Build GRCh38
rs566090807 Chr.14: 96285041-96285042 on Build GRCh38
rs561493587 Chr.14: 96284839-96284839 on Build GRCh38
rs948141430 Chr.14: 96284810-96284810 on Build GRCh38
rs929984878 Chr.14: 96284866-96284866 on Build GRCh38
rs961959293 Chr.14: 96284763-96284763 on Build GRCh38
rs944380999 Chr.14: 96284930-96284930 on Build GRCh38
rs1034348683 Chr.14: 96284739-96284739 on Build GRCh38
rs3809422 Chr.14: 96285021-96285021 on Build GRCh38
rs985868129 Chr.14: 96284879-96284879 on Build GRCh38
rs116785545 Chr.14: 96285055-96285055 on Build GRCh38
rs973328861 Chr.14: 96285148-96285148 on Build GRCh38
rs961249007 Chr.14: 96285125-96285125 on Build GRCh38
rs1001614786 Chr.14: 96285031-96285031 on Build GRCh38
rs771654305 Chr.14: 96284666-96284666 on Build GRCh38
rs900368910 Chr.14: 96285036-96285036 on Build GRCh38
rs867482070 Chr.14: 96285140-96285140 on Build GRCh38
rs1031186883 Chr.14: 96285102-96285102 on Build GRCh38
rs974403491 Chr.14: 96284685-96284685 on Build GRCh38
rs992802996 Chr.14: 96284760-96284760 on Build GRCh38
rs1015263584 Chr.14: 96284761-96284761 on Build GRCh38
rs573582694 Chr.14: 96284910-96284910 on Build GRCh38
rs895865467 Chr.14: 96285063-96285063 on Build GRCh38
rs568455939 Chr.14: 96284663-96284663 on Build GRCh38
rs1038486970 Chr.14: 96284812-96284813 on Build GRCh38
rs544118554 Chr.14: 96284809-96284809 on Build GRCh38

View Full Product Details