Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100467929-100468432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
UniGene
Hs.497600
Gene Chromosome Location
Chr.14: 100376410-100530303 on Build GRCh38
Transcripts
NM_001161476.1 NM_024515.4 XM_005268056.4 XM_005268057.3 XM_005268058.4 XM_011537150.2 XM_011537151.2 XM_017021639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4243323 c.916C>T p.R306C Chr.14: 100468114-100468114 on Build GRCh38
4049075 c.116G>T p.R39L Chr.14: 100468085-100468085 on Build GRCh38
5176815 c.111A>C p.A37A Chr.14: 100468080-100468080 on Build GRCh38
3885623 c.171C>T p.F57F Chr.14: 100468140-100468140 on Build GRCh38
6821666 c.831C>T p.N277N Chr.14: 100468029-100468029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375715238 Chr.14: 100468087-100468087 on Build GRCh38
rs745418254 Chr.14: 100468101-100468101 on Build GRCh38
rs1016249536 Chr.14: 100468404-100468404 on Build GRCh38
rs760824912 Chr.14: 100468014-100468014 on Build GRCh38
rs762801190 Chr.14: 100468064-100468064 on Build GRCh38
rs765497257 Chr.14: 100468130-100468130 on Build GRCh38
rs769004491 Chr.14: 100468259-100468259 on Build GRCh38
rs778400423 Chr.14: 100468032-100468032 on Build GRCh38
rs750461876 Chr.14: 100468005-100468005 on Build GRCh38
rs978354948 Chr.14: 100468369-100468369 on Build GRCh38
rs117591044 Chr.14: 100468191-100468191 on Build GRCh38
rs202134560 Chr.14: 100468089-100468089 on Build GRCh38
rs760634961 Chr.14: 100468075-100468075 on Build GRCh38
rs551230616 Chr.14: 100468260-100468260 on Build GRCh38
rs1015874036 Chr.14: 100468230-100468230 on Build GRCh38
rs772034827 Chr.14: 100468210-100468210 on Build GRCh38
rs759749617 Chr.14: 100468126-100468126 on Build GRCh38
rs963330565 Chr.14: 100468407-100468407 on Build GRCh38
rs900520831 Chr.14: 100468154-100468154 on Build GRCh38
rs771944170 Chr.14: 100468120-100468120 on Build GRCh38
rs1033331203 Chr.14: 100468079-100468079 on Build GRCh38
rs775806248 Chr.14: 100468217-100468217 on Build GRCh38
rs150647230 Chr.14: 100467957-100467957 on Build GRCh38
rs77046356 Chr.14: 100467999-100467999 on Build GRCh38
rs370067304 Chr.14: 100468090-100468090 on Build GRCh38
rs6575783 Chr.14: 100467989-100467989 on Build GRCh38
rs781070959 Chr.14: 100468152-100468152 on Build GRCh38
rs1011767898 Chr.14: 100467992-100467992 on Build GRCh38
rs767972981 Chr.14: 100468167-100468167 on Build GRCh38
rs770476522 Chr.14: 100467979-100467979 on Build GRCh38
rs369639749 Chr.14: 100468118-100468118 on Build GRCh38
rs115563176 Chr.14: 100468244-100468244 on Build GRCh38
rs35620735 Chr.14: 100468093-100468094 on Build GRCh38
rs764054927 Chr.14: 100468002-100468002 on Build GRCh38
rs371733197 Chr.14: 100468073-100468073 on Build GRCh38
rs746445610 Chr.14: 100468040-100468040 on Build GRCh38
rs139730965 Chr.14: 100468099-100468099 on Build GRCh38
rs779398443 Chr.14: 100468023-100468023 on Build GRCh38
rs770479232 Chr.14: 100468041-100468041 on Build GRCh38
rs766449819 Chr.14: 100468018-100468018 on Build GRCh38
rs199801334 Chr.14: 100468093-100468093 on Build GRCh38
rs1012932438 Chr.14: 100468397-100468397 on Build GRCh38
rs745875130 Chr.14: 100468212-100468212 on Build GRCh38
rs770023027 Chr.14: 100468213-100468213 on Build GRCh38
rs776229615 Chr.14: 100468191-100468192 on Build GRCh38
rs565627567 Chr.14: 100468340-100468340 on Build GRCh38
rs193004327 Chr.14: 100468273-100468273 on Build GRCh38
rs775232071 Chr.14: 100468060-100468060 on Build GRCh38
rs780668010 Chr.14: 100468042-100468042 on Build GRCh38
rs753155095 Chr.14: 100468028-100468028 on Build GRCh38
rs146541699 Chr.14: 100468094-100468094 on Build GRCh38
rs372743834 Chr.14: 100468172-100468172 on Build GRCh38
rs559488785 Chr.14: 100468111-100468111 on Build GRCh38
rs551792043 Chr.14: 100468141-100468141 on Build GRCh38
rs866576236 Chr.14: 100468297-100468297 on Build GRCh38
rs796052162 Chr.14: 100468038-100468038 on Build GRCh38
rs773202736 Chr.14: 100468331-100468331 on Build GRCh38
rs748965751 Chr.14: 100468196-100468196 on Build GRCh38
rs754125168 Chr.14: 100468020-100468020 on Build GRCh38
rs1004128214 Chr.14: 100468225-100468225 on Build GRCh38
rs912069384 Chr.14: 100468388-100468388 on Build GRCh38
rs768422606 Chr.14: 100468068-100468068 on Build GRCh38
rs906350419 Chr.14: 100468240-100468240 on Build GRCh38
rs745538384 Chr.14: 100468044-100468044 on Build GRCh38
rs199882869 Chr.14: 100467982-100467982 on Build GRCh38
rs776848385 Chr.14: 100468124-100468124 on Build GRCh38
rs555395521 Chr.14: 100468299-100468299 on Build GRCh38
rs766506215 Chr.14: 100468082-100468082 on Build GRCh38
rs138735540 Chr.14: 100468112-100468112 on Build GRCh38
rs755347286 Chr.14: 100468021-100468021 on Build GRCh38
rs754640546 Chr.14: 100468203-100468203 on Build GRCh38
rs778739472 Chr.14: 100468207-100468207 on Build GRCh38
rs376398808 Chr.14: 100468116-100468116 on Build GRCh38
rs528448819 Chr.14: 100468115-100468115 on Build GRCh38
rs4905966 Chr.14: 100468265-100468265 on Build GRCh38
rs758971019 Chr.14: 100468029-100468029 on Build GRCh38
rs775857537 Chr.14: 100468136-100468136 on Build GRCh38
rs763187661 Chr.14: 100468140-100468140 on Build GRCh38
rs775355059 Chr.14: 100467998-100467998 on Build GRCh38
rs746115534 Chr.14: 100468096-100468096 on Build GRCh38
rs529381627 Chr.14: 100468084-100468084 on Build GRCh38
rs755628133 Chr.14: 100468186-100468186 on Build GRCh38
rs930600339 Chr.14: 100468174-100468174 on Build GRCh38
rs751915088 Chr.14: 100468142-100468142 on Build GRCh38
rs768269075 Chr.14: 100468114-100468114 on Build GRCh38
rs967399418 Chr.14: 100468343-100468343 on Build GRCh38
rs563125299 Chr.14: 100468059-100468059 on Build GRCh38
rs376036953 Chr.14: 100468209-100468209 on Build GRCh38

View Full Product Details