Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100728289-100728795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLK1
Entrez Gene ID
8788
Gene Name
delta like non-canonical Notch ligand 1
Gene Aliases
DLK, DLK-1, Delta1, FA1, PREF1, Pref-1, ZOG, pG2
UniGene
Hs.116631
Gene Chromosome Location
Chr.14: 100726865-100738224 on Build GRCh38
Transcripts
NM_001317172.1 NM_003836.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6481424 c.78C>A p.C26* Chr.14: 100728406-100728406 on Build GRCh38
2261936 c.90C>A p.C30* Chr.14: 100728418-100728418 on Build GRCh38
6645036 c.93C>T p.N31N Chr.14: 100728421-100728421 on Build GRCh38
319910 c.118G>T p.D40Y Chr.14: 100728446-100728446 on Build GRCh38
5886835 c.131+7G>A p.? Chr.14: 100728466-100728466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535164022 Chr.14: 100728531-100728532 on Build GRCh38
rs932645396 Chr.14: 100728545-100728545 on Build GRCh38
rs1041656986 Chr.14: 100728721-100728721 on Build GRCh38
rs747243105 Chr.14: 100728399-100728399 on Build GRCh38
rs982957953 Chr.14: 100728722-100728722 on Build GRCh38
rs923901730 Chr.14: 100728726-100728726 on Build GRCh38
rs958544813 Chr.14: 100728580-100728580 on Build GRCh38
rs543495125 Chr.14: 100728699-100728699 on Build GRCh38
rs370270261 Chr.14: 100728472-100728472 on Build GRCh38
rs369649317 Chr.14: 100728363-100728363 on Build GRCh38
rs868429675 Chr.14: 100728623-100728623 on Build GRCh38
rs550567077 Chr.14: 100728441-100728441 on Build GRCh38
rs750303475 Chr.14: 100728450-100728450 on Build GRCh38
rs768148700 Chr.14: 100728433-100728433 on Build GRCh38
rs1006461177 Chr.14: 100728540-100728540 on Build GRCh38
rs577592121 Chr.14: 100728624-100728624 on Build GRCh38
rs746357653 Chr.14: 100728420-100728420 on Build GRCh38
rs960082985 Chr.14: 100728339-100728339 on Build GRCh38
rs904479114 Chr.14: 100728561-100728561 on Build GRCh38
rs376525727 Chr.14: 100728458-100728458 on Build GRCh38
rs779124291 Chr.14: 100728387-100728387 on Build GRCh38
rs914438545 Chr.14: 100728707-100728707 on Build GRCh38
rs563280590 Chr.14: 100728708-100728708 on Build GRCh38
rs528252372 Chr.14: 100728319-100728319 on Build GRCh38
rs767265253 Chr.14: 100728358-100728358 on Build GRCh38
rs1022670371 Chr.14: 100728618-100728618 on Build GRCh38
rs200447912 Chr.14: 100728476-100728476 on Build GRCh38
rs112878187 Chr.14: 100728628-100728629 on Build GRCh38
rs867051371 Chr.14: 100728571-100728571 on Build GRCh38
rs143382466 Chr.14: 100728440-100728440 on Build GRCh38
rs761694533 Chr.14: 100728353-100728353 on Build GRCh38
rs370957942 Chr.14: 100728497-100728497 on Build GRCh38
rs147089185 Chr.14: 100728329-100728329 on Build GRCh38
rs375822521 Chr.14: 100728477-100728477 on Build GRCh38
rs1014395777 Chr.14: 100728609-100728609 on Build GRCh38
rs10151914 Chr.14: 100728607-100728607 on Build GRCh38
rs751797755 Chr.14: 100728488-100728488 on Build GRCh38
rs374599665 Chr.14: 100728500-100728500 on Build GRCh38
rs1040433249 Chr.14: 100728730-100728730 on Build GRCh38
rs1042986093 Chr.14: 100728553-100728553 on Build GRCh38
rs953102899 Chr.14: 100728715-100728716 on Build GRCh38
rs746587134 Chr.14: 100728504-100728504 on Build GRCh38
rs557926884 Chr.14: 100728587-100728587 on Build GRCh38
rs553361176 Chr.14: 100728560-100728560 on Build GRCh38
rs1017481592 Chr.14: 100728666-100728666 on Build GRCh38
rs565890080 Chr.14: 100728318-100728318 on Build GRCh38
rs1027192660 Chr.14: 100728694-100728694 on Build GRCh38
rs757545372 Chr.14: 100728430-100728430 on Build GRCh38
rs962018042 Chr.14: 100728567-100728567 on Build GRCh38
rs576995874 Chr.14: 100728343-100728343 on Build GRCh38
rs13329039 Chr.14: 100728427-100728427 on Build GRCh38
rs976795354 Chr.14: 100728766-100728766 on Build GRCh38
rs778079144 Chr.14: 100728396-100728396 on Build GRCh38
rs867388893 Chr.14: 100728625-100728625 on Build GRCh38
rs760634125 Chr.14: 100728452-100728452 on Build GRCh38
rs373277301 Chr.14: 100728391-100728391 on Build GRCh38
rs947311024 Chr.14: 100728394-100728394 on Build GRCh38
rs200239189 Chr.14: 100728615-100728616 on Build GRCh38
rs142018292 Chr.14: 100728729-100728729 on Build GRCh38
rs994497580 Chr.14: 100728680-100728680 on Build GRCh38
rs537098005 Chr.14: 100728439-100728439 on Build GRCh38
rs781598740 Chr.14: 100728418-100728418 on Build GRCh38
rs944149464 Chr.14: 100728520-100728520 on Build GRCh38
rs758507505 Chr.14: 100728393-100728393 on Build GRCh38
rs999576468 Chr.14: 100728436-100728436 on Build GRCh38
rs762558665 Chr.14: 100728431-100728431 on Build GRCh38
rs545674033 Chr.14: 100728539-100728539 on Build GRCh38
rs914511408 Chr.14: 100728360-100728360 on Build GRCh38
rs567513580 Chr.14: 100728531-100728531 on Build GRCh38
rs768219462 Chr.14: 100728508-100728509 on Build GRCh38
rs113105360 Chr.14: 100728351-100728351 on Build GRCh38
rs750354698 Chr.14: 100728359-100728359 on Build GRCh38
rs542936640 Chr.14: 100728736-100728736 on Build GRCh38
rs766132033 Chr.14: 100728486-100728486 on Build GRCh38
rs921381459 Chr.14: 100728769-100728769 on Build GRCh38
rs186583984 Chr.14: 100728541-100728541 on Build GRCh38
rs866286349 Chr.14: 100728633-100728633 on Build GRCh38
rs573410025 Chr.14: 100728576-100728576 on Build GRCh38
rs756240258 Chr.14: 100728625-100728625 on Build GRCh38
rs755336735 Chr.14: 100728759-100728759 on Build GRCh38
rs965593215 Chr.14: 100728732-100728733 on Build GRCh38
rs189124625 Chr.14: 100728577-100728577 on Build GRCh38
rs370397678 Chr.14: 100728624-100728624 on Build GRCh38
rs757474355 Chr.14: 100728494-100728494 on Build GRCh38
rs774917244 Chr.14: 100728429-100728429 on Build GRCh38
rs771292634 Chr.14: 100728412-100728412 on Build GRCh38
rs901905360 Chr.14: 100728751-100728751 on Build GRCh38
rs895661649 Chr.14: 100728593-100728593 on Build GRCh38

View Full Product Details