Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104588167-104588670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf180
Entrez Gene ID
400258
Gene Name
chromosome 14 open reading frame 180
Gene Aliases
C14orf77, NRAC
UniGene
Hs.153827
Gene Chromosome Location
Chr.14: 104579684-104589847 on Build GRCh38
Transcripts
NM_001008404.2 NM_001286399.1 NM_001286400.1 XM_005267638.3 XM_011536764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971052 c.370C>T p.R124W Chr.14: 104588670-104588670 on Build GRCh38
5847647 c.277G>A p.V93M Chr.14: 104588309-104588309 on Build GRCh38
4486773 c.309C>T p.S103S Chr.14: 104588609-104588609 on Build GRCh38
5812801 c.264A>T p.T88T Chr.14: 104588296-104588296 on Build GRCh38
4147851 c.297A>G p.P99P Chr.14: 104588597-104588597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753361314 Chr.14: 104588643-104588643 on Build GRCh38
rs866690881 Chr.14: 104588357-104588357 on Build GRCh38
rs749585347 Chr.14: 104588448-104588448 on Build GRCh38
rs550307365 Chr.14: 104588385-104588385 on Build GRCh38
rs199822025 Chr.14: 104588263-104588263 on Build GRCh38
rs778147750 Chr.14: 104588229-104588229 on Build GRCh38
rs112419320 Chr.14: 104588206-104588206 on Build GRCh38
rs1031775583 Chr.14: 104588575-104588575 on Build GRCh38
rs142842253 Chr.14: 104588343-104588344 on Build GRCh38
rs751963195 Chr.14: 104588289-104588289 on Build GRCh38
rs374943787 Chr.14: 104588600-104588600 on Build GRCh38
rs767818995 Chr.14: 104588366-104588366 on Build GRCh38
rs771198714 Chr.14: 104588270-104588270 on Build GRCh38
rs964355229 Chr.14: 104588344-104588344 on Build GRCh38
rs969064990 Chr.14: 104588201-104588201 on Build GRCh38
rs746140869 Chr.14: 104588350-104588350 on Build GRCh38
rs374995646 Chr.14: 104588306-104588306 on Build GRCh38
rs769814414 Chr.14: 104588247-104588247 on Build GRCh38
rs73361827 Chr.14: 104588537-104588537 on Build GRCh38
rs935221695 Chr.14: 104588207-104588208 on Build GRCh38
rs771022547 Chr.14: 104588337-104588337 on Build GRCh38
rs557967233 Chr.14: 104588207-104588207 on Build GRCh38
rs1017198270 Chr.14: 104588331-104588331 on Build GRCh38
rs774538243 Chr.14: 104588362-104588362 on Build GRCh38
rs377073295 Chr.14: 104588352-104588352 on Build GRCh38
rs1009765947 Chr.14: 104588436-104588436 on Build GRCh38
rs779751469 Chr.14: 104588292-104588292 on Build GRCh38
rs775477825 Chr.14: 104588596-104588596 on Build GRCh38
rs548394000 Chr.14: 104588238-104588238 on Build GRCh38
rs533220862 Chr.14: 104588380-104588380 on Build GRCh38
rs964309917 Chr.14: 104588480-104588480 on Build GRCh38
rs1005725832 Chr.14: 104588530-104588530 on Build GRCh38
rs888308558 Chr.14: 104588311-104588311 on Build GRCh38
rs951676018 Chr.14: 104588631-104588631 on Build GRCh38
rs373864612 Chr.14: 104588291-104588291 on Build GRCh38
rs767921027 Chr.14: 104588606-104588606 on Build GRCh38
rs570203119 Chr.14: 104588423-104588423 on Build GRCh38
rs780160691 Chr.14: 104588259-104588259 on Build GRCh38
rs922572152 Chr.14: 104588268-104588268 on Build GRCh38
rs535846589 Chr.14: 104588452-104588452 on Build GRCh38
rs370582866 Chr.14: 104588604-104588604 on Build GRCh38
rs542453612 Chr.14: 104588389-104588389 on Build GRCh38
rs755274140 Chr.14: 104588306-104588306 on Build GRCh38
rs145751328 Chr.14: 104588290-104588290 on Build GRCh38
rs555826612 Chr.14: 104588471-104588471 on Build GRCh38
rs968165204 Chr.14: 104588460-104588460 on Build GRCh38
rs753168914 Chr.14: 104588286-104588286 on Build GRCh38
rs200465379 Chr.14: 104588242-104588242 on Build GRCh38
rs747983760 Chr.14: 104588325-104588325 on Build GRCh38
rs371187611 Chr.14: 104588540-104588540 on Build GRCh38
rs764586624 Chr.14: 104588361-104588361 on Build GRCh38
rs956474996 Chr.14: 104588211-104588211 on Build GRCh38
rs111529102 Chr.14: 104588584-104588584 on Build GRCh38
rs751164090 Chr.14: 104588235-104588235 on Build GRCh38
rs753412379 Chr.14: 104588539-104588539 on Build GRCh38
rs780506543 Chr.14: 104588241-104588241 on Build GRCh38
rs767824742 Chr.14: 104588221-104588221 on Build GRCh38
rs892119241 Chr.14: 104588381-104588381 on Build GRCh38
rs12880814 Chr.14: 104588597-104588597 on Build GRCh38
rs935196367 Chr.14: 104588412-104588412 on Build GRCh38
rs547639485 Chr.14: 104588365-104588365 on Build GRCh38
rs776511627 Chr.14: 104588277-104588277 on Build GRCh38
rs774899365 Chr.14: 104588601-104588601 on Build GRCh38
rs955379648 Chr.14: 104588550-104588550 on Build GRCh38
rs372128905 Chr.14: 104588225-104588225 on Build GRCh38
rs778515973 Chr.14: 104588319-104588319 on Build GRCh38
rs759696293 Chr.14: 104588281-104588281 on Build GRCh38
rs113812438 Chr.14: 104588497-104588497 on Build GRCh38
rs906241992 Chr.14: 104588477-104588477 on Build GRCh38
rs565966135 Chr.14: 104588474-104588474 on Build GRCh38
rs892171831 Chr.14: 104588582-104588582 on Build GRCh38
rs1029654490 Chr.14: 104588547-104588547 on Build GRCh38
rs976981412 Chr.14: 104588481-104588481 on Build GRCh38
rs9788432 Chr.14: 104588372-104588372 on Build GRCh38
rs753022039 Chr.14: 104588224-104588224 on Build GRCh38
rs909844837 Chr.14: 104588639-104588639 on Build GRCh38
rs968906832 Chr.14: 104588218-104588218 on Build GRCh38
rs765686270 Chr.14: 104588284-104588284 on Build GRCh38
rs978962396 Chr.14: 104588465-104588465 on Build GRCh38
rs778944156 Chr.14: 104588573-104588573 on Build GRCh38
rs913420872 Chr.14: 104588215-104588215 on Build GRCh38
rs569849919 Chr.14: 104588338-104588338 on Build GRCh38
rs977034607 Chr.14: 104588603-104588603 on Build GRCh38
rs929952259 Chr.14: 104588488-104588488 on Build GRCh38
rs376633325 Chr.14: 104588267-104588267 on Build GRCh38
rs78675373 Chr.14: 104588294-104588294 on Build GRCh38
rs763389400 Chr.14: 104588356-104588356 on Build GRCh38
rs369221387 Chr.14: 104588260-104588260 on Build GRCh38
rs763515847 Chr.14: 104588287-104588287 on Build GRCh38
rs201205928 Chr.14: 104588538-104588538 on Build GRCh38
rs562143106 Chr.14: 104588583-104588583 on Build GRCh38
rs1046822556 Chr.14: 104588505-104588505 on Build GRCh38
rs528020736 Chr.14: 104588351-104588351 on Build GRCh38
rs755073368 Chr.14: 104588214-104588214 on Build GRCh38
rs369790934 Chr.14: 104588269-104588269 on Build GRCh38
rs983309782 Chr.14: 104588507-104588507 on Build GRCh38
rs868014005 Chr.14: 104588613-104588613 on Build GRCh38
rs534932777 Chr.14: 104588482-104588482 on Build GRCh38
rs942898163 Chr.14: 104588253-104588253 on Build GRCh38
rs377369394 Chr.14: 104588642-104588642 on Build GRCh38
rs1022043255 Chr.14: 104588440-104588440 on Build GRCh38
rs373441764 Chr.14: 104588586-104588586 on Build GRCh38
rs773770243 Chr.14: 104588626-104588626 on Build GRCh38
rs557641271 Chr.14: 104588534-104588534 on Build GRCh38
rs115845341 Chr.14: 104588587-104588587 on Build GRCh38
rs573451584 Chr.14: 104588574-104588574 on Build GRCh38
rs541764061 Chr.14: 104588634-104588634 on Build GRCh38

View Full Product Details