Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104738361-104738855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_001320424.1 NM_152328.4 NM_199165.2 XM_006720026.3 XM_011536412.1 XM_011536414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
42752 c.448C>T p.P150S Chr.14: 104738399-104738399 on Build GRCh38
6294494 c.436G>T p.V146F Chr.14: 104738387-104738387 on Build GRCh38
696956 c.435G>T p.V145V Chr.14: 104738386-104738386 on Build GRCh38
6294495 c.307G>T p.V103F Chr.14: 104738387-104738387 on Build GRCh38
5847658 c.433G>A p.V145M Chr.14: 104738384-104738384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769839836 Chr.14: 104738399-104738399 on Build GRCh38
rs572848377 Chr.14: 104738757-104738757 on Build GRCh38
rs376632596 Chr.14: 104738467-104738467 on Build GRCh38
rs936588059 Chr.14: 104738579-104738579 on Build GRCh38
rs896233403 Chr.14: 104738581-104738581 on Build GRCh38
rs537303935 Chr.14: 104738776-104738776 on Build GRCh38
rs1036012514 Chr.14: 104738607-104738607 on Build GRCh38
rs923959839 Chr.14: 104738567-104738567 on Build GRCh38
rs560035859 Chr.14: 104738595-104738595 on Build GRCh38
rs995752835 Chr.14: 104738644-104738644 on Build GRCh38
rs978089795 Chr.14: 104738528-104738528 on Build GRCh38
rs1036558117 Chr.14: 104738821-104738821 on Build GRCh38
rs532127836 Chr.14: 104738596-104738596 on Build GRCh38
rs978620121 Chr.14: 104738690-104738690 on Build GRCh38
rs575850457 Chr.14: 104738502-104738502 on Build GRCh38
rs747748633 Chr.14: 104738489-104738489 on Build GRCh38
rs775702433 Chr.14: 104738402-104738402 on Build GRCh38
rs931774474 Chr.14: 104738589-104738589 on Build GRCh38
rs574659584 Chr.14: 104738526-104738526 on Build GRCh38
rs937949003 Chr.14: 104738739-104738739 on Build GRCh38
rs745947589 Chr.14: 104738807-104738807 on Build GRCh38
rs1043531339 Chr.14: 104738546-104738546 on Build GRCh38
rs527304996 Chr.14: 104738798-104738798 on Build GRCh38
rs749462695 Chr.14: 104738781-104738781 on Build GRCh38
rs749908378 Chr.14: 104738459-104738459 on Build GRCh38
rs1043606459 Chr.14: 104738694-104738694 on Build GRCh38
rs1027156715 Chr.14: 104738470-104738470 on Build GRCh38
rs753571697 Chr.14: 104738482-104738482 on Build GRCh38
rs559055917 Chr.14: 104738496-104738496 on Build GRCh38
rs895403957 Chr.14: 104738658-104738658 on Build GRCh38
rs989442961 Chr.14: 104738774-104738774 on Build GRCh38
rs539241656 Chr.14: 104738488-104738488 on Build GRCh38
rs373372027 Chr.14: 104738782-104738782 on Build GRCh38
rs562233860 Chr.14: 104738659-104738659 on Build GRCh38
rs376923979 Chr.14: 104738381-104738381 on Build GRCh38
rs921827017 Chr.14: 104738780-104738780 on Build GRCh38
rs1025578515 Chr.14: 104738673-104738673 on Build GRCh38
rs750099880 Chr.14: 104738477-104738488 on Build GRCh38
rs373050445 Chr.14: 104738477-104738477 on Build GRCh38
rs554691903 Chr.14: 104738517-104738517 on Build GRCh38
rs905101776 Chr.14: 104738704-104738704 on Build GRCh38
rs1001854615 Chr.14: 104738524-104738524 on Build GRCh38
rs772867061 Chr.14: 104738394-104738394 on Build GRCh38
rs762971500 Chr.14: 104738419-104738419 on Build GRCh38
rs758355304 Chr.14: 104738800-104738800 on Build GRCh38
rs970825271 Chr.14: 104738675-104738675 on Build GRCh38
rs369912171 Chr.14: 104738472-104738472 on Build GRCh38
rs912603813 Chr.14: 104738640-104738640 on Build GRCh38
rs544593137 Chr.14: 104738506-104738506 on Build GRCh38
rs1004329783 Chr.14: 104738593-104738593 on Build GRCh38
rs764190265 Chr.14: 104738424-104738424 on Build GRCh38
rs996066317 Chr.14: 104738795-104738795 on Build GRCh38
rs553061311 Chr.14: 104738594-104738594 on Build GRCh38
rs373504081 Chr.14: 104738580-104738580 on Build GRCh38
rs73360648 Chr.14: 104738655-104738655 on Build GRCh38
rs201006862 Chr.14: 104738437-104738437 on Build GRCh38
rs989270439 Chr.14: 104738656-104738656 on Build GRCh38
rs199832023 Chr.14: 104738454-104738454 on Build GRCh38
rs1028513318 Chr.14: 104738598-104738598 on Build GRCh38
rs182453389 Chr.14: 104738803-104738803 on Build GRCh38
rs921859769 Chr.14: 104738583-104738583 on Build GRCh38
rs1003895802 Chr.14: 104738770-104738770 on Build GRCh38
rs957997724 Chr.14: 104738761-104738761 on Build GRCh38
rs1015736222 Chr.14: 104738779-104738779 on Build GRCh38
rs567042560 Chr.14: 104738486-104738486 on Build GRCh38
rs371971986 Chr.14: 104738480-104738480 on Build GRCh38
rs79207846 Chr.14: 104738479-104738479 on Build GRCh38
rs531351764 Chr.14: 104738696-104738696 on Build GRCh38
rs903709096 Chr.14: 104738550-104738550 on Build GRCh38
rs200253621 Chr.14: 104738446-104738446 on Build GRCh38
rs898872547 Chr.14: 104738638-104738638 on Build GRCh38
rs760582840 Chr.14: 104738395-104738395 on Build GRCh38
rs375561909 Chr.14: 104738491-104738491 on Build GRCh38

View Full Product Details