Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104740494-104740996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_001320424.1 NM_152328.4 NM_199165.2 XM_006720026.3 XM_011536412.1 XM_011536414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3494129 c.751C>T p.P251S Chr.14: 104740876-104740876 on Build GRCh38
3885700 c.750C>T p.F250F Chr.14: 104740875-104740875 on Build GRCh38
6355597 c.549C>T p.C183C Chr.14: 104740673-104740673 on Build GRCh38
3494128 c.707C>T p.S236F Chr.14: 104740702-104740702 on Build GRCh38
6595656 c.502G>A p.G168R Chr.14: 104740626-104740626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765264298 Chr.14: 104740934-104740934 on Build GRCh38
rs533280528 Chr.14: 104740733-104740733 on Build GRCh38
rs758814291 Chr.14: 104740755-104740755 on Build GRCh38
rs753244916 Chr.14: 104740857-104740857 on Build GRCh38
rs779468129 Chr.14: 104740738-104740738 on Build GRCh38
rs772909262 Chr.14: 104740674-104740674 on Build GRCh38
rs570002015 Chr.14: 104740809-104740809 on Build GRCh38
rs556944473 Chr.14: 104740614-104740614 on Build GRCh38
rs773954704 Chr.14: 104740925-104740925 on Build GRCh38
rs376218660 Chr.14: 104740929-104740929 on Build GRCh38
rs923362443 Chr.14: 104740848-104740848 on Build GRCh38
rs929603850 Chr.14: 104740762-104740762 on Build GRCh38
rs751967273 Chr.14: 104740869-104740869 on Build GRCh38
rs377019000 Chr.14: 104740779-104740779 on Build GRCh38
rs779194965 Chr.14: 104740671-104740671 on Build GRCh38
rs3803307 Chr.14: 104740797-104740797 on Build GRCh38
rs549210865 Chr.14: 104740814-104740814 on Build GRCh38
rs149832793 Chr.14: 104740654-104740654 on Build GRCh38
rs377764596 Chr.14: 104740951-104740951 on Build GRCh38
rs113106305 Chr.14: 104740600-104740600 on Build GRCh38
rs777951779 Chr.14: 104740868-104740868 on Build GRCh38
rs202160058 Chr.14: 104740908-104740908 on Build GRCh38
rs748520253 Chr.14: 104740672-104740672 on Build GRCh38
rs760756315 Chr.14: 104740830-104740830 on Build GRCh38
rs368092518 Chr.14: 104740618-104740618 on Build GRCh38
rs372990767 Chr.14: 104740824-104740824 on Build GRCh38
rs942457557 Chr.14: 104740751-104740751 on Build GRCh38
rs369145808 Chr.14: 104740945-104740945 on Build GRCh38
rs866479068 Chr.14: 104740693-104740693 on Build GRCh38
rs766327167 Chr.14: 104740732-104740732 on Build GRCh38
rs12894569 Chr.14: 104740703-104740703 on Build GRCh38
rs1006502641 Chr.14: 104740801-104740801 on Build GRCh38
rs769475665 Chr.14: 104740885-104740885 on Build GRCh38
rs747975810 Chr.14: 104740928-104740928 on Build GRCh38
rs200306980 Chr.14: 104740927-104740927 on Build GRCh38
rs746605039 Chr.14: 104740796-104740796 on Build GRCh38
rs755859136 Chr.14: 104740966-104740966 on Build GRCh38
rs762907733 Chr.14: 104740719-104740719 on Build GRCh38
rs370413393 Chr.14: 104740935-104740935 on Build GRCh38
rs202096406 Chr.14: 104740950-104740950 on Build GRCh38
rs1054399252 Chr.14: 104740901-104740901 on Build GRCh38
rs1026445085 Chr.14: 104740812-104740812 on Build GRCh38
rs114109100 Chr.14: 104740585-104740585 on Build GRCh38
rs139898050 Chr.14: 104740608-104740608 on Build GRCh38
rs200830217 Chr.14: 104740624-104740624 on Build GRCh38
rs4307875 Chr.14: 104740938-104740938 on Build GRCh38
rs375512741 Chr.14: 104740730-104740730 on Build GRCh38
rs745899389 Chr.14: 104740809-104740809 on Build GRCh38
rs1020263753 Chr.14: 104740888-104740888 on Build GRCh38
rs886708006 Chr.14: 104740612-104740612 on Build GRCh38
rs146202938 Chr.14: 104740809-104740810 on Build GRCh38
rs1017798993 Chr.14: 104740825-104740825 on Build GRCh38
rs188251428 Chr.14: 104740823-104740823 on Build GRCh38
rs935809557 Chr.14: 104740900-104740900 on Build GRCh38
rs753050565 Chr.14: 104740749-104740749 on Build GRCh38
rs765714431 Chr.14: 104740854-104740854 on Build GRCh38
rs759344791 Chr.14: 104740701-104740701 on Build GRCh38
rs759674416 Chr.14: 104740841-104740841 on Build GRCh38
rs759582869 Chr.14: 104740932-104740932 on Build GRCh38
rs201337652 Chr.14: 104740720-104740720 on Build GRCh38
rs770388989 Chr.14: 104740682-104740682 on Build GRCh38
rs776483145 Chr.14: 104740615-104740615 on Build GRCh38
rs144265214 Chr.14: 104740880-104740880 on Build GRCh38
rs144784529 Chr.14: 104740702-104740702 on Build GRCh38
rs900869804 Chr.14: 104740549-104740549 on Build GRCh38
rs887975053 Chr.14: 104740955-104740955 on Build GRCh38
rs529290625 Chr.14: 104740810-104740810 on Build GRCh38
rs768733756 Chr.14: 104740909-104740909 on Build GRCh38
rs373243171 Chr.14: 104740756-104740756 on Build GRCh38
rs962226261 Chr.14: 104740527-104740527 on Build GRCh38
rs201862216 Chr.14: 104740589-104740589 on Build GRCh38
rs767855413 Chr.14: 104740637-104740637 on Build GRCh38
rs771131106 Chr.14: 104740728-104740729 on Build GRCh38
rs1054369546 Chr.14: 104740746-104740746 on Build GRCh38
rs776755733 Chr.14: 104740820-104740821 on Build GRCh38
rs773959408 Chr.14: 104740579-104740579 on Build GRCh38
rs750139746 Chr.14: 104740954-104740954 on Build GRCh38
rs527839589 Chr.14: 104740673-104740673 on Build GRCh38
rs906949934 Chr.14: 104740946-104740946 on Build GRCh38
rs372488352 Chr.14: 104740561-104740561 on Build GRCh38
rs1024047678 Chr.14: 104740522-104740522 on Build GRCh38
rs551647159 Chr.14: 104740931-104740931 on Build GRCh38
rs750525941 Chr.14: 104740647-104740647 on Build GRCh38
rs751704623 Chr.14: 104740784-104740784 on Build GRCh38
rs371663949 Chr.14: 104740653-104740653 on Build GRCh38
rs768947885 Chr.14: 104740564-104740564 on Build GRCh38
rs772243718 Chr.14: 104740810-104740811 on Build GRCh38
rs775624587 Chr.14: 104740816-104740816 on Build GRCh38
rs764667800 Chr.14: 104740940-104740940 on Build GRCh38
rs755536909 Chr.14: 104740666-104740666 on Build GRCh38
rs1012708513 Chr.14: 104740522-104740522 on Build GRCh38
rs375018442 Chr.14: 104740681-104740681 on Build GRCh38
rs757496228 Chr.14: 104740633-104740633 on Build GRCh38
rs112013617 Chr.14: 104740598-104740598 on Build GRCh38
rs567381063 Chr.14: 104740525-104740525 on Build GRCh38
rs752562610 Chr.14: 104740625-104740625 on Build GRCh38
rs200895321 Chr.14: 104740834-104740834 on Build GRCh38
rs1036146804 Chr.14: 104740866-104740866 on Build GRCh38
rs369995687 Chr.14: 104740699-104740699 on Build GRCh38
rs544157025 Chr.14: 104740650-104740650 on Build GRCh38
rs200192645 Chr.14: 104740665-104740665 on Build GRCh38
rs758936863 Chr.14: 104740865-104740865 on Build GRCh38
rs756794512 Chr.14: 104740877-104740877 on Build GRCh38
rs35614288 Chr.14: 104740783-104740783 on Build GRCh38
rs749479977 Chr.14: 104740676-104740678 on Build GRCh38

View Full Product Details