Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30634116-30634655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11B
Entrez Gene ID
89839
Gene Name
Rho GTPase activating protein 11B
Gene Aliases
B'-T, FAM7B1
UniGene
Hs.659621
Gene Chromosome Location
Chr.15: 30626676-30638810 on Build GRCh38
Transcripts
NM_001039841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578159 c.452A>G p.E151G Chr.15: 30634324-30634324 on Build GRCh38
4939368 c.454A>G p.K152E Chr.15: 30634326-30634326 on Build GRCh38
6603030 c.475C>A p.L159I Chr.15: 30634347-30634347 on Build GRCh38
268689 c.353C>T p.A118V Chr.15: 30634225-30634225 on Build GRCh38
960341 c.422T>C p.L141P Chr.15: 30634294-30634294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746073192 Chr.15: 30634322-30634322 on Build GRCh38
rs761150663 Chr.15: 30634352-30634352 on Build GRCh38
rs926154433 Chr.15: 30634223-30634223 on Build GRCh38
rs772402848 Chr.15: 30634323-30634323 on Build GRCh38
rs527373636 Chr.15: 30634591-30634591 on Build GRCh38
rs757851646 Chr.15: 30634261-30634261 on Build GRCh38
rs777716736 Chr.15: 30634297-30634297 on Build GRCh38
rs773138351 Chr.15: 30634196-30634196 on Build GRCh38
rs576610011 Chr.15: 30634465-30634465 on Build GRCh38
rs769839518 Chr.15: 30634185-30634185 on Build GRCh38
rs1037909636 Chr.15: 30634421-30634421 on Build GRCh38
rs774587817 Chr.15: 30634318-30634318 on Build GRCh38
rs764384036 Chr.15: 30634364-30634364 on Build GRCh38
rs916349482 Chr.15: 30634272-30634272 on Build GRCh38
rs775685313 Chr.15: 30634341-30634341 on Build GRCh38
rs745983464 Chr.15: 30634458-30634458 on Build GRCh38
rs565040066 Chr.15: 30634500-30634500 on Build GRCh38
rs751142575 Chr.15: 30634262-30634262 on Build GRCh38
rs926530576 Chr.15: 30634386-30634386 on Build GRCh38
rs560873162 Chr.15: 30634597-30634597 on Build GRCh38
rs755899255 Chr.15: 30634393-30634393 on Build GRCh38
rs749431787 Chr.15: 30634198-30634198 on Build GRCh38
rs988876051 Chr.15: 30634310-30634310 on Build GRCh38
rs1008975440 Chr.15: 30634526-30634526 on Build GRCh38
rs751765451 Chr.15: 30634223-30634224 on Build GRCh38
rs968996118 Chr.15: 30634283-30634283 on Build GRCh38
rs775979080 Chr.15: 30634240-30634240 on Build GRCh38
rs767551108 Chr.15: 30634303-30634304 on Build GRCh38
rs956097981 Chr.15: 30634285-30634285 on Build GRCh38
rs779024152 Chr.15: 30634450-30634452 on Build GRCh38
rs767252391 Chr.15: 30634388-30634388 on Build GRCh38
rs563006128 Chr.15: 30634199-30634199 on Build GRCh38
rs757324376 Chr.15: 30634430-30634430 on Build GRCh38
rs965316773 Chr.15: 30634563-30634563 on Build GRCh38
rs765814821 Chr.15: 30634379-30634379 on Build GRCh38
rs764701226 Chr.15: 30634248-30634248 on Build GRCh38
rs749879540 Chr.15: 30634255-30634255 on Build GRCh38
rs983795818 Chr.15: 30634266-30634266 on Build GRCh38
rs915040224 Chr.15: 30634417-30634417 on Build GRCh38
rs774675599 Chr.15: 30634224-30634224 on Build GRCh38
rs1033119170 Chr.15: 30634578-30634578 on Build GRCh38
rs777078462 Chr.15: 30634371-30634371 on Build GRCh38
rs753772842 Chr.15: 30634418-30634418 on Build GRCh38
rs1021722695 Chr.15: 30634172-30634172 on Build GRCh38
rs965027345 Chr.15: 30634180-30634180 on Build GRCh38
rs780847779 Chr.15: 30634276-30634276 on Build GRCh38
rs982211973 Chr.15: 30634306-30634306 on Build GRCh38
rs755219465 Chr.15: 30634235-30634236 on Build GRCh38
rs374158012 Chr.15: 30634453-30634453 on Build GRCh38
rs780274066 Chr.15: 30634464-30634464 on Build GRCh38
rs751138607 Chr.15: 30634380-30634380 on Build GRCh38
rs1006908725 Chr.15: 30634221-30634221 on Build GRCh38
rs776990404 Chr.15: 30634475-30634475 on Build GRCh38
rs1041483484 Chr.15: 30634518-30634518 on Build GRCh38
rs766705502 Chr.15: 30634213-30634215 on Build GRCh38
rs749269796 Chr.15: 30634305-30634305 on Build GRCh38
rs748025690 Chr.15: 30634181-30634181 on Build GRCh38
rs754725884 Chr.15: 30634165-30634165 on Build GRCh38
rs752835434 Chr.15: 30634391-30634393 on Build GRCh38
rs771068533 Chr.15: 30634314-30634314 on Build GRCh38
rs755987238 Chr.15: 30634296-30634296 on Build GRCh38
rs545251678 Chr.15: 30634494-30634494 on Build GRCh38
rs927882934 Chr.15: 30634339-30634339 on Build GRCh38
rs768996065 Chr.15: 30634472-30634472 on Build GRCh38
rs952858653 Chr.15: 30634599-30634599 on Build GRCh38
rs1047519766 Chr.15: 30634462-30634462 on Build GRCh38
rs756195105 Chr.15: 30634441-30634442 on Build GRCh38
rs781042733 Chr.15: 30634178-30634178 on Build GRCh38
rs759843396 Chr.15: 30634225-30634225 on Build GRCh38
rs371509806 Chr.15: 30634377-30634377 on Build GRCh38
rs1574382 Chr.15: 30634565-30634565 on Build GRCh38
rs1000467266 Chr.15: 30634573-30634573 on Build GRCh38
rs748018819 Chr.15: 30634290-30634290 on Build GRCh38
rs889903473 Chr.15: 30634491-30634491 on Build GRCh38
rs752319675 Chr.15: 30634391-30634391 on Build GRCh38
rs777435170 Chr.15: 30634411-30634411 on Build GRCh38
rs1017292982 Chr.15: 30634529-30634529 on Build GRCh38
rs761080096 Chr.15: 30634243-30634243 on Build GRCh38
rs547475867 Chr.15: 30634595-30634595 on Build GRCh38
rs993206743 Chr.15: 30634456-30634456 on Build GRCh38
rs772161108 Chr.15: 30634461-30634461 on Build GRCh38
rs376891539 Chr.15: 30634227-30634227 on Build GRCh38
rs746682624 Chr.15: 30634157-30634157 on Build GRCh38
rs952889751 Chr.15: 30634263-30634263 on Build GRCh38
rs887045914 Chr.15: 30634257-30634271 on Build GRCh38
rs754636191 Chr.15: 30634268-30634268 on Build GRCh38
rs901415722 Chr.15: 30634531-30634531 on Build GRCh38

View Full Product Details