Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32636576-32637080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11A
Entrez Gene ID
9824
Gene Name
Rho GTPase activating protein 11A
Gene Aliases
GAP (1-12)
UniGene
Hs.591130
Gene Chromosome Location
Chr.15: 32615144-32639949 on Build GRCh38
Transcripts
NM_001286479.1 NM_001286480.1 NM_014783.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6603024 c.2247G>A p.P749P Chr.15: 32637020-32637020 on Build GRCh38
960464 c.2060C>T p.T687I Chr.15: 32636833-32636833 on Build GRCh38
960465 c.2151C>A p.F717L Chr.15: 32636924-32636924 on Build GRCh38
4417573 c.2191G>T p.D731Y Chr.15: 32636964-32636964 on Build GRCh38
4441443 c.1805C>T p.A602V Chr.15: 32636578-32636578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555394373 Chr.15: 32637025-32637025 on Build GRCh38
rs928432739 Chr.15: 32637044-32637044 on Build GRCh38
rs61758695 Chr.15: 32636652-32636652 on Build GRCh38
rs770071785 Chr.15: 32637033-32637033 on Build GRCh38
rs377731590 Chr.15: 32636874-32636874 on Build GRCh38
rs768376712 Chr.15: 32636942-32636945 on Build GRCh38
rs1047865002 Chr.15: 32636782-32636782 on Build GRCh38
rs111425460 Chr.15: 32636903-32636903 on Build GRCh38
rs776380445 Chr.15: 32637054-32637056 on Build GRCh38
rs1011442846 Chr.15: 32636882-32636882 on Build GRCh38
rs766787717 Chr.15: 32636916-32636916 on Build GRCh38
rs780749897 Chr.15: 32636651-32636651 on Build GRCh38
rs374275026 Chr.15: 32637055-32637055 on Build GRCh38
rs749141290 Chr.15: 32636808-32636808 on Build GRCh38
rs1019715198 Chr.15: 32636918-32636918 on Build GRCh38
rs771829860 Chr.15: 32636632-32636633 on Build GRCh38
rs763447291 Chr.15: 32636914-32636914 on Build GRCh38
rs1045487941 Chr.15: 32636702-32636702 on Build GRCh38
rs747141868 Chr.15: 32637018-32637018 on Build GRCh38
rs752293185 Chr.15: 32636794-32636794 on Build GRCh38
rs991063085 Chr.15: 32636991-32636991 on Build GRCh38
rs746725915 Chr.15: 32636903-32636906 on Build GRCh38
rs561697586 Chr.15: 32636724-32636724 on Build GRCh38
rs763646762 Chr.15: 32637049-32637049 on Build GRCh38
rs766956038 Chr.15: 32637045-32637045 on Build GRCh38
rs573125613 Chr.15: 32637024-32637024 on Build GRCh38
rs753500032 Chr.15: 32636946-32636946 on Build GRCh38
rs202105243 Chr.15: 32636700-32636700 on Build GRCh38
rs757085835 Chr.15: 32636822-32636822 on Build GRCh38
rs774965589 Chr.15: 32637047-32637047 on Build GRCh38
rs565643568 Chr.15: 32636801-32636801 on Build GRCh38
rs536928735 Chr.15: 32637020-32637020 on Build GRCh38
rs138279948 Chr.15: 32636998-32636998 on Build GRCh38
rs778745127 Chr.15: 32636824-32636824 on Build GRCh38
rs779388959 Chr.15: 32636612-32636612 on Build GRCh38
rs775574246 Chr.15: 32636877-32636877 on Build GRCh38
rs199884674 Chr.15: 32636607-32636607 on Build GRCh38
rs189559276 Chr.15: 32637043-32637043 on Build GRCh38
rs773617500 Chr.15: 32636913-32636913 on Build GRCh38
rs201555862 Chr.15: 32636797-32636797 on Build GRCh38
rs370856022 Chr.15: 32636878-32636878 on Build GRCh38
rs1028144966 Chr.15: 32636889-32636889 on Build GRCh38
rs753557646 Chr.15: 32637053-32637053 on Build GRCh38
rs764059949 Chr.15: 32636897-32636897 on Build GRCh38
rs200377132 Chr.15: 32636934-32636934 on Build GRCh38
rs545619705 Chr.15: 32636647-32636647 on Build GRCh38
rs952980251 Chr.15: 32636988-32636988 on Build GRCh38
rs769722243 Chr.15: 32636656-32636656 on Build GRCh38
rs757140912 Chr.15: 32636964-32636964 on Build GRCh38
rs1052705158 Chr.15: 32636800-32636800 on Build GRCh38
rs375838141 Chr.15: 32636976-32636976 on Build GRCh38
rs528898505 Chr.15: 32636785-32636785 on Build GRCh38
rs772151113 Chr.15: 32636718-32636718 on Build GRCh38
rs764447695 Chr.15: 32636745-32636745 on Build GRCh38
rs765600768 Chr.15: 32636908-32636908 on Build GRCh38
rs768693478 Chr.15: 32637019-32637019 on Build GRCh38
rs373354756 Chr.15: 32636663-32636663 on Build GRCh38
rs531220822 Chr.15: 32636688-32636688 on Build GRCh38
rs761448800 Chr.15: 32637056-32637057 on Build GRCh38
rs970930708 Chr.15: 32636763-32636763 on Build GRCh38
rs771047764 Chr.15: 32636667-32636667 on Build GRCh38
rs776814252 Chr.15: 32636890-32636890 on Build GRCh38
rs775597191 Chr.15: 32636720-32636720 on Build GRCh38
rs530913844 Chr.15: 32636880-32636880 on Build GRCh38
rs961733199 Chr.15: 32636919-32636919 on Build GRCh38
rs776718156 Chr.15: 32636606-32636606 on Build GRCh38
rs367982621 Chr.15: 32636993-32636993 on Build GRCh38
rs1033200713 Chr.15: 32636654-32636654 on Build GRCh38
rs914090958 Chr.15: 32636725-32636725 on Build GRCh38
rs751092235 Chr.15: 32636632-32636632 on Build GRCh38
rs775359909 Chr.15: 32636897-32636897 on Build GRCh38
rs893077627 Chr.15: 32636850-32636850 on Build GRCh38
rs765716119 Chr.15: 32636775-32636775 on Build GRCh38
rs754481526 Chr.15: 32636786-32636786 on Build GRCh38
rs147044309 Chr.15: 32636970-32636970 on Build GRCh38
rs370589918 Chr.15: 32637048-32637048 on Build GRCh38
rs773689327 Chr.15: 32637041-32637041 on Build GRCh38
rs988279997 Chr.15: 32636694-32636694 on Build GRCh38
rs774321509 Chr.15: 32636686-32636686 on Build GRCh38
rs767007185 Chr.15: 32636790-32636790 on Build GRCh38
rs752311090 Chr.15: 32636927-32636927 on Build GRCh38
rs906922118 Chr.15: 32636752-32636752 on Build GRCh38
rs754269816 Chr.15: 32636769-32636769 on Build GRCh38
rs745768456 Chr.15: 32636846-32636846 on Build GRCh38
rs755660320 Chr.15: 32636932-32636932 on Build GRCh38
rs199734120 Chr.15: 32636608-32636608 on Build GRCh38

View Full Product Details