Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32766539-32767048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN1
Entrez Gene ID
342184
Gene Name
formin 1
Gene Aliases
FMN, LD
UniGene
Hs.657649
Gene Chromosome Location
Chr.15: 32765544-33194765 on Build GRCh38
Transcripts
NM_001103184.3 NM_001277313.1 XM_011521504.2 XM_011521506.2 XM_011521509.2 XM_011521511.2 XM_017022130.1 XM_017022132.1 XM_017022133.1 XM_017022134.1 XM_017022135.1

SNP Detail

SNP ID Chromosome Location
rs536403671 Chr.15: 32766977-32766977 on Build GRCh38
rs1045484682 Chr.15: 32766806-32766806 on Build GRCh38
rs1019070964 Chr.15: 32766889-32766889 on Build GRCh38
rs988339860 Chr.15: 32766664-32766664 on Build GRCh38
rs563907630 Chr.15: 32766922-32766922 on Build GRCh38
rs549024062 Chr.15: 32766861-32766861 on Build GRCh38
rs751560953 Chr.15: 32766979-32766979 on Build GRCh38
rs573208604 Chr.15: 32766598-32766598 on Build GRCh38
rs773394600 Chr.15: 32766817-32766817 on Build GRCh38
rs1005889829 Chr.15: 32766839-32766839 on Build GRCh38
rs3812932 Chr.15: 32766709-32766709 on Build GRCh38
rs930786761 Chr.15: 32767015-32767015 on Build GRCh38
rs3812927 Chr.15: 32767024-32767024 on Build GRCh38
rs1046919373 Chr.15: 32766776-32766776 on Build GRCh38
rs1049365908 Chr.15: 32767022-32767022 on Build GRCh38
rs1038677157 Chr.15: 32766967-32766967 on Build GRCh38
rs374159584 Chr.15: 32766960-32766960 on Build GRCh38
rs557170073 Chr.15: 32766943-32766943 on Build GRCh38
rs906928527 Chr.15: 32766811-32766811 on Build GRCh38
rs571653534 Chr.15: 32766873-32766873 on Build GRCh38
rs747053065 Chr.15: 32766865-32766865 on Build GRCh38
rs923354132 Chr.15: 32766924-32766924 on Build GRCh38
rs2204722 Chr.15: 32766762-32766762 on Build GRCh38
rs941265022 Chr.15: 32766963-32766963 on Build GRCh38
rs370758784 Chr.15: 32766754-32766754 on Build GRCh38
rs528418058 Chr.15: 32766860-32766860 on Build GRCh38
rs937668660 Chr.15: 32766987-32766987 on Build GRCh38
rs960481223 Chr.15: 32766869-32766870 on Build GRCh38
rs540587472 Chr.15: 32766621-32766621 on Build GRCh38
rs895403311 Chr.15: 32766842-32766842 on Build GRCh38
rs966519885 Chr.15: 32766891-32766891 on Build GRCh38
rs937052810 Chr.15: 32766981-32766981 on Build GRCh38
rs989994690 Chr.15: 32766928-32766928 on Build GRCh38
rs1055316945 Chr.15: 32767004-32767004 on Build GRCh38
rs865931532 Chr.15: 32766674-32766674 on Build GRCh38
rs957301978 Chr.15: 32766914-32766914 on Build GRCh38
rs1055692822 Chr.15: 32766823-32766823 on Build GRCh38
rs994872565 Chr.15: 32766572-32766572 on Build GRCh38
rs968290677 Chr.15: 32766669-32766670 on Build GRCh38
rs115147687 Chr.15: 32766640-32766640 on Build GRCh38
rs3812928 Chr.15: 32766951-32766951 on Build GRCh38
rs554794991 Chr.15: 32766569-32766569 on Build GRCh38
rs752614571 Chr.15: 32766737-32766737 on Build GRCh38
rs916706847 Chr.15: 32767006-32767006 on Build GRCh38
rs982545010 Chr.15: 32766948-32766948 on Build GRCh38
rs765155566 Chr.15: 32766677-32766677 on Build GRCh38
rs16958537 Chr.15: 32766665-32766665 on Build GRCh38
rs28595770 Chr.15: 32766656-32766656 on Build GRCh38
rs935885168 Chr.15: 32766714-32766714 on Build GRCh38
rs886469028 Chr.15: 32766863-32766863 on Build GRCh38
rs186082678 Chr.15: 32766787-32766787 on Build GRCh38
rs3812929 Chr.15: 32766862-32766862 on Build GRCh38
rs554756241 Chr.15: 32766984-32766984 on Build GRCh38
rs996034050 Chr.15: 32766895-32766895 on Build GRCh38
rs976142532 Chr.15: 32766921-32766921 on Build GRCh38
rs527681123 Chr.15: 32766852-32766852 on Build GRCh38
rs912873984 Chr.15: 32766721-32766721 on Build GRCh38
rs3812931 Chr.15: 32766720-32766720 on Build GRCh38
rs965059063 Chr.15: 32766917-32766918 on Build GRCh38
rs913135611 Chr.15: 32766941-32766941 on Build GRCh38
rs968298560 Chr.15: 32766898-32766898 on Build GRCh38
rs73378952 Chr.15: 32766905-32766905 on Build GRCh38
rs969132333 Chr.15: 32766569-32766573 on Build GRCh38
rs950797831 Chr.15: 32766623-32766623 on Build GRCh38

View Full Product Details