Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34147108-34147517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNBL1
Entrez Gene ID
79768
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29
UniGene
Hs.633566
Gene Chromosome Location
Chr.15: 34140674-34210146 on Build GRCh38
Transcripts
NM_024713.2 XM_005254668.3 XM_017022571.1 XM_017022572.1 XM_017022573.1 XM_017022574.1 XM_017022575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245791 c.615G>T p.Q205H Chr.15: 34147283-34147283 on Build GRCh38
4148661 c.667G>T p.V223L Chr.15: 34147231-34147231 on Build GRCh38
6690293 c.665T>C p.L222P Chr.15: 34147233-34147233 on Build GRCh38
1659293 c.680T>A p.L227H Chr.15: 34147218-34147218 on Build GRCh38
6865100 c.680T>G p.L227R Chr.15: 34147218-34147218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770241291 Chr.15: 34147346-34147346 on Build GRCh38
rs201403507 Chr.15: 34147487-34147487 on Build GRCh38
rs547225796 Chr.15: 34147134-34147134 on Build GRCh38
rs767011372 Chr.15: 34147202-34147202 on Build GRCh38
rs756890707 Chr.15: 34147457-34147457 on Build GRCh38
rs773912306 Chr.15: 34147347-34147347 on Build GRCh38
rs935801562 Chr.15: 34147443-34147443 on Build GRCh38
rs35167124 Chr.15: 34147161-34147162 on Build GRCh38
rs538528685 Chr.15: 34147165-34147165 on Build GRCh38
rs573863174 Chr.15: 34147192-34147192 on Build GRCh38
rs780394428 Chr.15: 34147463-34147463 on Build GRCh38
rs772347327 Chr.15: 34147302-34147302 on Build GRCh38
rs962567745 Chr.15: 34147492-34147492 on Build GRCh38
rs756015153 Chr.15: 34147352-34147352 on Build GRCh38
rs779156933 Chr.15: 34147265-34147265 on Build GRCh38
rs139830158 Chr.15: 34147209-34147209 on Build GRCh38
rs372407960 Chr.15: 34147306-34147306 on Build GRCh38
rs962864511 Chr.15: 34147174-34147174 on Build GRCh38
rs536283658 Chr.15: 34147407-34147407 on Build GRCh38
rs769007847 Chr.15: 34147325-34147325 on Build GRCh38
rs916106090 Chr.15: 34147465-34147465 on Build GRCh38
rs146813634 Chr.15: 34147332-34147332 on Build GRCh38
rs556864214 Chr.15: 34147206-34147206 on Build GRCh38
rs79165488 Chr.15: 34147323-34147323 on Build GRCh38
rs777444047 Chr.15: 34147352-34147354 on Build GRCh38
rs368534152 Chr.15: 34147176-34147176 on Build GRCh38
rs1028853983 Chr.15: 34147244-34147244 on Build GRCh38
rs750131336 Chr.15: 34147338-34147338 on Build GRCh38
rs765021681 Chr.15: 34147432-34147432 on Build GRCh38
rs765849647 Chr.15: 34147181-34147181 on Build GRCh38
rs369718128 Chr.15: 34147414-34147414 on Build GRCh38
rs887127143 Chr.15: 34147135-34147135 on Build GRCh38
rs953315112 Chr.15: 34147308-34147308 on Build GRCh38
rs202128690 Chr.15: 34147488-34147488 on Build GRCh38
rs749063148 Chr.15: 34147351-34147352 on Build GRCh38
rs755141963 Chr.15: 34147480-34147480 on Build GRCh38
rs372467198 Chr.15: 34147449-34147449 on Build GRCh38
rs760585192 Chr.15: 34147405-34147405 on Build GRCh38
rs139279812 Chr.15: 34147454-34147454 on Build GRCh38
rs780948214 Chr.15: 34147155-34147155 on Build GRCh38
rs750981421 Chr.15: 34147188-34147188 on Build GRCh38
rs755959023 Chr.15: 34147249-34147249 on Build GRCh38
rs747173629 Chr.15: 34147478-34147478 on Build GRCh38
rs750533927 Chr.15: 34147441-34147441 on Build GRCh38
rs967026502 Chr.15: 34147371-34147371 on Build GRCh38
rs767258189 Chr.15: 34147382-34147382 on Build GRCh38
rs758910903 Chr.15: 34147370-34147370 on Build GRCh38
rs757292160 Chr.15: 34147429-34147429 on Build GRCh38
rs774999787 Chr.15: 34147384-34147384 on Build GRCh38
rs571173722 Chr.15: 34147142-34147142 on Build GRCh38
rs777921653 Chr.15: 34147253-34147253 on Build GRCh38
rs777168245 Chr.15: 34147330-34147330 on Build GRCh38
rs745896586 Chr.15: 34147278-34147278 on Build GRCh38
rs530124944 Chr.15: 34147473-34147473 on Build GRCh38
rs769167528 Chr.15: 34147149-34147149 on Build GRCh38
rs757463531 Chr.15: 34147258-34147258 on Build GRCh38
rs760885172 Chr.15: 34147147-34147147 on Build GRCh38

View Full Product Details