Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38699211-38699716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf53
Entrez Gene ID
400359
Gene Name
chromosome 15 open reading frame 53
Gene Aliases
-
UniGene
Hs.448785
Gene Chromosome Location
Chr.15: 38696598-38700038 on Build GRCh38
Transcripts
NM_207444.2

SNP Detail

SNP ID Chromosome Location
rs907290781 Chr.15: 38699584-38699584 on Build GRCh38
rs915880565 Chr.15: 38699527-38699527 on Build GRCh38
rs113538057 Chr.15: 38699582-38699582 on Build GRCh38
rs1495187 Chr.15: 38699506-38699506 on Build GRCh38
rs762088197 Chr.15: 38699307-38699307 on Build GRCh38
rs866398201 Chr.15: 38699533-38699533 on Build GRCh38
rs74589865 Chr.15: 38699682-38699682 on Build GRCh38
rs138429733 Chr.15: 38699438-38699438 on Build GRCh38
rs1045889517 Chr.15: 38699397-38699401 on Build GRCh38
rs574012032 Chr.15: 38699608-38699608 on Build GRCh38
rs1495185 Chr.15: 38699235-38699235 on Build GRCh38
rs1001545384 Chr.15: 38699404-38699404 on Build GRCh38
rs1026354410 Chr.15: 38699672-38699672 on Build GRCh38
rs183670690 Chr.15: 38699649-38699649 on Build GRCh38
rs555539309 Chr.15: 38699280-38699280 on Build GRCh38
rs193017735 Chr.15: 38699414-38699414 on Build GRCh38
rs1038667437 Chr.15: 38699299-38699299 on Build GRCh38
rs191420719 Chr.15: 38699648-38699648 on Build GRCh38
rs766410068 Chr.15: 38699493-38699493 on Build GRCh38
rs913146371 Chr.15: 38699578-38699578 on Build GRCh38
rs576636762 Chr.15: 38699278-38699278 on Build GRCh38
rs1004345062 Chr.15: 38699358-38699358 on Build GRCh38
rs1034313501 Chr.15: 38699406-38699406 on Build GRCh38
rs924699284 Chr.15: 38699254-38699254 on Build GRCh38
rs547251238 Chr.15: 38699494-38699494 on Build GRCh38
rs531693829 Chr.15: 38699563-38699563 on Build GRCh38
rs889423036 Chr.15: 38699678-38699678 on Build GRCh38
rs974381745 Chr.15: 38699504-38699504 on Build GRCh38
rs769471650 Chr.15: 38699682-38699683 on Build GRCh38
rs573926866 Chr.15: 38699289-38699289 on Build GRCh38
rs987101165 Chr.15: 38699424-38699424 on Build GRCh38
rs990280498 Chr.15: 38699524-38699524 on Build GRCh38
rs878906628 Chr.15: 38699569-38699569 on Build GRCh38
rs754818272 Chr.15: 38699577-38699577 on Build GRCh38
rs140784970 Chr.15: 38699393-38699393 on Build GRCh38
rs186584483 Chr.15: 38699587-38699587 on Build GRCh38
rs12916379 Chr.15: 38699319-38699319 on Build GRCh38
rs150123050 Chr.15: 38699402-38699402 on Build GRCh38
rs907351865 Chr.15: 38699345-38699345 on Build GRCh38
rs898768629 Chr.15: 38699412-38699412 on Build GRCh38
rs563539576 Chr.15: 38699389-38699389 on Build GRCh38
rs145245220 Chr.15: 38699250-38699250 on Build GRCh38
rs202156900 Chr.15: 38699449-38699449 on Build GRCh38
rs937387866 Chr.15: 38699607-38699607 on Build GRCh38
rs948761922 Chr.15: 38699536-38699536 on Build GRCh38
rs78750057 Chr.15: 38699676-38699676 on Build GRCh38
rs118067070 Chr.15: 38699679-38699679 on Build GRCh38
rs185880376 Chr.15: 38699583-38699583 on Build GRCh38
rs534679364 Chr.15: 38699330-38699330 on Build GRCh38
rs1495186 Chr.15: 38699255-38699255 on Build GRCh38
rs188668611 Chr.15: 38699378-38699378 on Build GRCh38
rs966616085 Chr.15: 38699692-38699692 on Build GRCh38
rs944413601 Chr.15: 38699294-38699294 on Build GRCh38
rs898516669 Chr.15: 38699645-38699645 on Build GRCh38
rs75046697 Chr.15: 38699683-38699683 on Build GRCh38
rs555676244 Chr.15: 38699588-38699588 on Build GRCh38
rs559767064 Chr.15: 38699324-38699324 on Build GRCh38
rs1041797436 Chr.15: 38699253-38699253 on Build GRCh38

View Full Product Details