Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44427671-44428178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL2
Entrez Gene ID
51496
Gene Name
CTD small phosphatase like 2
Gene Aliases
HSPC058, HSPC129
UniGene
Hs.497967
Gene Chromosome Location
Chr.15: 44427329-44527774 on Build GRCh38
Transcripts
NM_016396.2 XM_005254441.2 XM_006720565.2 XM_011521665.2 XM_017022306.1 XM_017022307.1

SNP Detail

SNP ID Chromosome Location
rs1054085552 Chr.15: 44427996-44427996 on Build GRCh38
rs1046131980 Chr.15: 44427958-44427958 on Build GRCh38
rs3743219 Chr.15: 44427835-44427835 on Build GRCh38
rs116908763 Chr.15: 44427813-44427813 on Build GRCh38
rs907677389 Chr.15: 44427966-44427966 on Build GRCh38
rs999974708 Chr.15: 44427718-44427718 on Build GRCh38
rs1050834398 Chr.15: 44428062-44428062 on Build GRCh38
rs1033164852 Chr.15: 44427791-44427791 on Build GRCh38
rs1046184595 Chr.15: 44427725-44427725 on Build GRCh38
rs1004715121 Chr.15: 44427769-44427770 on Build GRCh38
rs145528616 Chr.15: 44427700-44427700 on Build GRCh38
rs936367612 Chr.15: 44427704-44427704 on Build GRCh38
rs984927465 Chr.15: 44427855-44427855 on Build GRCh38
rs1054878788 Chr.15: 44427709-44427709 on Build GRCh38
rs917059922 Chr.15: 44427918-44427918 on Build GRCh38
rs949968103 Chr.15: 44427957-44427957 on Build GRCh38
rs996413464 Chr.15: 44428078-44428078 on Build GRCh38
rs57843471 Chr.15: 44427747-44427747 on Build GRCh38
rs561657545 Chr.15: 44427849-44427849 on Build GRCh38
rs771401492 Chr.15: 44427872-44427872 on Build GRCh38
rs905559252 Chr.15: 44427988-44427988 on Build GRCh38
rs1029192195 Chr.15: 44428088-44428088 on Build GRCh38
rs1028973038 Chr.15: 44427836-44427836 on Build GRCh38
rs375314623 Chr.15: 44428147-44428147 on Build GRCh38
rs940570226 Chr.15: 44427980-44427980 on Build GRCh38
rs764041355 Chr.15: 44428056-44428056 on Build GRCh38
rs544050020 Chr.15: 44428139-44428139 on Build GRCh38
rs774898559 Chr.15: 44427993-44427993 on Build GRCh38
rs972570184 Chr.15: 44427910-44427910 on Build GRCh38
rs1020425901 Chr.15: 44427816-44427818 on Build GRCh38
rs940747371 Chr.15: 44427900-44427900 on Build GRCh38
rs148856339 Chr.15: 44427716-44427716 on Build GRCh38
rs375866378 Chr.15: 44427714-44427714 on Build GRCh38
rs886289818 Chr.15: 44427757-44427757 on Build GRCh38
rs899164577 Chr.15: 44427777-44427777 on Build GRCh38
rs949232354 Chr.15: 44427715-44427715 on Build GRCh38
rs181653074 Chr.15: 44427805-44427805 on Build GRCh38
rs891545910 Chr.15: 44428042-44428042 on Build GRCh38
rs539341080 Chr.15: 44428060-44428060 on Build GRCh38
rs1044321365 Chr.15: 44427986-44427986 on Build GRCh38
rs990919678 Chr.15: 44427874-44427874 on Build GRCh38
rs3743218 Chr.15: 44427734-44427734 on Build GRCh38
rs957708569 Chr.15: 44427851-44427851 on Build GRCh38
rs562522957 Chr.15: 44427778-44427778 on Build GRCh38
rs1020414395 Chr.15: 44428117-44428118 on Build GRCh38
rs1037607908 Chr.15: 44427990-44427990 on Build GRCh38
rs956269157 Chr.15: 44427798-44427798 on Build GRCh38
rs74658224 Chr.15: 44427809-44427809 on Build GRCh38
rs143480180 Chr.15: 44427736-44427736 on Build GRCh38
rs959182242 Chr.15: 44428137-44428137 on Build GRCh38
rs949114875 Chr.15: 44427913-44427913 on Build GRCh38
rs967547676 Chr.15: 44427829-44427829 on Build GRCh38
rs567135949 Chr.15: 44428052-44428052 on Build GRCh38
rs991771247 Chr.15: 44428143-44428143 on Build GRCh38
rs890465128 Chr.15: 44428096-44428096 on Build GRCh38
rs766420865 Chr.15: 44427819-44427820 on Build GRCh38
rs747705648 Chr.15: 44427780-44427780 on Build GRCh38
rs916260183 Chr.15: 44427878-44427878 on Build GRCh38
rs116317537 Chr.15: 44428131-44428131 on Build GRCh38
rs775537531 Chr.15: 44427843-44427843 on Build GRCh38
rs1009957816 Chr.15: 44428051-44428051 on Build GRCh38
rs926238100 Chr.15: 44427844-44427844 on Build GRCh38
rs147982201 Chr.15: 44427823-44427823 on Build GRCh38
rs759639448 Chr.15: 44428057-44428057 on Build GRCh38
rs547252683 Chr.15: 44428044-44428044 on Build GRCh38
rs141771355 Chr.15: 44428118-44428118 on Build GRCh38
rs1008953107 Chr.15: 44428117-44428117 on Build GRCh38
rs759618509 Chr.15: 44427854-44427854 on Build GRCh38
rs1000389782 Chr.15: 44428118-44428118 on Build GRCh38
rs1037618641 Chr.15: 44427770-44427770 on Build GRCh38
rs996605046 Chr.15: 44427792-44427792 on Build GRCh38

View Full Product Details