Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45120012-45120523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOXA1
Entrez Gene ID
90527
Gene Name
dual oxidase maturation factor 1
Gene Aliases
NIP, NUMBIP, mol
UniGene
Hs.356664
Gene Chromosome Location
Chr.15: 45117366-45129938 on Build GRCh38
Transcripts
NM_001276264.1 NM_001276265.1 NM_001276266.1 NM_001276267.1 NM_001276268.1 NM_144565.3 XM_006720743.3 XM_006720745.1 XM_006720746.1 XM_006720747.3 XM_006720749.1 XM_006720751.3 XM_006720752.3 XM_006720754.1 XM_006720755.1 XM_011522181.2 XM_011522182.1 XM_011522183.2 XM_011522184.1 XM_011522185.1 XM_011522186.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871472 c.604G>A p.V202M Chr.15: 45120271-45120271 on Build GRCh38
4251526 c.717C>T p.G239G Chr.15: 45120158-45120158 on Build GRCh38
4909338 c.726G>A p.V242V Chr.15: 45120149-45120149 on Build GRCh38
4909285 c.638C>T p.T213M Chr.15: 45120237-45120237 on Build GRCh38
4251530 c.634G>A p.A212T Chr.15: 45120241-45120241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760767564 Chr.15: 45120289-45120289 on Build GRCh38
rs751723195 Chr.15: 45120095-45120095 on Build GRCh38
rs751297912 Chr.15: 45120336-45120336 on Build GRCh38
rs747637552 Chr.15: 45120369-45120369 on Build GRCh38
rs567061569 Chr.15: 45120157-45120157 on Build GRCh38
rs755147038 Chr.15: 45120197-45120197 on Build GRCh38
rs758196241 Chr.15: 45120320-45120320 on Build GRCh38
rs777904853 Chr.15: 45120112-45120112 on Build GRCh38
rs764615428 Chr.15: 45120052-45120052 on Build GRCh38
rs773723948 Chr.15: 45120249-45120249 on Build GRCh38
rs751812698 Chr.15: 45120192-45120192 on Build GRCh38
rs1015148261 Chr.15: 45120113-45120113 on Build GRCh38
rs146181954 Chr.15: 45120135-45120135 on Build GRCh38
rs550198186 Chr.15: 45120392-45120392 on Build GRCh38
rs769327636 Chr.15: 45120373-45120373 on Build GRCh38
rs187988297 Chr.15: 45120481-45120481 on Build GRCh38
rs192944506 Chr.15: 45120079-45120079 on Build GRCh38
rs201757094 Chr.15: 45120148-45120148 on Build GRCh38
rs759472083 Chr.15: 45120275-45120275 on Build GRCh38
rs777190569 Chr.15: 45120161-45120161 on Build GRCh38
rs574036594 Chr.15: 45120462-45120462 on Build GRCh38
rs528501658 Chr.15: 45120393-45120393 on Build GRCh38
rs757152121 Chr.15: 45120209-45120209 on Build GRCh38
rs200642821 Chr.15: 45120191-45120191 on Build GRCh38
rs200164792 Chr.15: 45120213-45120213 on Build GRCh38
rs532727492 Chr.15: 45120428-45120428 on Build GRCh38
rs112792333 Chr.15: 45120402-45120402 on Build GRCh38
rs569682046 Chr.15: 45120436-45120436 on Build GRCh38
rs959298089 Chr.15: 45120166-45120166 on Build GRCh38
rs946139552 Chr.15: 45120454-45120454 on Build GRCh38
rs535394171 Chr.15: 45120477-45120477 on Build GRCh38
rs1040453262 Chr.15: 45120040-45120040 on Build GRCh38
rs753881760 Chr.15: 45120204-45120204 on Build GRCh38
rs762295474 Chr.15: 45120163-45120163 on Build GRCh38
rs768214542 Chr.15: 45120236-45120236 on Build GRCh38
rs771413812 Chr.15: 45120202-45120204 on Build GRCh38
rs371203619 Chr.15: 45120084-45120084 on Build GRCh38
rs528246024 Chr.15: 45120039-45120039 on Build GRCh38
rs779812256 Chr.15: 45120333-45120333 on Build GRCh38
rs146941277 Chr.15: 45120129-45120129 on Build GRCh38
rs369510098 Chr.15: 45120082-45120082 on Build GRCh38
rs754630018 Chr.15: 45120339-45120339 on Build GRCh38
rs746662073 Chr.15: 45120231-45120231 on Build GRCh38
rs367933259 Chr.15: 45120265-45120265 on Build GRCh38
rs1024633431 Chr.15: 45120444-45120444 on Build GRCh38
rs891857413 Chr.15: 45120425-45120425 on Build GRCh38
rs768124734 Chr.15: 45120158-45120158 on Build GRCh38
rs578186395 Chr.15: 45120394-45120394 on Build GRCh38
rs770239459 Chr.15: 45120241-45120241 on Build GRCh38
rs1040402705 Chr.15: 45120285-45120285 on Build GRCh38
rs780787516 Chr.15: 45120355-45120355 on Build GRCh38
rs771627364 Chr.15: 45120143-45120143 on Build GRCh38
rs1024699599 Chr.15: 45120235-45120235 on Build GRCh38
rs762207584 Chr.15: 45120074-45120074 on Build GRCh38
rs763340879 Chr.15: 45120187-45120187 on Build GRCh38
rs375934424 Chr.15: 45120287-45120287 on Build GRCh38
rs778555105 Chr.15: 45120133-45120133 on Build GRCh38
rs932071315 Chr.15: 45120329-45120329 on Build GRCh38
rs1051784648 Chr.15: 45120413-45120413 on Build GRCh38
rs766561153 Chr.15: 45120085-45120085 on Build GRCh38
rs765107564 Chr.15: 45120299-45120299 on Build GRCh38
rs988035611 Chr.15: 45120261-45120261 on Build GRCh38
rs267604232 Chr.15: 45120116-45120116 on Build GRCh38
rs763279284 Chr.15: 45120252-45120252 on Build GRCh38
rs770838058 Chr.15: 45120230-45120230 on Build GRCh38
rs182466172 Chr.15: 45120440-45120440 on Build GRCh38
rs757181935 Chr.15: 45120131-45120131 on Build GRCh38
rs756679960 Chr.15: 45120075-45120076 on Build GRCh38
rs775169480 Chr.15: 45120498-45120498 on Build GRCh38
rs767697506 Chr.15: 45120199-45120199 on Build GRCh38
rs1009326616 Chr.15: 45120127-45120127 on Build GRCh38
rs371597352 Chr.15: 45120228-45120228 on Build GRCh38
rs759815184 Chr.15: 45120268-45120268 on Build GRCh38
rs745432029 Chr.15: 45120293-45120294 on Build GRCh38
rs766732857 Chr.15: 45120255-45120255 on Build GRCh38
rs774777765 Chr.15: 45120263-45120263 on Build GRCh38
rs551669749 Chr.15: 45120051-45120051 on Build GRCh38
rs750350747 Chr.15: 45120317-45120317 on Build GRCh38
rs139046370 Chr.15: 45120171-45120171 on Build GRCh38
rs774865691 Chr.15: 45120397-45120397 on Build GRCh38
rs765717743 Chr.15: 45120170-45120170 on Build GRCh38
rs149960164 Chr.15: 45120237-45120237 on Build GRCh38
rs909386171 Chr.15: 45120284-45120284 on Build GRCh38
rs969490464 Chr.15: 45120452-45120452 on Build GRCh38

View Full Product Details