Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52120822-52121330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB5
Entrez Gene ID
10681
Gene Name
G protein subunit beta 5
Gene Aliases
GB5
UniGene
Hs.155090
Gene Chromosome Location
Chr.15: 52120926-52191368 on Build GRCh38
Transcripts
NM_006578.3 NM_016194.3 XM_011521162.2 XM_011521163.2 XM_017021867.1

SNP Detail

SNP ID Chromosome Location
rs958060819 Chr.15: 52121206-52121206 on Build GRCh38
rs747666703 Chr.15: 52120942-52120942 on Build GRCh38
rs1028202583 Chr.15: 52121269-52121269 on Build GRCh38
rs1048750677 Chr.15: 52121104-52121104 on Build GRCh38
rs528595733 Chr.15: 52120851-52120851 on Build GRCh38
rs926291824 Chr.15: 52121117-52121117 on Build GRCh38
rs966212629 Chr.15: 52121001-52121001 on Build GRCh38
rs913141654 Chr.15: 52120876-52120876 on Build GRCh38
rs746340016 Chr.15: 52121015-52121015 on Build GRCh38
rs911957393 Chr.15: 52121000-52121000 on Build GRCh38
rs574013044 Chr.15: 52121183-52121183 on Build GRCh38
rs953197872 Chr.15: 52121274-52121274 on Build GRCh38
rs985881363 Chr.15: 52121294-52121294 on Build GRCh38
rs534995751 Chr.15: 52121025-52121025 on Build GRCh38
rs12396 Chr.15: 52121119-52121119 on Build GRCh38
rs1000730216 Chr.15: 52121240-52121240 on Build GRCh38
rs28570348 Chr.15: 52121092-52121092 on Build GRCh38
rs953371529 Chr.15: 52120940-52120940 on Build GRCh38
rs544738476 Chr.15: 52121184-52121184 on Build GRCh38
rs779587307 Chr.15: 52121004-52121004 on Build GRCh38
rs1032355959 Chr.15: 52120845-52120845 on Build GRCh38
rs923159594 Chr.15: 52121270-52121270 on Build GRCh38
rs147851158 Chr.15: 52121100-52121101 on Build GRCh38
rs575356921 Chr.15: 52121276-52121276 on Build GRCh38
rs1002315963 Chr.15: 52121136-52121136 on Build GRCh38
rs967925655 Chr.15: 52120861-52120861 on Build GRCh38
rs1050122224 Chr.15: 52121035-52121035 on Build GRCh38
rs545796744 Chr.15: 52121308-52121308 on Build GRCh38
rs1037055175 Chr.15: 52121082-52121082 on Build GRCh38
rs908905539 Chr.15: 52121053-52121053 on Build GRCh38
rs1009580900 Chr.15: 52121211-52121211 on Build GRCh38
rs41277702 Chr.15: 52121087-52121087 on Build GRCh38
rs903819024 Chr.15: 52121234-52121234 on Build GRCh38
rs552868900 Chr.15: 52120931-52120931 on Build GRCh38
rs1042185639 Chr.15: 52121220-52121220 on Build GRCh38
rs986622535 Chr.15: 52120957-52120957 on Build GRCh38
rs570879593 Chr.15: 52120968-52120968 on Build GRCh38
rs1007528996 Chr.15: 52121291-52121291 on Build GRCh38
rs1032426636 Chr.15: 52121143-52121143 on Build GRCh38
rs556882266 Chr.15: 52121178-52121178 on Build GRCh38
rs919531085 Chr.15: 52121045-52121045 on Build GRCh38
rs1009385388 Chr.15: 52121215-52121215 on Build GRCh38
rs967668006 Chr.15: 52120887-52120887 on Build GRCh38
rs553538709 Chr.15: 52121039-52121039 on Build GRCh38
rs1000177233 Chr.15: 52120911-52120911 on Build GRCh38
rs1046575411 Chr.15: 52121112-52121112 on Build GRCh38
rs953522066 Chr.15: 52121287-52121287 on Build GRCh38
rs7177966 Chr.15: 52120853-52120853 on Build GRCh38
rs769373210 Chr.15: 52120951-52120951 on Build GRCh38
rs941696683 Chr.15: 52121057-52121057 on Build GRCh38
rs1033424144 Chr.15: 52120932-52120932 on Build GRCh38

View Full Product Details