Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52549976-52550480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

SNP Detail

SNP ID Chromosome Location
rs779783648 Chr.15: 52550098-52550098 on Build GRCh38
rs964682750 Chr.15: 52550107-52550107 on Build GRCh38
rs559692753 Chr.15: 52550163-52550163 on Build GRCh38
rs1055277944 Chr.15: 52550243-52550243 on Build GRCh38
rs752208206 Chr.15: 52550359-52550362 on Build GRCh38
rs999754377 Chr.15: 52550035-52550035 on Build GRCh38
rs564179437 Chr.15: 52550320-52550320 on Build GRCh38
rs192347686 Chr.15: 52550346-52550346 on Build GRCh38
rs6493566 Chr.15: 52550362-52550362 on Build GRCh38
rs999406221 Chr.15: 52550330-52550332 on Build GRCh38
rs955473744 Chr.15: 52550069-52550069 on Build GRCh38
rs774348109 Chr.15: 52550198-52550198 on Build GRCh38
rs35885704 Chr.15: 52550431-52550431 on Build GRCh38
rs376584537 Chr.15: 52550020-52550020 on Build GRCh38
rs894835433 Chr.15: 52550262-52550262 on Build GRCh38
rs189253343 Chr.15: 52550185-52550185 on Build GRCh38
rs201162765 Chr.15: 52550283-52550283 on Build GRCh38
rs772171607 Chr.15: 52550276-52550276 on Build GRCh38
rs563536810 Chr.15: 52550268-52550268 on Build GRCh38
rs184792598 Chr.15: 52550141-52550141 on Build GRCh38
rs1548220 Chr.15: 52550140-52550140 on Build GRCh38
rs568715984 Chr.15: 52550087-52550087 on Build GRCh38
rs964733917 Chr.15: 52550432-52550432 on Build GRCh38
rs768418255 Chr.15: 52550158-52550158 on Build GRCh38
rs200129115 Chr.15: 52550273-52550273 on Build GRCh38
rs557192150 Chr.15: 52550265-52550265 on Build GRCh38
rs902253747 Chr.15: 52550329-52550329 on Build GRCh38
rs1006830078 Chr.15: 52550416-52550416 on Build GRCh38
rs973506187 Chr.15: 52550124-52550128 on Build GRCh38
rs542138502 Chr.15: 52550237-52550237 on Build GRCh38
rs558764038 Chr.15: 52550095-52550095 on Build GRCh38
rs1043485669 Chr.15: 52550312-52550312 on Build GRCh38
rs946368591 Chr.15: 52550303-52550303 on Build GRCh38
rs1029615629 Chr.15: 52550040-52550040 on Build GRCh38
rs928116034 Chr.15: 52550210-52550210 on Build GRCh38
rs939608334 Chr.15: 52550231-52550231 on Build GRCh38
rs192076646 Chr.15: 52550081-52550081 on Build GRCh38
rs766733614 Chr.15: 52550435-52550435 on Build GRCh38

View Full Product Details