Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52584472-52585016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214A
Entrez Gene ID
56204
Gene Name
family with sequence similarity 214 member A
Gene Aliases
KIAA1370
UniGene
Hs.152385
Gene Chromosome Location
Chr.15: 52581321-52709817 on Build GRCh38
Transcripts
NM_001286495.1 NM_019600.3 NR_104457.1 XM_005254547.2 XM_005254548.2 XM_011521789.2 XM_011521790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638636 c.3009G>C p.K1003N Chr.15: 52584813-52584813 on Build GRCh38
3386941 c.2899A>C p.T967P Chr.15: 52584923-52584923 on Build GRCh38
4055593 c.2988A>G p.V996V Chr.15: 52584834-52584834 on Build GRCh38
1373517 c.2969G>A p.R990Q Chr.15: 52584853-52584853 on Build GRCh38
4820582 c.3004G>A p.V1002M Chr.15: 52584818-52584818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566905949 Chr.15: 52584739-52584739 on Build GRCh38
rs746208680 Chr.15: 52584974-52584975 on Build GRCh38
rs990919183 Chr.15: 52584678-52584678 on Build GRCh38
rs1044957542 Chr.15: 52584659-52584659 on Build GRCh38
rs778778734 Chr.15: 52584724-52584724 on Build GRCh38
rs12909204 Chr.15: 52584626-52584626 on Build GRCh38
rs1051296466 Chr.15: 52584576-52584576 on Build GRCh38
rs772224606 Chr.15: 52584788-52584788 on Build GRCh38
rs775606973 Chr.15: 52584953-52584953 on Build GRCh38
rs564079979 Chr.15: 52584655-52584655 on Build GRCh38
rs906486255 Chr.15: 52584662-52584662 on Build GRCh38
rs542275719 Chr.15: 52584530-52584530 on Build GRCh38
rs764736400 Chr.15: 52584809-52584809 on Build GRCh38
rs530344968 Chr.15: 52584648-52584648 on Build GRCh38
rs975001859 Chr.15: 52584570-52584570 on Build GRCh38
rs779785053 Chr.15: 52584741-52584741 on Build GRCh38
rs754305984 Chr.15: 52584714-52584714 on Build GRCh38
rs568882477 Chr.15: 52584933-52584933 on Build GRCh38
rs759467569 Chr.15: 52584787-52584787 on Build GRCh38
rs764920633 Chr.15: 52584854-52584854 on Build GRCh38
rs951368458 Chr.15: 52584654-52584654 on Build GRCh38
rs898283507 Chr.15: 52584883-52584883 on Build GRCh38
rs377630038 Chr.15: 52584886-52584886 on Build GRCh38
rs749943704 Chr.15: 52584725-52584725 on Build GRCh38
rs749403842 Chr.15: 52584719-52584720 on Build GRCh38
rs1029234345 Chr.15: 52584831-52584831 on Build GRCh38
rs531592953 Chr.15: 52584564-52584564 on Build GRCh38
rs942413773 Chr.15: 52584818-52584818 on Build GRCh38
rs770995987 Chr.15: 52584808-52584812 on Build GRCh38
rs746123749 Chr.15: 52584924-52584924 on Build GRCh38
rs757810101 Chr.15: 52584882-52584882 on Build GRCh38
rs781174399 Chr.15: 52584913-52584913 on Build GRCh38
rs375974954 Chr.15: 52584701-52584701 on Build GRCh38
rs759728110 Chr.15: 52584696-52584696 on Build GRCh38
rs760909579 Chr.15: 52584704-52584704 on Build GRCh38
rs750109132 Chr.15: 52584857-52584857 on Build GRCh38
rs61731680 Chr.15: 52584819-52584819 on Build GRCh38
rs769269209 Chr.15: 52584979-52584979 on Build GRCh38
rs747366445 Chr.15: 52584959-52584959 on Build GRCh38
rs774704938 Chr.15: 52584819-52584823 on Build GRCh38
rs907930552 Chr.15: 52584600-52584600 on Build GRCh38
rs935496906 Chr.15: 52584575-52584575 on Build GRCh38
rs747978471 Chr.15: 52584773-52584773 on Build GRCh38
rs762206482 Chr.15: 52584990-52584990 on Build GRCh38
rs754915516 Chr.15: 52584903-52584903 on Build GRCh38
rs748268872 Chr.15: 52584762-52584762 on Build GRCh38
rs747926495 Chr.15: 52584918-52584918 on Build GRCh38
rs369270208 Chr.15: 52584771-52584771 on Build GRCh38
rs879221810 Chr.15: 52584677-52584678 on Build GRCh38
rs779067395 Chr.15: 52584922-52584922 on Build GRCh38
rs557100113 Chr.15: 52584539-52584539 on Build GRCh38
rs12909037 Chr.15: 52584572-52584572 on Build GRCh38
rs762322962 Chr.15: 52584852-52584852 on Build GRCh38
rs12910551 Chr.15: 52584677-52584677 on Build GRCh38
rs756941411 Chr.15: 52584722-52584722 on Build GRCh38
rs184952260 Chr.15: 52584599-52584599 on Build GRCh38
rs764543020 Chr.15: 52584706-52584706 on Build GRCh38
rs904696563 Chr.15: 52584812-52584812 on Build GRCh38
rs528992766 Chr.15: 52584789-52584789 on Build GRCh38
rs757935154 Chr.15: 52584737-52584737 on Build GRCh38
rs968933169 Chr.15: 52584565-52584565 on Build GRCh38
rs1021822424 Chr.15: 52584515-52584515 on Build GRCh38
rs973181647 Chr.15: 52584630-52584630 on Build GRCh38
rs751082116 Chr.15: 52584899-52584899 on Build GRCh38
rs561903308 Chr.15: 52584702-52584702 on Build GRCh38
rs777287550 Chr.15: 52584982-52584982 on Build GRCh38
rs774614658 Chr.15: 52584782-52584782 on Build GRCh38
rs987852038 Chr.15: 52584917-52584917 on Build GRCh38
rs754741307 Chr.15: 52584973-52584973 on Build GRCh38
rs892189180 Chr.15: 52584719-52584719 on Build GRCh38
rs12324699 Chr.15: 52584678-52584678 on Build GRCh38
rs12909472 Chr.15: 52584608-52584608 on Build GRCh38
rs979498440 Chr.15: 52584717-52584717 on Build GRCh38
rs575194172 Chr.15: 52584583-52584583 on Build GRCh38
rs924252206 Chr.15: 52584568-52584568 on Build GRCh38
rs768283661 Chr.15: 52584764-52584764 on Build GRCh38
rs935626711 Chr.15: 52584749-52584749 on Build GRCh38
rs753459045 Chr.15: 52584944-52584944 on Build GRCh38
rs755088118 Chr.15: 52584558-52584558 on Build GRCh38
rs1035555506 Chr.15: 52584660-52584663 on Build GRCh38
rs962404142 Chr.15: 52584981-52584981 on Build GRCh38
rs954864535 Chr.15: 52584590-52584590 on Build GRCh38
rs746995127 Chr.15: 52584758-52584758 on Build GRCh38
rs761220407 Chr.15: 52584793-52584793 on Build GRCh38
rs920436531 Chr.15: 52584635-52584635 on Build GRCh38
rs771138466 Chr.15: 52584780-52584780 on Build GRCh38

View Full Product Details