Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52601039-52601576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214A
Entrez Gene ID
56204
Gene Name
family with sequence similarity 214 member A
Gene Aliases
KIAA1370
UniGene
Hs.152385
Gene Chromosome Location
Chr.15: 52581321-52709817 on Build GRCh38
Transcripts
NM_001286495.1 NM_019600.3 NR_104457.1 XM_005254547.2 XM_005254548.2 XM_011521789.2 XM_011521790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404680 c.2524A>T p.S842C Chr.15: 52601102-52601102 on Build GRCh38
4793868 c.2525G>A p.S842N Chr.15: 52601101-52601101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867773527 Chr.15: 52601143-52601143 on Build GRCh38
rs796702623 Chr.15: 52601530-52601533 on Build GRCh38
rs928856566 Chr.15: 52601483-52601483 on Build GRCh38
rs982366651 Chr.15: 52601464-52601464 on Build GRCh38
rs893328318 Chr.15: 52601206-52601206 on Build GRCh38
rs200981712 Chr.15: 52601530-52601530 on Build GRCh38
rs113059311 Chr.15: 52601542-52601542 on Build GRCh38
rs923924133 Chr.15: 52601553-52601553 on Build GRCh38
rs753990738 Chr.15: 52601145-52601145 on Build GRCh38
rs746171201 Chr.15: 52601190-52601190 on Build GRCh38
rs538035818 Chr.15: 52601114-52601114 on Build GRCh38
rs757419346 Chr.15: 52601206-52601207 on Build GRCh38
rs756179539 Chr.15: 52601104-52601104 on Build GRCh38
rs751377677 Chr.15: 52601173-52601174 on Build GRCh38
rs893286643 Chr.15: 52601552-52601552 on Build GRCh38
rs764058348 Chr.15: 52601108-52601108 on Build GRCh38
rs867019435 Chr.15: 52601129-52601129 on Build GRCh38
rs767793196 Chr.15: 52601098-52601098 on Build GRCh38
rs757839805 Chr.15: 52601503-52601503 on Build GRCh38
rs929951084 Chr.15: 52601477-52601477 on Build GRCh38
rs372913031 Chr.15: 52601135-52601135 on Build GRCh38
rs749328036 Chr.15: 52601205-52601206 on Build GRCh38
rs750895250 Chr.15: 52601480-52601480 on Build GRCh38
rs986336442 Chr.15: 52601384-52601384 on Build GRCh38
rs77767490 Chr.15: 52601531-52601531 on Build GRCh38
rs748117465 Chr.15: 52601490-52601491 on Build GRCh38
rs953279607 Chr.15: 52601340-52601340 on Build GRCh38
rs752973274 Chr.15: 52601101-52601101 on Build GRCh38
rs869278782 Chr.15: 52601543-52601545 on Build GRCh38
rs1018584350 Chr.15: 52601131-52601131 on Build GRCh38
rs951052161 Chr.15: 52601251-52601251 on Build GRCh38
rs1006737396 Chr.15: 52601067-52601067 on Build GRCh38
rs540476771 Chr.15: 52601490-52601490 on Build GRCh38
rs997196062 Chr.15: 52601322-52601322 on Build GRCh38
rs983859503 Chr.15: 52601289-52601289 on Build GRCh38
rs949652142 Chr.15: 52601437-52601437 on Build GRCh38
rs201371932 Chr.15: 52601328-52601332 on Build GRCh38
rs991008709 Chr.15: 52601356-52601356 on Build GRCh38
rs781198305 Chr.15: 52601194-52601195 on Build GRCh38
rs757699422 Chr.15: 52601146-52601146 on Build GRCh38
rs56856069 Chr.15: 52601545-52601545 on Build GRCh38
rs186633306 Chr.15: 52601205-52601205 on Build GRCh38
rs1016707740 Chr.15: 52601300-52601300 on Build GRCh38
rs963859559 Chr.15: 52601323-52601323 on Build GRCh38
rs553319406 Chr.15: 52601404-52601404 on Build GRCh38
rs930003310 Chr.15: 52601085-52601085 on Build GRCh38
rs561951915 Chr.15: 52601520-52601520 on Build GRCh38
rs1042544074 Chr.15: 52601546-52601546 on Build GRCh38
rs754852759 Chr.15: 52601195-52601195 on Build GRCh38
rs779453731 Chr.15: 52601178-52601178 on Build GRCh38
rs529233905 Chr.15: 52601280-52601282 on Build GRCh38
rs865874344 Chr.15: 52601347-52601347 on Build GRCh38
rs538292422 Chr.15: 52601336-52601336 on Build GRCh38
rs148622329 Chr.15: 52601235-52601235 on Build GRCh38
rs1020474033 Chr.15: 52601274-52601274 on Build GRCh38
rs901093180 Chr.15: 52601200-52601200 on Build GRCh38
rs918700508 Chr.15: 52601466-52601466 on Build GRCh38
rs573220926 Chr.15: 52601413-52601413 on Build GRCh38
rs963583639 Chr.15: 52601408-52601408 on Build GRCh38

View Full Product Details