Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64165136-64165659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G1
Entrez Gene ID
53944
Gene Name
casein kinase 1 gamma 1
Gene Aliases
CK1gamma1
UniGene
Hs.646508
Gene Chromosome Location
Chr.15: 64165517-64356243 on Build GRCh38
Transcripts
NM_022048.3

SNP Detail

SNP ID Chromosome Location
rs541362883 Chr.15: 64165231-64165231 on Build GRCh38
rs998415500 Chr.15: 64165247-64165247 on Build GRCh38
rs181167351 Chr.15: 64165472-64165472 on Build GRCh38
rs903476170 Chr.15: 64165439-64165439 on Build GRCh38
rs970096805 Chr.15: 64165243-64165243 on Build GRCh38
rs752013683 Chr.15: 64165544-64165544 on Build GRCh38
rs901356104 Chr.15: 64165220-64165220 on Build GRCh38
rs867042673 Chr.15: 64165446-64165446 on Build GRCh38
rs1031486847 Chr.15: 64165421-64165421 on Build GRCh38
rs930295236 Chr.15: 64165345-64165345 on Build GRCh38
rs371199936 Chr.15: 64165473-64165473 on Build GRCh38
rs150397261 Chr.15: 64165256-64165256 on Build GRCh38
rs903974426 Chr.15: 64165384-64165385 on Build GRCh38
rs993324690 Chr.15: 64165629-64165629 on Build GRCh38
rs1024605982 Chr.15: 64165440-64165440 on Build GRCh38
rs980209834 Chr.15: 64165479-64165479 on Build GRCh38
rs190051091 Chr.15: 64165187-64165187 on Build GRCh38
rs762057745 Chr.15: 64165286-64165286 on Build GRCh38
rs934158343 Chr.15: 64165420-64165420 on Build GRCh38
rs951800154 Chr.15: 64165488-64165488 on Build GRCh38
rs773182060 Chr.15: 64165518-64165519 on Build GRCh38
rs548949099 Chr.15: 64165339-64165339 on Build GRCh38
rs999546067 Chr.15: 64165408-64165408 on Build GRCh38
rs971915468 Chr.15: 64165335-64165335 on Build GRCh38
rs910281652 Chr.15: 64165370-64165370 on Build GRCh38
rs1000236739 Chr.15: 64165266-64165266 on Build GRCh38
rs979878720 Chr.15: 64165244-64165244 on Build GRCh38
rs944816747 Chr.15: 64165553-64165553 on Build GRCh38
rs146092609 Chr.15: 64165338-64165338 on Build GRCh38
rs1023988427 Chr.15: 64165236-64165236 on Build GRCh38
rs533173384 Chr.15: 64165277-64165277 on Build GRCh38
rs773618282 Chr.15: 64165386-64165386 on Build GRCh38
rs567178814 Chr.15: 64165422-64165422 on Build GRCh38
rs1039924782 Chr.15: 64165215-64165215 on Build GRCh38
rs1024346166 Chr.15: 64165533-64165533 on Build GRCh38
rs776789513 Chr.15: 64165280-64165280 on Build GRCh38
rs894478053 Chr.15: 64165497-64165497 on Build GRCh38
rs766978258 Chr.15: 64165527-64165527 on Build GRCh38
rs1026646806 Chr.15: 64165630-64165630 on Build GRCh38
rs368769128 Chr.15: 64165434-64165434 on Build GRCh38
rs971170795 Chr.15: 64165566-64165566 on Build GRCh38
rs539647130 Chr.15: 64165551-64165551 on Build GRCh38
rs555517068 Chr.15: 64165176-64165176 on Build GRCh38
rs1040166411 Chr.15: 64165395-64165395 on Build GRCh38
rs922728982 Chr.15: 64165400-64165400 on Build GRCh38
rs867174233 Chr.15: 64165163-64165163 on Build GRCh38
rs1027312198 Chr.15: 64165482-64165482 on Build GRCh38
rs908018718 Chr.15: 64165306-64165306 on Build GRCh38
rs549243761 Chr.15: 64165427-64165427 on Build GRCh38
rs942829138 Chr.15: 64165204-64165204 on Build GRCh38
rs958839637 Chr.15: 64165274-64165274 on Build GRCh38
rs1043723685 Chr.15: 64165341-64165341 on Build GRCh38
rs984498865 Chr.15: 64165365-64165365 on Build GRCh38
rs565895069 Chr.15: 64165526-64165526 on Build GRCh38
rs768618753 Chr.15: 64165260-64165260 on Build GRCh38
rs879706607 Chr.15: 64165269-64165269 on Build GRCh38
rs138228809 Chr.15: 64165305-64165305 on Build GRCh38
rs955978448 Chr.15: 64165186-64165189 on Build GRCh38
rs1020553674 Chr.15: 64165254-64165254 on Build GRCh38
rs28380607 Chr.15: 64165312-64165312 on Build GRCh38
rs779981389 Chr.15: 64165206-64165206 on Build GRCh38
rs908048090 Chr.15: 64165531-64165531 on Build GRCh38

View Full Product Details