Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64167548-64168043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G1
Entrez Gene ID
53944
Gene Name
casein kinase 1 gamma 1
Gene Aliases
CK1gamma1
UniGene
Hs.646508
Gene Chromosome Location
Chr.15: 64165517-64356243 on Build GRCh38
Transcripts
NM_022048.3

SNP Detail

SNP ID Chromosome Location
rs893484438 Chr.15: 64167607-64167607 on Build GRCh38
rs974789377 Chr.15: 64167699-64167699 on Build GRCh38
rs970193790 Chr.15: 64167644-64167644 on Build GRCh38
rs934026272 Chr.15: 64167975-64167976 on Build GRCh38
rs556984581 Chr.15: 64167744-64167744 on Build GRCh38
rs922043502 Chr.15: 64167702-64167702 on Build GRCh38
rs571692233 Chr.15: 64167717-64167717 on Build GRCh38
rs538892162 Chr.15: 64167741-64167741 on Build GRCh38
rs755539403 Chr.15: 64167919-64167919 on Build GRCh38
rs927738129 Chr.15: 64167859-64167859 on Build GRCh38
rs572586774 Chr.15: 64167934-64167934 on Build GRCh38
rs1002984744 Chr.15: 64167925-64167925 on Build GRCh38
rs935215805 Chr.15: 64167805-64167805 on Build GRCh38
rs539966839 Chr.15: 64167935-64167935 on Build GRCh38
rs749494627 Chr.15: 64167597-64167597 on Build GRCh38
rs1011903932 Chr.15: 64167866-64167866 on Build GRCh38
rs545158873 Chr.15: 64167672-64167672 on Build GRCh38
rs897158385 Chr.15: 64167962-64167962 on Build GRCh38
rs1037055636 Chr.15: 64167678-64167678 on Build GRCh38
rs966738979 Chr.15: 64167815-64167815 on Build GRCh38
rs528633913 Chr.15: 64167660-64167660 on Build GRCh38
rs941577687 Chr.15: 64167675-64167675 on Build GRCh38
rs1017373051 Chr.15: 64167785-64167785 on Build GRCh38
rs941608961 Chr.15: 64167909-64167909 on Build GRCh38
rs771286692 Chr.15: 64167624-64167624 on Build GRCh38
rs772868123 Chr.15: 64167908-64167908 on Build GRCh38
rs923868043 Chr.15: 64167761-64167761 on Build GRCh38
rs72756915 Chr.15: 64167829-64167829 on Build GRCh38
rs957966146 Chr.15: 64167786-64167786 on Build GRCh38
rs1053891494 Chr.15: 64167598-64167598 on Build GRCh38
rs145082805 Chr.15: 64168014-64168014 on Build GRCh38
rs889876430 Chr.15: 64167754-64167754 on Build GRCh38
rs910028229 Chr.15: 64167886-64167886 on Build GRCh38
rs1023349433 Chr.15: 64167633-64167633 on Build GRCh38
rs998289279 Chr.15: 64167701-64167701 on Build GRCh38
rs974778016 Chr.15: 64167988-64167988 on Build GRCh38
rs963374399 Chr.15: 64167682-64167682 on Build GRCh38
rs756192635 Chr.15: 64167587-64167587 on Build GRCh38
rs772761513 Chr.15: 64167783-64167783 on Build GRCh38
rs139479622 Chr.15: 64167640-64167640 on Build GRCh38
rs761337842 Chr.15: 64167821-64167821 on Build GRCh38
rs1029062594 Chr.15: 64167997-64167997 on Build GRCh38
rs776274828 Chr.15: 64167806-64167806 on Build GRCh38
rs1016522415 Chr.15: 64167948-64167948 on Build GRCh38
rs951065527 Chr.15: 64167661-64167661 on Build GRCh38
rs989325750 Chr.15: 64167791-64167791 on Build GRCh38
rs1011912418 Chr.15: 64167631-64167631 on Build GRCh38
rs536055186 Chr.15: 64167873-64167873 on Build GRCh38
rs1044790303 Chr.15: 64167898-64167898 on Build GRCh38
rs906317150 Chr.15: 64167906-64167906 on Build GRCh38
rs777993697 Chr.15: 64167589-64167589 on Build GRCh38
rs762557676 Chr.15: 64167960-64167960 on Build GRCh38
rs746362969 Chr.15: 64167680-64167680 on Build GRCh38
rs991010681 Chr.15: 64167878-64167878 on Build GRCh38
rs933136564 Chr.15: 64167705-64167705 on Build GRCh38
rs1003036739 Chr.15: 64167650-64167650 on Build GRCh38
rs554382398 Chr.15: 64167928-64167928 on Build GRCh38
rs766177340 Chr.15: 64167982-64167982 on Build GRCh38

View Full Product Details