Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65388357-65388865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGDCC4
Entrez Gene ID
57722
Gene Name
immunoglobulin superfamily DCC subclass member 4
Gene Aliases
DDM36, NOPE
UniGene
Hs.458607
Gene Chromosome Location
Chr.15: 65381480-65423072 on Build GRCh38
Transcripts
NM_020962.2 XM_011521845.2 XM_011521846.2 XM_011521847.1 XM_017022448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
352566 c.2784G>T p.E928D Chr.15: 65388510-65388510 on Build GRCh38
7426758 c.2738T>A p.L913Q Chr.15: 65388556-65388556 on Build GRCh38
6684169 c.2807G>A p.R936H Chr.15: 65388487-65388487 on Build GRCh38
2256509 c.2776C>T p.R926C Chr.15: 65388518-65388518 on Build GRCh38
4918035 c.2767A>T p.M923L Chr.15: 65388527-65388527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037606374 Chr.15: 65388664-65388664 on Build GRCh38
rs757089246 Chr.15: 65388622-65388622 on Build GRCh38
rs375451176 Chr.15: 65388552-65388552 on Build GRCh38
rs771231127 Chr.15: 65388812-65388812 on Build GRCh38
rs115076941 Chr.15: 65388487-65388487 on Build GRCh38
rs941708635 Chr.15: 65388658-65388658 on Build GRCh38
rs1012930000 Chr.15: 65388731-65388731 on Build GRCh38
rs751629777 Chr.15: 65388512-65388512 on Build GRCh38
rs748065941 Chr.15: 65388783-65388783 on Build GRCh38
rs758468988 Chr.15: 65388657-65388657 on Build GRCh38
rs771163753 Chr.15: 65388567-65388567 on Build GRCh38
rs746652250 Chr.15: 65388570-65388570 on Build GRCh38
rs140602610 Chr.15: 65388488-65388488 on Build GRCh38
rs562429008 Chr.15: 65388692-65388692 on Build GRCh38
rs749620830 Chr.15: 65388811-65388811 on Build GRCh38
rs746574899 Chr.15: 65388770-65388770 on Build GRCh38
rs59936444 Chr.15: 65388712-65388712 on Build GRCh38
rs759694694 Chr.15: 65388823-65388823 on Build GRCh38
rs372985723 Chr.15: 65388580-65388580 on Build GRCh38
rs764083284 Chr.15: 65388592-65388592 on Build GRCh38
rs752611906 Chr.15: 65388517-65388517 on Build GRCh38
rs895654837 Chr.15: 65388724-65388724 on Build GRCh38
rs954406549 Chr.15: 65388600-65388600 on Build GRCh38
rs749653210 Chr.15: 65388389-65388389 on Build GRCh38
rs202027229 Chr.15: 65388426-65388426 on Build GRCh38
rs996432299 Chr.15: 65388820-65388820 on Build GRCh38
rs781032031 Chr.15: 65388421-65388421 on Build GRCh38
rs202023951 Chr.15: 65388541-65388541 on Build GRCh38
rs769171850 Chr.15: 65388468-65388468 on Build GRCh38
rs559083416 Chr.15: 65388744-65388744 on Build GRCh38
rs773895737 Chr.15: 65388462-65388462 on Build GRCh38
rs544354391 Chr.15: 65388648-65388648 on Build GRCh38
rs201797562 Chr.15: 65388381-65388381 on Build GRCh38
rs904520138 Chr.15: 65388782-65388782 on Build GRCh38
rs759624587 Chr.15: 65388573-65388573 on Build GRCh38
rs1057335088 Chr.15: 65388719-65388719 on Build GRCh38
rs928861990 Chr.15: 65388749-65388749 on Build GRCh38
rs763453024 Chr.15: 65388566-65388566 on Build GRCh38
rs755929194 Chr.15: 65388435-65388435 on Build GRCh38
rs756897098 Chr.15: 65388528-65388528 on Build GRCh38
rs777050099 Chr.15: 65388480-65388480 on Build GRCh38
rs762218926 Chr.15: 65388482-65388482 on Build GRCh38
rs528128173 Chr.15: 65388757-65388757 on Build GRCh38
rs946239292 Chr.15: 65388448-65388448 on Build GRCh38
rs770394102 Chr.15: 65388781-65388781 on Build GRCh38
rs567417450 Chr.15: 65388403-65388403 on Build GRCh38
rs755062661 Chr.15: 65388513-65388513 on Build GRCh38
rs897646835 Chr.15: 65388384-65388384 on Build GRCh38
rs73478540 Chr.15: 65388742-65388742 on Build GRCh38
rs1011584840 Chr.15: 65388694-65388694 on Build GRCh38
rs919207549 Chr.15: 65388690-65388690 on Build GRCh38
rs200877185 Chr.15: 65388813-65388813 on Build GRCh38
rs376547484 Chr.15: 65388625-65388625 on Build GRCh38
rs778466995 Chr.15: 65388531-65388531 on Build GRCh38
rs750073405 Chr.15: 65388626-65388626 on Build GRCh38
rs575344635 Chr.15: 65388614-65388614 on Build GRCh38
rs113065376 Chr.15: 65388543-65388543 on Build GRCh38
rs1032368410 Chr.15: 65388545-65388545 on Build GRCh38
rs182317592 Chr.15: 65388519-65388519 on Build GRCh38
rs1012962744 Chr.15: 65388521-65388521 on Build GRCh38
rs770373738 Chr.15: 65388461-65388461 on Build GRCh38
rs374349637 Chr.15: 65388516-65388516 on Build GRCh38
rs142343031 Chr.15: 65388549-65388549 on Build GRCh38
rs754940989 Chr.15: 65388410-65388410 on Build GRCh38
rs774459548 Chr.15: 65388571-65388571 on Build GRCh38
rs777368442 Chr.15: 65388441-65388441 on Build GRCh38
rs753712156 Chr.15: 65388639-65388640 on Build GRCh38
rs1000423055 Chr.15: 65388542-65388542 on Build GRCh38
rs372069978 Chr.15: 65388590-65388590 on Build GRCh38
rs371304078 Chr.15: 65388750-65388750 on Build GRCh38
rs189426939 Chr.15: 65388391-65388391 on Build GRCh38
rs748947003 Chr.15: 65388449-65388449 on Build GRCh38
rs751485318 Chr.15: 65388727-65388727 on Build GRCh38
rs561167961 Chr.15: 65388652-65388652 on Build GRCh38
rs200414898 Chr.15: 65388483-65388483 on Build GRCh38
rs202153933 Chr.15: 65388518-65388518 on Build GRCh38
rs745357948 Chr.15: 65388540-65388540 on Build GRCh38
rs778474498 Chr.15: 65388798-65388798 on Build GRCh38
rs200113249 Chr.15: 65388548-65388548 on Build GRCh38

View Full Product Details