Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65916045-65916535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEGF11
Entrez Gene ID
84465
Gene Name
multiple EGF like domains 11
Gene Aliases
-
UniGene
Hs.712886
Gene Chromosome Location
Chr.15: 65895296-66253737 on Build GRCh38
Transcripts
NM_032445.2 XM_017022670.1 XM_017022671.1 XM_017022672.1 XM_017022673.1 XM_017022674.1 XM_017022675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809393 c.2290A>G p.I764V Chr.15: 65916202-65916202 on Build GRCh38
964160 c.2225C>A p.A742E Chr.15: 65916267-65916267 on Build GRCh38
162446 c.2065A>G p.I689V Chr.15: 65916202-65916202 on Build GRCh38
6025605 c.2312G>A p.R771H Chr.15: 65916180-65916180 on Build GRCh38
4747399 c.2235delT p.F745fs*55 Chr.15: 65916257-65916257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747220792 Chr.15: 65916263-65916265 on Build GRCh38
rs878937802 Chr.15: 65916441-65916441 on Build GRCh38
rs199886545 Chr.15: 65916190-65916190 on Build GRCh38
rs765193130 Chr.15: 65916181-65916181 on Build GRCh38
rs750174130 Chr.15: 65916187-65916187 on Build GRCh38
rs899125708 Chr.15: 65916310-65916310 on Build GRCh38
rs768650534 Chr.15: 65916282-65916283 on Build GRCh38
rs890752166 Chr.15: 65916258-65916258 on Build GRCh38
rs779506642 Chr.15: 65916202-65916202 on Build GRCh38
rs201139943 Chr.15: 65916130-65916130 on Build GRCh38
rs751572321 Chr.15: 65916239-65916239 on Build GRCh38
rs773158124 Chr.15: 65916145-65916145 on Build GRCh38
rs775566970 Chr.15: 65916295-65916295 on Build GRCh38
rs778140068 Chr.15: 65916118-65916118 on Build GRCh38
rs761013773 Chr.15: 65916299-65916299 on Build GRCh38
rs980619787 Chr.15: 65916513-65916513 on Build GRCh38
rs745935417 Chr.15: 65916067-65916067 on Build GRCh38
rs545155487 Chr.15: 65916498-65916498 on Build GRCh38
rs754434534 Chr.15: 65916218-65916218 on Build GRCh38
rs542430844 Chr.15: 65916429-65916429 on Build GRCh38
rs953723117 Chr.15: 65916362-65916362 on Build GRCh38
rs377093610 Chr.15: 65916153-65916153 on Build GRCh38
rs746447677 Chr.15: 65916100-65916100 on Build GRCh38
rs983117458 Chr.15: 65916109-65916109 on Build GRCh38
rs931000173 Chr.15: 65916260-65916260 on Build GRCh38
rs756477825 Chr.15: 65916461-65916461 on Build GRCh38
rs1048505168 Chr.15: 65916278-65916278 on Build GRCh38
rs573311927 Chr.15: 65916460-65916460 on Build GRCh38
rs767965074 Chr.15: 65916422-65916422 on Build GRCh38
rs373932693 Chr.15: 65916253-65916253 on Build GRCh38
rs761782026 Chr.15: 65916180-65916180 on Build GRCh38
rs1039061865 Chr.15: 65916303-65916303 on Build GRCh38
rs759625663 Chr.15: 65916240-65916240 on Build GRCh38
rs970549544 Chr.15: 65916514-65916514 on Build GRCh38
rs760602131 Chr.15: 65916165-65916165 on Build GRCh38
rs113004184 Chr.15: 65916354-65916357 on Build GRCh38
rs774894453 Chr.15: 65916140-65916140 on Build GRCh38
rs768590062 Chr.15: 65916301-65916301 on Build GRCh38
rs775790898 Chr.15: 65916158-65916158 on Build GRCh38
rs1032736549 Chr.15: 65916401-65916401 on Build GRCh38
rs764299442 Chr.15: 65916172-65916172 on Build GRCh38
rs995349118 Chr.15: 65916395-65916395 on Build GRCh38
rs377576849 Chr.15: 65916279-65916279 on Build GRCh38
rs775300026 Chr.15: 65916130-65916130 on Build GRCh38
rs144231820 Chr.15: 65916238-65916238 on Build GRCh38
rs28559977 Chr.15: 65916126-65916126 on Build GRCh38
rs753755756 Chr.15: 65916174-65916174 on Build GRCh38
rs902013389 Chr.15: 65916298-65916298 on Build GRCh38
rs1036516876 Chr.15: 65916297-65916297 on Build GRCh38
rs945596540 Chr.15: 65916244-65916244 on Build GRCh38
rs113008166 Chr.15: 65916302-65916302 on Build GRCh38
rs780801469 Chr.15: 65916224-65916224 on Build GRCh38
rs1026369704 Chr.15: 65916427-65916427 on Build GRCh38
rs760580124 Chr.15: 65916203-65916205 on Build GRCh38
rs746173752 Chr.15: 65916241-65916241 on Build GRCh38
rs1004072512 Chr.15: 65916433-65916433 on Build GRCh38
rs768217555 Chr.15: 65916104-65916104 on Build GRCh38
rs776558546 Chr.15: 65916302-65916302 on Build GRCh38
rs553258108 Chr.15: 65916065-65916065 on Build GRCh38
rs1029249738 Chr.15: 65916316-65916316 on Build GRCh38
rs201567733 Chr.15: 65916211-65916211 on Build GRCh38
rs140454685 Chr.15: 65916503-65916503 on Build GRCh38
rs772136365 Chr.15: 65916157-65916157 on Build GRCh38
rs11632730 Chr.15: 65916084-65916084 on Build GRCh38

View Full Product Details