Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68301555-68302049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
UniGene
Hs.436416
Gene Chromosome Location
Chr.15: 68297433-68432312 on Build GRCh38
Transcripts
NM_001004439.1 XM_005254228.3 XM_011521363.2

SNP Detail

SNP ID Chromosome Location
rs555753150 Chr.15: 68301975-68301975 on Build GRCh38
rs193082863 Chr.15: 68301960-68301960 on Build GRCh38
rs918177626 Chr.15: 68301590-68301590 on Build GRCh38
rs571080219 Chr.15: 68301811-68301815 on Build GRCh38
rs969544027 Chr.15: 68301926-68301926 on Build GRCh38
rs912976243 Chr.15: 68301958-68301958 on Build GRCh38
rs887162405 Chr.15: 68301857-68301857 on Build GRCh38
rs1026159870 Chr.15: 68301856-68301856 on Build GRCh38
rs145957262 Chr.15: 68301650-68301650 on Build GRCh38
rs527514294 Chr.15: 68301756-68301756 on Build GRCh38
rs942516773 Chr.15: 68301967-68301967 on Build GRCh38
rs567240009 Chr.15: 68301988-68301992 on Build GRCh38
rs796945267 Chr.15: 68301600-68301600 on Build GRCh38
rs1014181647 Chr.15: 68301905-68301905 on Build GRCh38
rs891049137 Chr.15: 68301779-68301779 on Build GRCh38
rs547679578 Chr.15: 68301907-68301907 on Build GRCh38
rs931309148 Chr.15: 68301598-68301598 on Build GRCh38
rs531244944 Chr.15: 68301665-68301665 on Build GRCh38
rs570118697 Chr.15: 68301961-68301961 on Build GRCh38
rs1050844702 Chr.15: 68301770-68301770 on Build GRCh38
rs957274972 Chr.15: 68301947-68301947 on Build GRCh38
rs951658030 Chr.15: 68301576-68301576 on Build GRCh38
rs992700839 Chr.15: 68301908-68301908 on Build GRCh38
rs1039592633 Chr.15: 68301825-68301825 on Build GRCh38
rs770283206 Chr.15: 68301721-68301721 on Build GRCh38
rs533346929 Chr.15: 68301946-68301946 on Build GRCh38
rs898381165 Chr.15: 68301991-68301991 on Build GRCh38
rs561499742 Chr.15: 68301742-68301742 on Build GRCh38
rs1032583475 Chr.15: 68301936-68301936 on Build GRCh38
rs945888004 Chr.15: 68301752-68301752 on Build GRCh38
rs1039623861 Chr.15: 68301973-68301973 on Build GRCh38
rs372375690 Chr.15: 68301788-68301788 on Build GRCh38
rs142114044 Chr.15: 68302030-68302030 on Build GRCh38
rs931311041 Chr.15: 68301998-68301998 on Build GRCh38
rs562598442 Chr.15: 68301652-68301652 on Build GRCh38
rs771194118 Chr.15: 68301964-68301964 on Build GRCh38
rs547645478 Chr.15: 68301787-68301787 on Build GRCh38
rs774912749 Chr.15: 68301995-68301995 on Build GRCh38
rs548186092 Chr.15: 68301692-68301692 on Build GRCh38
rs942653139 Chr.15: 68301800-68301800 on Build GRCh38
rs989809331 Chr.15: 68301955-68301955 on Build GRCh38
rs915862032 Chr.15: 68301939-68301939 on Build GRCh38
rs1041195959 Chr.15: 68301607-68301607 on Build GRCh38
rs981045131 Chr.15: 68301930-68301930 on Build GRCh38
rs565042442 Chr.15: 68301584-68301584 on Build GRCh38
rs565886498 Chr.15: 68302002-68302002 on Build GRCh38
rs1031312136 Chr.15: 68301819-68301819 on Build GRCh38
rs535542954 Chr.15: 68301965-68301965 on Build GRCh38
rs570752673 Chr.15: 68301847-68301847 on Build GRCh38
rs138965769 Chr.15: 68302015-68302015 on Build GRCh38
rs1054642868 Chr.15: 68301740-68301740 on Build GRCh38
rs763308495 Chr.15: 68301792-68301792 on Build GRCh38
rs981013889 Chr.15: 68301614-68301614 on Build GRCh38
rs773603111 Chr.15: 68301747-68301747 on Build GRCh38
rs898523983 Chr.15: 68301828-68301828 on Build GRCh38
rs542978496 Chr.15: 68301649-68301649 on Build GRCh38

View Full Product Details