Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69455308-69455814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPLP1
Entrez Gene ID
6176
Gene Name
ribosomal protein lateral stalk subunit P1
Gene Aliases
LP1, P1, RPP1
UniGene
Hs.356502
Gene Chromosome Location
Chr.15: 69452820-69455545 on Build GRCh38
Transcripts
NM_001003.2 NM_213725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582928 c.292A>G p.K98E Chr.15: 69455454-69455454 on Build GRCh38
4056540 c.299A>G p.E100G Chr.15: 69455461-69455461 on Build GRCh38
3711735 c.273G>A p.E91E Chr.15: 69455435-69455435 on Build GRCh38
1374295 c.335delT p.F113fs*>2 Chr.15: 69455497-69455497 on Build GRCh38
1323869 c.309G>C p.E103D Chr.15: 69455471-69455471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547455572 Chr.15: 69455665-69455665 on Build GRCh38
rs1004883858 Chr.15: 69455482-69455482 on Build GRCh38
rs923709615 Chr.15: 69455666-69455666 on Build GRCh38
rs752174225 Chr.15: 69455552-69455552 on Build GRCh38
rs142400150 Chr.15: 69455737-69455737 on Build GRCh38
rs781682538 Chr.15: 69455439-69455439 on Build GRCh38
rs570755023 Chr.15: 69455695-69455695 on Build GRCh38
rs780740996 Chr.15: 69455412-69455412 on Build GRCh38
rs930118598 Chr.15: 69455363-69455363 on Build GRCh38
rs139070897 Chr.15: 69455661-69455661 on Build GRCh38
rs202225518 Chr.15: 69455518-69455518 on Build GRCh38
rs1050913476 Chr.15: 69455622-69455622 on Build GRCh38
rs201563838 Chr.15: 69455553-69455553 on Build GRCh38
rs1025901530 Chr.15: 69455708-69455708 on Build GRCh38
rs951698043 Chr.15: 69455710-69455710 on Build GRCh38
rs200720703 Chr.15: 69455545-69455545 on Build GRCh38
rs763723146 Chr.15: 69455387-69455387 on Build GRCh38
rs201726748 Chr.15: 69455334-69455334 on Build GRCh38
rs549904170 Chr.15: 69455752-69455752 on Build GRCh38
rs867697105 Chr.15: 69455706-69455706 on Build GRCh38
rs748470409 Chr.15: 69455444-69455444 on Build GRCh38
rs927481703 Chr.15: 69455773-69455773 on Build GRCh38
rs755410929 Chr.15: 69455424-69455424 on Build GRCh38
rs760411129 Chr.15: 69455525-69455525 on Build GRCh38
rs762069154 Chr.15: 69455430-69455432 on Build GRCh38
rs544246404 Chr.15: 69455411-69455411 on Build GRCh38
rs765659388 Chr.15: 69455510-69455511 on Build GRCh38
rs992380171 Chr.15: 69455368-69455368 on Build GRCh38
rs749351343 Chr.15: 69455468-69455468 on Build GRCh38
rs918052935 Chr.15: 69455765-69455765 on Build GRCh38
rs539130213 Chr.15: 69455697-69455698 on Build GRCh38
rs1028850976 Chr.15: 69455592-69455592 on Build GRCh38
rs995197145 Chr.15: 69455707-69455707 on Build GRCh38
rs752047646 Chr.15: 69455416-69455416 on Build GRCh38
rs777960502 Chr.15: 69455466-69455466 on Build GRCh38
rs562950236 Chr.15: 69455465-69455465 on Build GRCh38
rs1018048078 Chr.15: 69455686-69455686 on Build GRCh38
rs1463911 Chr.15: 69455336-69455336 on Build GRCh38
rs768035130 Chr.15: 69455563-69455563 on Build GRCh38
rs754137512 Chr.15: 69455415-69455418 on Build GRCh38
rs769295132 Chr.15: 69455740-69455740 on Build GRCh38
rs116311376 Chr.15: 69455337-69455337 on Build GRCh38
rs753479085 Chr.15: 69455528-69455528 on Build GRCh38
rs774274098 Chr.15: 69455488-69455488 on Build GRCh38
rs576544725 Chr.15: 69455414-69455415 on Build GRCh38
rs752982175 Chr.15: 69455568-69455568 on Build GRCh38
rs761416059 Chr.15: 69455540-69455540 on Build GRCh38
rs760839391 Chr.15: 69455415-69455415 on Build GRCh38
rs564613487 Chr.15: 69455630-69455630 on Build GRCh38
rs11554438 Chr.15: 69455478-69455478 on Build GRCh38
rs891749437 Chr.15: 69455532-69455532 on Build GRCh38
rs906017074 Chr.15: 69455405-69455405 on Build GRCh38
rs796295422 Chr.15: 69455334-69455336 on Build GRCh38
rs1054280719 Chr.15: 69455493-69455493 on Build GRCh38
rs764191639 Chr.15: 69455415-69455417 on Build GRCh38
rs548004580 Chr.15: 69455527-69455527 on Build GRCh38
rs370055255 Chr.15: 69455483-69455483 on Build GRCh38
rs201427547 Chr.15: 69455512-69455512 on Build GRCh38
rs974630507 Chr.15: 69455573-69455573 on Build GRCh38
rs201272949 Chr.15: 69455452-69455452 on Build GRCh38
rs969979577 Chr.15: 69455776-69455776 on Build GRCh38
rs771001182 Chr.15: 69455336-69455336 on Build GRCh38
rs753253861 Chr.15: 69455406-69455406 on Build GRCh38
rs967890130 Chr.15: 69455631-69455631 on Build GRCh38

View Full Product Details