Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 70655626-70656155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UACA
Entrez Gene ID
55075
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
NUCLING
UniGene
Hs.108049
Gene Chromosome Location
Chr.15: 70654554-70778823 on Build GRCh38
Transcripts
NM_001008224.1 NM_018003.2 XM_011521752.2

SNP Detail

SNP ID Chromosome Location
rs530090733 Chr.15: 70655816-70655816 on Build GRCh38
rs902606299 Chr.15: 70656055-70656055 on Build GRCh38
rs948796091 Chr.15: 70655840-70655840 on Build GRCh38
rs752380957 Chr.15: 70655732-70655732 on Build GRCh38
rs979438692 Chr.15: 70655818-70655818 on Build GRCh38
rs986340483 Chr.15: 70655706-70655706 on Build GRCh38
rs981582011 Chr.15: 70655942-70655942 on Build GRCh38
rs1021064890 Chr.15: 70655807-70655807 on Build GRCh38
rs71152306 Chr.15: 70655779-70655779 on Build GRCh38
rs574208509 Chr.15: 70655859-70655859 on Build GRCh38
rs1028389714 Chr.15: 70655987-70655987 on Build GRCh38
rs949943559 Chr.15: 70655930-70655930 on Build GRCh38
rs919053375 Chr.15: 70655944-70655944 on Build GRCh38
rs998020336 Chr.15: 70655781-70655781 on Build GRCh38
rs997848877 Chr.15: 70655689-70655689 on Build GRCh38
rs538685435 Chr.15: 70655704-70655704 on Build GRCh38
rs181691919 Chr.15: 70656028-70656028 on Build GRCh38
rs1021453371 Chr.15: 70656054-70656055 on Build GRCh38
rs181191060 Chr.15: 70655799-70655799 on Build GRCh38
rs767361500 Chr.15: 70655652-70655652 on Build GRCh38
rs965005725 Chr.15: 70655669-70655669 on Build GRCh38
rs888063091 Chr.15: 70655708-70655708 on Build GRCh38
rs398027823 Chr.15: 70655781-70655781 on Build GRCh38
rs777532110 Chr.15: 70655984-70655984 on Build GRCh38
rs184837742 Chr.15: 70655824-70655824 on Build GRCh38
rs915743282 Chr.15: 70655922-70655922 on Build GRCh38
rs1896430 Chr.15: 70656111-70656111 on Build GRCh38
rs189012551 Chr.15: 70655985-70655985 on Build GRCh38
rs938720605 Chr.15: 70655809-70655809 on Build GRCh38
rs1016440644 Chr.15: 70656049-70656049 on Build GRCh38
rs370819736 Chr.15: 70655692-70655692 on Build GRCh38
rs998777235 Chr.15: 70655872-70655872 on Build GRCh38
rs557296507 Chr.15: 70655837-70655837 on Build GRCh38
rs904324587 Chr.15: 70655854-70655854 on Build GRCh38
rs1042910074 Chr.15: 70655853-70655853 on Build GRCh38
rs1028107879 Chr.15: 70655702-70655702 on Build GRCh38
rs949860297 Chr.15: 70655662-70655662 on Build GRCh38
rs901083054 Chr.15: 70655983-70655983 on Build GRCh38
rs963474823 Chr.15: 70656090-70656090 on Build GRCh38
rs901993360 Chr.15: 70655995-70655995 on Build GRCh38
rs953917167 Chr.15: 70655705-70655705 on Build GRCh38
rs1026555875 Chr.15: 70656114-70656114 on Build GRCh38
rs972254101 Chr.15: 70655766-70655766 on Build GRCh38
rs963610347 Chr.15: 70655747-70655747 on Build GRCh38
rs889561353 Chr.15: 70656023-70656023 on Build GRCh38
rs943619219 Chr.15: 70655986-70655988 on Build GRCh38
rs1032326108 Chr.15: 70656085-70656085 on Build GRCh38
rs1000784435 Chr.15: 70656062-70656062 on Build GRCh38
rs1053064825 Chr.15: 70655896-70655896 on Build GRCh38
rs1007697272 Chr.15: 70655750-70655750 on Build GRCh38
rs34923449 Chr.15: 70655707-70655707 on Build GRCh38
rs991353434 Chr.15: 70656125-70656125 on Build GRCh38
rs928947422 Chr.15: 70655965-70655965 on Build GRCh38

View Full Product Details