Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89316938-89317457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
UniGene
Hs.513126
Gene Chromosome Location
Chr.15: 89243954-89317132 on Build GRCh38
Transcripts
NM_001113378.1 NM_018193.2 XM_011521756.2 XM_011521757.2 XM_011521764.2 XM_011521767.2 NM_001126131.1 NM_002693.2

Gene Symbol
POLG
Entrez Gene ID
5428
Gene Name
DNA polymerase gamma, catalytic subunit
Gene Aliases
MDP1, MIRAS, MTDPS4A, MTDPS4B, PEO, POLG1, POLGA, SANDO, SCAE
UniGene
Hs.706868
Gene Chromosome Location
Chr.15: 89316305-89334795 on Build GRCh38
Transcripts
NM_001113378.1 NM_018193.2 XM_011521756.2 XM_011521757.2 XM_011521764.2 XM_011521767.2 NM_001126131.1 NM_002693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653350 c.3581C>T p.T1194I Chr.15: 89317438-89317438 on Build GRCh38
5750700 c.3631G>T p.G1211W Chr.15: 89317388-89317388 on Build GRCh38
7298521 c.3583A>G p.M1195V Chr.15: 89317436-89317436 on Build GRCh38
6752086 c.3641A>G p.Q1214R Chr.15: 89317378-89317378 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765344513 Chr.15: 89317433-89317433 on Build GRCh38
rs563913302 Chr.15: 89317160-89317160 on Build GRCh38
rs986944201 Chr.15: 89317301-89317301 on Build GRCh38
rs974235750 Chr.15: 89317076-89317076 on Build GRCh38
rs929400394 Chr.15: 89317305-89317305 on Build GRCh38
rs781256643 Chr.15: 89317379-89317379 on Build GRCh38
rs1045761501 Chr.15: 89317098-89317098 on Build GRCh38
rs780754049 Chr.15: 89317041-89317044 on Build GRCh38
rs756156336 Chr.15: 89317380-89317380 on Build GRCh38
rs912740266 Chr.15: 89317302-89317302 on Build GRCh38
rs766919082 Chr.15: 89317421-89317421 on Build GRCh38
rs765549612 Chr.15: 89317364-89317364 on Build GRCh38
rs749123350 Chr.15: 89317383-89317383 on Build GRCh38
rs1007946732 Chr.15: 89317327-89317327 on Build GRCh38
rs139562274 Chr.15: 89317389-89317389 on Build GRCh38
rs751149954 Chr.15: 89316991-89316991 on Build GRCh38
rs747627023 Chr.15: 89317391-89317391 on Build GRCh38
rs545559636 Chr.15: 89317362-89317362 on Build GRCh38
rs771520663 Chr.15: 89317396-89317396 on Build GRCh38
rs748855817 Chr.15: 89317331-89317331 on Build GRCh38
rs935080245 Chr.15: 89317086-89317086 on Build GRCh38
rs113994101 Chr.15: 89317388-89317389 on Build GRCh38
rs908234860 Chr.15: 89317038-89317038 on Build GRCh38
rs750747321 Chr.15: 89317128-89317128 on Build GRCh38
rs1012689797 Chr.15: 89317156-89317156 on Build GRCh38
rs2307443 Chr.15: 89317422-89317422 on Build GRCh38
rs917582933 Chr.15: 89317245-89317245 on Build GRCh38
rs1004087702 Chr.15: 89317248-89317248 on Build GRCh38
rs1000909206 Chr.15: 89317138-89317138 on Build GRCh38
rs768181189 Chr.15: 89317388-89317388 on Build GRCh38
rs1061316 Chr.15: 89317119-89317119 on Build GRCh38
rs886051519 Chr.15: 89317006-89317007 on Build GRCh38
rs993992126 Chr.15: 89317142-89317142 on Build GRCh38
rs1045501252 Chr.15: 89317368-89317368 on Build GRCh38
rs189937835 Chr.15: 89317134-89317134 on Build GRCh38
rs115758754 Chr.15: 89317011-89317011 on Build GRCh38
rs568031019 Chr.15: 89317221-89317221 on Build GRCh38
rs192368879 Chr.15: 89317284-89317284 on Build GRCh38
rs932808252 Chr.15: 89317093-89317093 on Build GRCh38
rs990138732 Chr.15: 89317005-89317015 on Build GRCh38
rs942296016 Chr.15: 89317290-89317290 on Build GRCh38
rs781311846 Chr.15: 89317418-89317418 on Build GRCh38
rs956781405 Chr.15: 89317376-89317376 on Build GRCh38
rs41545724 Chr.15: 89317261-89317261 on Build GRCh38
rs886043241 Chr.15: 89317406-89317407 on Build GRCh38
rs386786249 Chr.15: 89317118-89317119 on Build GRCh38
rs970516573 Chr.15: 89317423-89317423 on Build GRCh38
rs1018230135 Chr.15: 89317155-89317155 on Build GRCh38
rs1860022 Chr.15: 89317150-89317150 on Build GRCh38
rs56206444 Chr.15: 89317318-89317318 on Build GRCh38
rs890446286 Chr.15: 89317323-89317323 on Build GRCh38
rs3176236 Chr.15: 89317316-89317316 on Build GRCh38
rs2307454 Chr.15: 89317328-89317328 on Build GRCh38
rs886051520 Chr.15: 89317002-89317003 on Build GRCh38
rs987789430 Chr.15: 89317089-89317089 on Build GRCh38
rs772737979 Chr.15: 89317405-89317405 on Build GRCh38
rs367876885 Chr.15: 89317345-89317345 on Build GRCh38
rs1055159712 Chr.15: 89317344-89317344 on Build GRCh38
rs3176238 Chr.15: 89317196-89317196 on Build GRCh38
rs886051521 Chr.15: 89317073-89317073 on Build GRCh38
rs963971744 Chr.15: 89317194-89317194 on Build GRCh38
rs758720361 Chr.15: 89317435-89317435 on Build GRCh38
rs761373092 Chr.15: 89317340-89317340 on Build GRCh38
rs751695170 Chr.15: 89317350-89317351 on Build GRCh38
rs949657346 Chr.15: 89317095-89317095 on Build GRCh38
rs565584613 Chr.15: 89317264-89317264 on Build GRCh38
rs780470425 Chr.15: 89317147-89317147 on Build GRCh38
rs778756546 Chr.15: 89317221-89317222 on Build GRCh38
rs941078547 Chr.15: 89317069-89317069 on Build GRCh38
rs572636339 Chr.15: 89317358-89317358 on Build GRCh38
rs1051297210 Chr.15: 89317113-89317113 on Build GRCh38
rs754053297 Chr.15: 89317078-89317078 on Build GRCh38
rs987163246 Chr.15: 89317006-89317006 on Build GRCh38
rs540371794 Chr.15: 89317019-89317019 on Build GRCh38
rs1030756836 Chr.15: 89316982-89316986 on Build GRCh38
rs375815816 Chr.15: 89317330-89317330 on Build GRCh38
rs376292486 Chr.15: 89317287-89317287 on Build GRCh38
rs761103159 Chr.15: 89317409-89317409 on Build GRCh38
rs973803189 Chr.15: 89317285-89317285 on Build GRCh38
rs921002510 Chr.15: 89317289-89317289 on Build GRCh38
rs752712345 Chr.15: 89317371-89317371 on Build GRCh38
rs753020294 Chr.15: 89317434-89317434 on Build GRCh38
rs1024158080 Chr.15: 89317009-89317009 on Build GRCh38
rs182146869 Chr.15: 89317206-89317206 on Build GRCh38
rs758587842 Chr.15: 89317373-89317373 on Build GRCh38
rs570733403 Chr.15: 89317304-89317304 on Build GRCh38
rs761191143 Chr.15: 89317350-89317350 on Build GRCh38
rs74842339 Chr.15: 89317351-89317351 on Build GRCh38
rs560246181 Chr.15: 89317121-89317121 on Build GRCh38
rs907052263 Chr.15: 89317370-89317370 on Build GRCh38
rs187943166 Chr.15: 89317232-89317232 on Build GRCh38
rs905356629 Chr.15: 89317110-89317110 on Build GRCh38
rs1008135462 Chr.15: 89317153-89317153 on Build GRCh38
rs773676697 Chr.15: 89317339-89317339 on Build GRCh38
rs116194994 Chr.15: 89317080-89317080 on Build GRCh38
rs768360551 Chr.15: 89317333-89317333 on Build GRCh38
rs2307427 Chr.15: 89317297-89317297 on Build GRCh38
rs1860021 Chr.15: 89317118-89317118 on Build GRCh38
rs768009806 Chr.15: 89317342-89317342 on Build GRCh38
rs988334790 Chr.15: 89317208-89317208 on Build GRCh38
rs755054845 Chr.15: 89317389-89317390 on Build GRCh38

View Full Product Details