Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90271102-90271614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGRN
Entrez Gene ID
51335
Gene Name
neugrin, neurite outgrowth associated
Gene Aliases
DSC92
UniGene
Hs.135471
Gene Chromosome Location
Chr.15: 90265663-90272211 on Build GRCh38
Transcripts
NM_001033088.1 NR_028052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6042911 c.324T>C p.A108A Chr.15: 90271236-90271236 on Build GRCh38
53775 c.374_375insCAGA p.T126fs*16 Chr.15: 90271502-90271503 on Build GRCh38
4833486 c.287C>T p.S96F Chr.15: 90271415-90271415 on Build GRCh38
5920855 c.79C>T p.P27S Chr.15: 90271207-90271207 on Build GRCh38
265591 c.503_512del10 p.V169fs*20 Chr.15: 90271415-90271424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150300063 Chr.15: 90271281-90271281 on Build GRCh38
rs772969264 Chr.15: 90271392-90271392 on Build GRCh38
rs749675944 Chr.15: 90271168-90271168 on Build GRCh38
rs755513289 Chr.15: 90271198-90271198 on Build GRCh38
rs574771556 Chr.15: 90271473-90271473 on Build GRCh38
rs760873705 Chr.15: 90271273-90271273 on Build GRCh38
rs761458046 Chr.15: 90271298-90271298 on Build GRCh38
rs781451166 Chr.15: 90271555-90271555 on Build GRCh38
rs756225182 Chr.15: 90271565-90271565 on Build GRCh38
rs772915506 Chr.15: 90271470-90271470 on Build GRCh38
rs961589602 Chr.15: 90271377-90271377 on Build GRCh38
rs148169177 Chr.15: 90271560-90271560 on Build GRCh38
rs755710210 Chr.15: 90271322-90271322 on Build GRCh38
rs11547112 Chr.15: 90271267-90271267 on Build GRCh38
rs1023438245 Chr.15: 90271359-90271361 on Build GRCh38
rs763852781 Chr.15: 90271187-90271187 on Build GRCh38
rs777966118 Chr.15: 90271427-90271427 on Build GRCh38
rs922376635 Chr.15: 90271185-90271185 on Build GRCh38
rs758843759 Chr.15: 90271419-90271419 on Build GRCh38
rs764622782 Chr.15: 90271515-90271515 on Build GRCh38
rs749802564 Chr.15: 90271202-90271202 on Build GRCh38
rs762108295 Chr.15: 90271177-90271177 on Build GRCh38
rs753299210 Chr.15: 90271507-90271507 on Build GRCh38
rs939248040 Chr.15: 90271492-90271493 on Build GRCh38
rs763979131 Chr.15: 90271288-90271288 on Build GRCh38
rs772087440 Chr.15: 90271179-90271179 on Build GRCh38
rs932786281 Chr.15: 90271424-90271424 on Build GRCh38
rs778554885 Chr.15: 90271494-90271495 on Build GRCh38
rs759187525 Chr.15: 90271493-90271493 on Build GRCh38
rs1015827230 Chr.15: 90271376-90271376 on Build GRCh38
rs143514320 Chr.15: 90271188-90271188 on Build GRCh38
rs775453400 Chr.15: 90271282-90271282 on Build GRCh38
rs749337538 Chr.15: 90271577-90271577 on Build GRCh38
rs202223531 Chr.15: 90271549-90271549 on Build GRCh38
rs753936358 Chr.15: 90271417-90271417 on Build GRCh38
rs372378482 Chr.15: 90271378-90271378 on Build GRCh38
rs772347544 Chr.15: 90271260-90271260 on Build GRCh38
rs748015628 Chr.15: 90271465-90271465 on Build GRCh38
rs773376624 Chr.15: 90271180-90271180 on Build GRCh38
rs758682740 Chr.15: 90271359-90271359 on Build GRCh38
rs200072806 Chr.15: 90271183-90271183 on Build GRCh38
rs745952128 Chr.15: 90271443-90271443 on Build GRCh38
rs373465100 Chr.15: 90271164-90271164 on Build GRCh38
rs767205069 Chr.15: 90271311-90271311 on Build GRCh38
rs367968713 Chr.15: 90271318-90271318 on Build GRCh38
rs771944806 Chr.15: 90271469-90271469 on Build GRCh38
rs766093649 Chr.15: 90271402-90271402 on Build GRCh38
rs747511189 Chr.15: 90271511-90271511 on Build GRCh38
rs769140302 Chr.15: 90271169-90271169 on Build GRCh38
rs375937415 Chr.15: 90271184-90271184 on Build GRCh38
rs200330412 Chr.15: 90271391-90271391 on Build GRCh38
rs774331317 Chr.15: 90271459-90271459 on Build GRCh38
rs150627206 Chr.15: 90271566-90271566 on Build GRCh38
rs778112125 Chr.15: 90271361-90271361 on Build GRCh38
rs745862168 Chr.15: 90271516-90271517 on Build GRCh38
rs757559649 Chr.15: 90271544-90271544 on Build GRCh38
rs772218656 Chr.15: 90271423-90271423 on Build GRCh38
rs779239381 Chr.15: 90271358-90271358 on Build GRCh38
rs984390722 Chr.15: 90271166-90271166 on Build GRCh38
rs548014673 Chr.15: 90271167-90271167 on Build GRCh38
rs771931722 Chr.15: 90271591-90271591 on Build GRCh38
rs11547106 Chr.15: 90271395-90271395 on Build GRCh38
rs745328987 Chr.15: 90271157-90271157 on Build GRCh38
rs116320466 Chr.15: 90271346-90271346 on Build GRCh38
rs201227519 Chr.15: 90271372-90271372 on Build GRCh38
rs748878820 Chr.15: 90271333-90271334 on Build GRCh38
rs533701618 Chr.15: 90271504-90271504 on Build GRCh38
rs751377639 Chr.15: 90271297-90271297 on Build GRCh38
rs766130378 Chr.15: 90271480-90271480 on Build GRCh38
rs779394011 Chr.15: 90271220-90271220 on Build GRCh38
rs529788409 Chr.15: 90271141-90271141 on Build GRCh38
rs764812147 Chr.15: 90271497-90271497 on Build GRCh38
rs1003409078 Chr.15: 90271312-90271312 on Build GRCh38
rs751874414 Chr.15: 90271541-90271541 on Build GRCh38
rs146882162 Chr.15: 90271352-90271352 on Build GRCh38
rs780721618 Chr.15: 90271143-90271143 on Build GRCh38
rs1012468635 Chr.15: 90271588-90271588 on Build GRCh38
rs768594996 Chr.15: 90271385-90271385 on Build GRCh38
rs144820175 Chr.15: 90271522-90271522 on Build GRCh38
rs760467158 Chr.15: 90271175-90271177 on Build GRCh38
rs765908627 Chr.15: 90271415-90271415 on Build GRCh38
rs752178638 Chr.15: 90271149-90271151 on Build GRCh38
rs117040512 Chr.15: 90271201-90271201 on Build GRCh38
rs370598919 Chr.15: 90271196-90271196 on Build GRCh38
rs753424795 Chr.15: 90271412-90271412 on Build GRCh38
rs199831326 Chr.15: 90271286-90271286 on Build GRCh38
rs1056346542 Chr.15: 90271505-90271505 on Build GRCh38
rs745898763 Chr.15: 90271373-90271373 on Build GRCh38
rs143871202 Chr.15: 90271327-90271327 on Build GRCh38
rs760217610 Chr.15: 90271401-90271401 on Build GRCh38
rs747175405 Chr.15: 90271365-90271365 on Build GRCh38
rs141815208 Chr.15: 90271561-90271561 on Build GRCh38
rs375789739 Chr.15: 90271355-90271355 on Build GRCh38
rs777695911 Chr.15: 90271592-90271592 on Build GRCh38
rs757181730 Chr.15: 90271473-90271474 on Build GRCh38
rs77207433 Chr.15: 90271501-90271501 on Build GRCh38
rs370660196 Chr.15: 90271438-90271438 on Build GRCh38
rs774208396 Chr.15: 90271388-90271388 on Build GRCh38
rs777578191 Chr.15: 90271301-90271302 on Build GRCh38
rs374229382 Chr.15: 90271208-90271208 on Build GRCh38
rs566973503 Chr.15: 90271472-90271472 on Build GRCh38
rs139094960 Chr.15: 90271191-90271191 on Build GRCh38
rs776414747 Chr.15: 90271488-90271488 on Build GRCh38
rs143233045 Chr.15: 90271456-90271456 on Build GRCh38
rs11073922 Chr.15: 90271432-90271432 on Build GRCh38
rs376127405 Chr.15: 90271182-90271182 on Build GRCh38
rs773238382 Chr.15: 90271262-90271262 on Build GRCh38

View Full Product Details