Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90617898-90618408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1708548 c.631C>T p.P211S Chr.15: 90617900-90617900 on Build GRCh38
966511 c.662C>T p.P221L Chr.15: 90617931-90617931 on Build GRCh38
4822972 c.646A>G p.N216D Chr.15: 90617915-90617915 on Build GRCh38
7248319 c.663G>A p.P221P Chr.15: 90617932-90617932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368205917 Chr.15: 90617997-90617997 on Build GRCh38
rs749351359 Chr.15: 90617972-90617972 on Build GRCh38
rs973713477 Chr.15: 90618114-90618114 on Build GRCh38
rs773174310 Chr.15: 90617995-90617995 on Build GRCh38
rs200894306 Chr.15: 90618206-90618206 on Build GRCh38
rs778306323 Chr.15: 90618331-90618331 on Build GRCh38
rs576311286 Chr.15: 90618141-90618141 on Build GRCh38
rs1007401145 Chr.15: 90618239-90618239 on Build GRCh38
rs980531259 Chr.15: 90618139-90618139 on Build GRCh38
rs1049360586 Chr.15: 90618228-90618228 on Build GRCh38
rs11372294 Chr.15: 90618206-90618207 on Build GRCh38
rs543085250 Chr.15: 90617950-90617950 on Build GRCh38
rs1013959423 Chr.15: 90618301-90618302 on Build GRCh38
rs1039776637 Chr.15: 90618308-90618308 on Build GRCh38
rs776413991 Chr.15: 90618003-90618003 on Build GRCh38
rs72224908 Chr.15: 90618216-90618217 on Build GRCh38
rs564525059 Chr.15: 90618097-90618097 on Build GRCh38
rs1000431527 Chr.15: 90618217-90618218 on Build GRCh38
rs60991614 Chr.15: 90618222-90618223 on Build GRCh38
rs3743406 Chr.15: 90618316-90618316 on Build GRCh38
rs915443286 Chr.15: 90618116-90618116 on Build GRCh38
rs144818109 Chr.15: 90617959-90617959 on Build GRCh38
rs919148684 Chr.15: 90618216-90618216 on Build GRCh38
rs927722997 Chr.15: 90618140-90618140 on Build GRCh38
rs919500317 Chr.15: 90618120-90618120 on Build GRCh38
rs116154571 Chr.15: 90618075-90618075 on Build GRCh38
rs930604719 Chr.15: 90618223-90618223 on Build GRCh38
rs891699993 Chr.15: 90618231-90618231 on Build GRCh38
rs1018285557 Chr.15: 90618027-90618027 on Build GRCh38
rs543567307 Chr.15: 90618315-90618315 on Build GRCh38
rs942156657 Chr.15: 90618111-90618111 on Build GRCh38
rs1048908529 Chr.15: 90617971-90617971 on Build GRCh38
rs77748519 Chr.15: 90617932-90617932 on Build GRCh38
rs957165727 Chr.15: 90618347-90618347 on Build GRCh38
rs374979539 Chr.15: 90618002-90618002 on Build GRCh38
rs1039489670 Chr.15: 90618187-90618187 on Build GRCh38
rs781120011 Chr.15: 90618077-90618077 on Build GRCh38
rs146224437 Chr.15: 90617939-90617939 on Build GRCh38
rs755295040 Chr.15: 90617980-90617980 on Build GRCh38
rs998194766 Chr.15: 90618065-90618065 on Build GRCh38
rs979823688 Chr.15: 90618085-90618085 on Build GRCh38
rs1036003881 Chr.15: 90618151-90618151 on Build GRCh38
rs139851694 Chr.15: 90617945-90617945 on Build GRCh38
rs990038066 Chr.15: 90618105-90618105 on Build GRCh38
rs769707256 Chr.15: 90618235-90618235 on Build GRCh38
rs904365165 Chr.15: 90618214-90618215 on Build GRCh38
rs969537926 Chr.15: 90618119-90618119 on Build GRCh38
rs149408148 Chr.15: 90617954-90617954 on Build GRCh38
rs965332617 Chr.15: 90618046-90618046 on Build GRCh38
rs969806577 Chr.15: 90618078-90618078 on Build GRCh38
rs758226599 Chr.15: 90617931-90617931 on Build GRCh38

View Full Product Details