Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 91251684-91252224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SV2B
Entrez Gene ID
9899
Gene Name
synaptic vesicle glycoprotein 2B
Gene Aliases
HsT19680
UniGene
Hs.21754
Gene Chromosome Location
Chr.15: 91099588-91301309 on Build GRCh38
Transcripts
NM_001167580.1 NM_001323031.1 NM_001323032.1 NM_001323033.1 NM_001323034.1 NM_001323036.1 NM_001323037.1 NM_001323038.1 NM_001323039.1 NM_001323040.1 NM_014848.5 XM_005254998.3 XM_017022761.1 XM_017022762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608844 c.485C>A p.A162D Chr.15: 91251852-91251852 on Build GRCh38
7307776 c.519A>G p.G173G Chr.15: 91251886-91251886 on Build GRCh38
6484824 c.632G>C p.G211A Chr.15: 91251999-91251999 on Build GRCh38
1608845 c.486C>A p.A162A Chr.15: 91251853-91251853 on Build GRCh38
4128594 c.606C>G p.L202L Chr.15: 91251973-91251973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999792123 Chr.15: 91252020-91252020 on Build GRCh38
rs17693706 Chr.15: 91251973-91251973 on Build GRCh38
rs1042000586 Chr.15: 91251826-91251826 on Build GRCh38
rs566828523 Chr.15: 91251965-91251965 on Build GRCh38
rs992939017 Chr.15: 91252079-91252079 on Build GRCh38
rs948772859 Chr.15: 91252129-91252129 on Build GRCh38
rs767247777 Chr.15: 91251980-91251980 on Build GRCh38
rs772911915 Chr.15: 91252043-91252043 on Build GRCh38
rs939959719 Chr.15: 91251946-91251946 on Build GRCh38
rs373474055 Chr.15: 91252049-91252049 on Build GRCh38
rs945039904 Chr.15: 91252001-91252001 on Build GRCh38
rs746131718 Chr.15: 91252025-91252025 on Build GRCh38
rs990093560 Chr.15: 91251807-91251807 on Build GRCh38
rs771765158 Chr.15: 91252041-91252041 on Build GRCh38
rs567477143 Chr.15: 91252158-91252158 on Build GRCh38
rs148460168 Chr.15: 91251916-91251916 on Build GRCh38
rs146271108 Chr.15: 91251846-91251846 on Build GRCh38
rs915668107 Chr.15: 91251907-91251907 on Build GRCh38
rs778818870 Chr.15: 91251823-91251823 on Build GRCh38
rs372020741 Chr.15: 91251851-91251851 on Build GRCh38
rs757965786 Chr.15: 91252015-91252015 on Build GRCh38
rs766155861 Chr.15: 91251993-91251993 on Build GRCh38
rs894680548 Chr.15: 91252125-91252125 on Build GRCh38
rs781199439 Chr.15: 91251812-91251812 on Build GRCh38
rs769309827 Chr.15: 91252033-91252033 on Build GRCh38
rs1015476941 Chr.15: 91251924-91251924 on Build GRCh38
rs201626625 Chr.15: 91252013-91252013 on Build GRCh38
rs776050433 Chr.15: 91251950-91251950 on Build GRCh38
rs547706465 Chr.15: 91251938-91251938 on Build GRCh38
rs771169038 Chr.15: 91251939-91251939 on Build GRCh38
rs555716365 Chr.15: 91252149-91252149 on Build GRCh38
rs866862427 Chr.15: 91251820-91251820 on Build GRCh38
rs752269407 Chr.15: 91251788-91251788 on Build GRCh38
rs758503750 Chr.15: 91251931-91251931 on Build GRCh38
rs79725910 Chr.15: 91251894-91251894 on Build GRCh38
rs369772157 Chr.15: 91251904-91251904 on Build GRCh38
rs921030364 Chr.15: 91251790-91251790 on Build GRCh38
rs370316095 Chr.15: 91251809-91251809 on Build GRCh38
rs369065381 Chr.15: 91251957-91251957 on Build GRCh38
rs555419265 Chr.15: 91251729-91251729 on Build GRCh38
rs757364690 Chr.15: 91252016-91252016 on Build GRCh38
rs764598588 Chr.15: 91251776-91251776 on Build GRCh38
rs1015550850 Chr.15: 91251751-91251751 on Build GRCh38
rs368302511 Chr.15: 91251805-91251805 on Build GRCh38
rs760507104 Chr.15: 91251986-91251986 on Build GRCh38
rs756099651 Chr.15: 91251816-91251816 on Build GRCh38
rs962523834 Chr.15: 91251723-91251723 on Build GRCh38
rs77204250 Chr.15: 91251921-91251921 on Build GRCh38
rs944904093 Chr.15: 91251824-91251824 on Build GRCh38
rs866100604 Chr.15: 91251806-91251806 on Build GRCh38
rs770360729 Chr.15: 91251893-91251893 on Build GRCh38
rs377576894 Chr.15: 91251795-91251795 on Build GRCh38
rs759983195 Chr.15: 91251879-91251879 on Build GRCh38
rs750319525 Chr.15: 91251813-91251813 on Build GRCh38
rs369745123 Chr.15: 91252040-91252040 on Build GRCh38
rs754888193 Chr.15: 91251998-91251998 on Build GRCh38
rs749746440 Chr.15: 91252031-91252031 on Build GRCh38
rs201523826 Chr.15: 91251850-91251850 on Build GRCh38
rs144587459 Chr.15: 91251800-91251800 on Build GRCh38
rs748229273 Chr.15: 91251836-91251836 on Build GRCh38
rs774010844 Chr.15: 91251966-91251966 on Build GRCh38
rs1055023184 Chr.15: 91252061-91252061 on Build GRCh38
rs756494343 Chr.15: 91252029-91252029 on Build GRCh38
rs12909119 Chr.15: 91251943-91251943 on Build GRCh38
rs534889358 Chr.15: 91252188-91252188 on Build GRCh38
rs143979505 Chr.15: 91251917-91251917 on Build GRCh38
rs901161078 Chr.15: 91251780-91251780 on Build GRCh38
rs745434724 Chr.15: 91251755-91251755 on Build GRCh38
rs775037521 Chr.15: 91252035-91252035 on Build GRCh38
rs750150251 Chr.15: 91251981-91251981 on Build GRCh38
rs753780491 Chr.15: 91251925-91251925 on Build GRCh38
rs888399144 Chr.15: 91251720-91251721 on Build GRCh38
rs530472457 Chr.15: 91251913-91251913 on Build GRCh38
rs574805145 Chr.15: 91251949-91251949 on Build GRCh38
rs765964793 Chr.15: 91251905-91251905 on Build GRCh38
rs764114595 Chr.15: 91252008-91252008 on Build GRCh38
rs769277305 Chr.15: 91251750-91251750 on Build GRCh38
rs200150672 Chr.15: 91251932-91251932 on Build GRCh38
rs61731608 Chr.15: 91251997-91251997 on Build GRCh38
rs781367448 Chr.15: 91252023-91252023 on Build GRCh38
rs538490934 Chr.15: 91251847-91251847 on Build GRCh38
rs781492337 Chr.15: 91251940-91251940 on Build GRCh38
rs536730683 Chr.15: 91251906-91251906 on Build GRCh38
rs537090850 Chr.15: 91252143-91252143 on Build GRCh38
rs977649315 Chr.15: 91252002-91252002 on Build GRCh38

View Full Product Details