Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92465178-92465673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST8SIA2
Entrez Gene ID
8128
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
HsT19690, SIAT8B, ST8SIA-II, STX
UniGene
Hs.302341
Gene Chromosome Location
Chr.15: 92393910-92468728 on Build GRCh38
Transcripts
NM_006011.3 XM_017022642.1 XM_017022643.1

SNP Detail

SNP ID Chromosome Location
rs528459103 Chr.15: 92465651-92465651 on Build GRCh38
rs911565656 Chr.15: 92465629-92465629 on Build GRCh38
rs899349991 Chr.15: 92465375-92465375 on Build GRCh38
rs144973310 Chr.15: 92465510-92465511 on Build GRCh38
rs975563565 Chr.15: 92465328-92465328 on Build GRCh38
rs188486480 Chr.15: 92465548-92465548 on Build GRCh38
rs562673541 Chr.15: 92465248-92465248 on Build GRCh38
rs780942195 Chr.15: 92465346-92465346 on Build GRCh38
rs767023383 Chr.15: 92465625-92465625 on Build GRCh38
rs565532140 Chr.15: 92465300-92465300 on Build GRCh38
rs988431733 Chr.15: 92465367-92465367 on Build GRCh38
rs8035760 Chr.15: 92465242-92465242 on Build GRCh38
rs899907332 Chr.15: 92465582-92465582 on Build GRCh38
rs191844716 Chr.15: 92465571-92465571 on Build GRCh38
rs555486851 Chr.15: 92465260-92465260 on Build GRCh38
rs1022354135 Chr.15: 92465543-92465543 on Build GRCh38
rs913520704 Chr.15: 92465380-92465380 on Build GRCh38
rs1035017920 Chr.15: 92465490-92465490 on Build GRCh38
rs943110093 Chr.15: 92465311-92465311 on Build GRCh38
rs887152734 Chr.15: 92465213-92465213 on Build GRCh38
rs1051449183 Chr.15: 92465298-92465298 on Build GRCh38
rs116854288 Chr.15: 92465615-92465615 on Build GRCh38
rs556979997 Chr.15: 92465212-92465212 on Build GRCh38
rs534649532 Chr.15: 92465315-92465315 on Build GRCh38
rs924521917 Chr.15: 92465241-92465241 on Build GRCh38
rs1037483705 Chr.15: 92465496-92465496 on Build GRCh38
rs886716832 Chr.15: 92465468-92465468 on Build GRCh38
rs34214859 Chr.15: 92465436-92465436 on Build GRCh38
rs543754556 Chr.15: 92465558-92465558 on Build GRCh38
rs1039209033 Chr.15: 92465639-92465639 on Build GRCh38
rs1012747197 Chr.15: 92465528-92465528 on Build GRCh38
rs1048349363 Chr.15: 92465465-92465465 on Build GRCh38
rs959332303 Chr.15: 92465503-92465503 on Build GRCh38
rs1032542505 Chr.15: 92465329-92465329 on Build GRCh38
rs139771950 Chr.15: 92465459-92465459 on Build GRCh38
rs1041468634 Chr.15: 92465572-92465572 on Build GRCh38
rs558061791 Chr.15: 92465438-92465438 on Build GRCh38
rs949089332 Chr.15: 92465452-92465452 on Build GRCh38
rs770951764 Chr.15: 92465559-92465559 on Build GRCh38
rs1047640773 Chr.15: 92465512-92465512 on Build GRCh38
rs958270027 Chr.15: 92465345-92465345 on Build GRCh38
rs61648904 Chr.15: 92465620-92465620 on Build GRCh38
rs762987090 Chr.15: 92465460-92465460 on Build GRCh38
rs1038743590 Chr.15: 92465356-92465356 on Build GRCh38
rs1003748890 Chr.15: 92465476-92465476 on Build GRCh38
rs150056584 Chr.15: 92465634-92465634 on Build GRCh38

View Full Product Details