Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 94315432-94315980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCTP2
Entrez Gene ID
55784
Gene Name
multiple C2 and transmembrane domain containing 2
Gene Aliases
-
UniGene
Hs.33368
Gene Chromosome Location
Chr.15: 94231184-94483952 on Build GRCh38
Transcripts
NM_001159643.1 NM_018349.3 XM_005254955.3 XM_006720603.2 XM_011521770.1 XM_011521771.2 XM_011521772.2 XM_011521773.2 XM_011521774.2 XM_017022403.1 XM_017022404.1 XM_017022405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237938 c.534G>A p.P178P Chr.15: 94315534-94315534 on Build GRCh38
1608865 c.551G>T p.G184V Chr.15: 94315551-94315551 on Build GRCh38
4701247 c.578C>T p.A193V Chr.15: 94315578-94315578 on Build GRCh38
2141129 c.611G>A p.R204Q Chr.15: 94315611-94315611 on Build GRCh38
7198559 c.573delT p.F192fs*9 Chr.15: 94315573-94315573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751695616 Chr.15: 94315645-94315645 on Build GRCh38
rs145586255 Chr.15: 94315566-94315566 on Build GRCh38
rs780765420 Chr.15: 94315659-94315659 on Build GRCh38
rs931795445 Chr.15: 94315904-94315904 on Build GRCh38
rs36041166 Chr.15: 94315928-94315929 on Build GRCh38
rs8024196 Chr.15: 94315751-94315751 on Build GRCh38
rs542318098 Chr.15: 94315532-94315532 on Build GRCh38
rs73462089 Chr.15: 94315490-94315490 on Build GRCh38
rs542410047 Chr.15: 94315827-94315827 on Build GRCh38
rs913169805 Chr.15: 94315590-94315590 on Build GRCh38
rs545052593 Chr.15: 94315858-94315858 on Build GRCh38
rs294549 Chr.15: 94315839-94315839 on Build GRCh38
rs294550 Chr.15: 94315822-94315822 on Build GRCh38
rs12101974 Chr.15: 94315660-94315660 on Build GRCh38
rs150277459 Chr.15: 94315632-94315632 on Build GRCh38
rs764394024 Chr.15: 94315925-94315925 on Build GRCh38
rs200389318 Chr.15: 94315625-94315625 on Build GRCh38
rs750195259 Chr.15: 94315638-94315639 on Build GRCh38
rs1033491101 Chr.15: 94315796-94315796 on Build GRCh38
rs142093007 Chr.15: 94315539-94315539 on Build GRCh38
rs146431277 Chr.15: 94315579-94315579 on Build GRCh38
rs202000611 Chr.15: 94315584-94315584 on Build GRCh38
rs746330087 Chr.15: 94315605-94315605 on Build GRCh38
rs191343327 Chr.15: 94315665-94315665 on Build GRCh38
rs556790768 Chr.15: 94315820-94315820 on Build GRCh38
rs530476261 Chr.15: 94315879-94315879 on Build GRCh38
rs780700713 Chr.15: 94315731-94315731 on Build GRCh38
rs746278697 Chr.15: 94315657-94315657 on Build GRCh38
rs201530723 Chr.15: 94315608-94315608 on Build GRCh38
rs140081019 Chr.15: 94315564-94315564 on Build GRCh38
rs764699724 Chr.15: 94315553-94315553 on Build GRCh38
rs183487854 Chr.15: 94315748-94315748 on Build GRCh38
rs535339662 Chr.15: 94315749-94315749 on Build GRCh38
rs368719680 Chr.15: 94315638-94315638 on Build GRCh38
rs373612545 Chr.15: 94315478-94315478 on Build GRCh38
rs767257496 Chr.15: 94315626-94315626 on Build GRCh38
rs1017590621 Chr.15: 94315888-94315888 on Build GRCh38
rs372349402 Chr.15: 94315644-94315644 on Build GRCh38
rs776602463 Chr.15: 94315593-94315593 on Build GRCh38
rs760127443 Chr.15: 94315540-94315540 on Build GRCh38
rs763508120 Chr.15: 94315542-94315542 on Build GRCh38
rs1655455 Chr.15: 94315457-94315457 on Build GRCh38
rs758943172 Chr.15: 94315889-94315890 on Build GRCh38
rs1001653848 Chr.15: 94315760-94315760 on Build GRCh38
rs539052922 Chr.15: 94315721-94315721 on Build GRCh38
rs748329818 Chr.15: 94315482-94315482 on Build GRCh38
rs550997934 Chr.15: 94315903-94315903 on Build GRCh38
rs762438951 Chr.15: 94315619-94315619 on Build GRCh38
rs761740643 Chr.15: 94315523-94315524 on Build GRCh38
rs775166158 Chr.15: 94315549-94315549 on Build GRCh38
rs376414265 Chr.15: 94315631-94315631 on Build GRCh38
rs762648518 Chr.15: 94315561-94315561 on Build GRCh38
rs1024482559 Chr.15: 94315707-94315707 on Build GRCh38
rs763694321 Chr.15: 94315563-94315563 on Build GRCh38
rs143311478 Chr.15: 94315859-94315859 on Build GRCh38
rs528157570 Chr.15: 94315848-94315848 on Build GRCh38
rs767360399 Chr.15: 94315570-94315570 on Build GRCh38
rs562037950 Chr.15: 94315813-94315813 on Build GRCh38
rs980465652 Chr.15: 94315719-94315719 on Build GRCh38
rs980556611 Chr.15: 94315546-94315546 on Build GRCh38
rs758612626 Chr.15: 94315581-94315581 on Build GRCh38
rs559014388 Chr.15: 94315538-94315538 on Build GRCh38
rs543376197 Chr.15: 94315818-94315818 on Build GRCh38
rs1021794466 Chr.15: 94315948-94315948 on Build GRCh38
rs1048827542 Chr.15: 94315911-94315911 on Build GRCh38
rs769391620 Chr.15: 94315545-94315545 on Build GRCh38
rs758462632 Chr.15: 94315642-94315642 on Build GRCh38
rs747704454 Chr.15: 94315680-94315680 on Build GRCh38
rs940425385 Chr.15: 94315928-94315928 on Build GRCh38
rs374591712 Chr.15: 94315582-94315582 on Build GRCh38
rs758360143 Chr.15: 94315660-94315660 on Build GRCh38
rs1056360892 Chr.15: 94315783-94315783 on Build GRCh38
rs571941802 Chr.15: 94315578-94315578 on Build GRCh38
rs757524354 Chr.15: 94315648-94315648 on Build GRCh38
rs752962634 Chr.15: 94315873-94315873 on Build GRCh38
rs771760017 Chr.15: 94315686-94315686 on Build GRCh38
rs372217936 Chr.15: 94315534-94315534 on Build GRCh38
rs1003452248 Chr.15: 94315549-94315550 on Build GRCh38
rs768342134 Chr.15: 94315620-94315620 on Build GRCh38
rs767985238 Chr.15: 94315663-94315663 on Build GRCh38
rs926262764 Chr.15: 94315750-94315750 on Build GRCh38
rs765157540 Chr.15: 94315533-94315534 on Build GRCh38
rs756685766 Chr.15: 94315658-94315658 on Build GRCh38
rs148617412 Chr.15: 94315535-94315535 on Build GRCh38
rs147741772 Chr.15: 94315571-94315571 on Build GRCh38
rs781716197 Chr.15: 94315595-94315595 on Build GRCh38
rs781676290 Chr.15: 94315653-94315653 on Build GRCh38
rs747383664 Chr.15: 94315588-94315588 on Build GRCh38
rs146697301 Chr.15: 94315938-94315938 on Build GRCh38
rs775112883 Chr.15: 94315611-94315611 on Build GRCh38

View Full Product Details