Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 96331612-96332130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F2
Entrez Gene ID
7026
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
UniGene
Hs.347991
Gene Chromosome Location
Chr.15: 96325928-96340263 on Build GRCh38
Transcripts
NM_001145155.1 NM_021005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6374537 c.2T>A p.M1K Chr.15: 96332107-96332107 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573904102 Chr.15: 96331877-96331877 on Build GRCh38
rs866894374 Chr.15: 96331977-96331977 on Build GRCh38
rs947976341 Chr.15: 96331798-96331798 on Build GRCh38
rs879036736 Chr.15: 96331915-96331915 on Build GRCh38
rs1023600045 Chr.15: 96331973-96331973 on Build GRCh38
rs563105053 Chr.15: 96331886-96331886 on Build GRCh38
rs1046256360 Chr.15: 96331636-96331636 on Build GRCh38
rs1050126221 Chr.15: 96331917-96331917 on Build GRCh38
rs940444040 Chr.15: 96331646-96331653 on Build GRCh38
rs930296576 Chr.15: 96331903-96331903 on Build GRCh38
rs927814635 Chr.15: 96331821-96331821 on Build GRCh38
rs958267561 Chr.15: 96331766-96331766 on Build GRCh38
rs1050054182 Chr.15: 96332044-96332051 on Build GRCh38
rs370551634 Chr.15: 96331948-96331948 on Build GRCh38
rs112110943 Chr.15: 96331665-96331665 on Build GRCh38
rs571888761 Chr.15: 96331707-96331707 on Build GRCh38
rs573021579 Chr.15: 96331933-96331933 on Build GRCh38
rs920334841 Chr.15: 96332041-96332051 on Build GRCh38
rs544781079 Chr.15: 96331940-96331940 on Build GRCh38
rs1036000086 Chr.15: 96331658-96331659 on Build GRCh38
rs548346364 Chr.15: 96331950-96331950 on Build GRCh38
rs544777514 Chr.15: 96331848-96331848 on Build GRCh38
rs146141012 Chr.15: 96331896-96331896 on Build GRCh38
rs1027636201 Chr.15: 96331662-96331665 on Build GRCh38
rs982012477 Chr.15: 96331999-96332000 on Build GRCh38
rs898943409 Chr.15: 96331881-96331881 on Build GRCh38
rs890001575 Chr.15: 96331919-96331919 on Build GRCh38
rs965417933 Chr.15: 96331697-96331697 on Build GRCh38
rs867022143 Chr.15: 96331806-96331806 on Build GRCh38
rs954446673 Chr.15: 96331646-96331646 on Build GRCh38
rs560263849 Chr.15: 96332055-96332055 on Build GRCh38
rs1036503128 Chr.15: 96331863-96331866 on Build GRCh38
rs942904631 Chr.15: 96332059-96332059 on Build GRCh38
rs1041307591 Chr.15: 96332067-96332067 on Build GRCh38
rs1028552850 Chr.15: 96331638-96331638 on Build GRCh38
rs561241591 Chr.15: 96332009-96332009 on Build GRCh38
rs79544641 Chr.15: 96331649-96331649 on Build GRCh38
rs989531492 Chr.15: 96331767-96331767 on Build GRCh38
rs962172505 Chr.15: 96332022-96332022 on Build GRCh38
rs376766307 Chr.15: 96331692-96331692 on Build GRCh38
rs916496367 Chr.15: 96331770-96331770 on Build GRCh38
rs529676483 Chr.15: 96332035-96332035 on Build GRCh38
rs529811394 Chr.15: 96331930-96331930 on Build GRCh38
rs978592282 Chr.15: 96331726-96331729 on Build GRCh38
rs530820855 Chr.15: 96331966-96331966 on Build GRCh38
rs971956567 Chr.15: 96332040-96332042 on Build GRCh38
rs12916548 Chr.15: 96332028-96332028 on Build GRCh38
rs984272987 Chr.15: 96331655-96331655 on Build GRCh38
rs558091667 Chr.15: 96331687-96331687 on Build GRCh38
rs940428570 Chr.15: 96331849-96331850 on Build GRCh38
rs79964084 Chr.15: 96331819-96331819 on Build GRCh38
rs142499350 Chr.15: 96332041-96332041 on Build GRCh38
rs994536530 Chr.15: 96331658-96331658 on Build GRCh38
rs901353840 Chr.15: 96331932-96331932 on Build GRCh38
rs1031110504 Chr.15: 96331752-96331752 on Build GRCh38
rs1007154119 Chr.15: 96331926-96331926 on Build GRCh38
rs184941768 Chr.15: 96331980-96331980 on Build GRCh38
rs111316474 Chr.15: 96331691-96331692 on Build GRCh38
rs890075137 Chr.15: 96331685-96331685 on Build GRCh38
rs112438450 Chr.15: 96331703-96331703 on Build GRCh38
rs898845007 Chr.15: 96331658-96331661 on Build GRCh38
rs112575789 Chr.15: 96332000-96332000 on Build GRCh38

View Full Product Details