Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99729043-99729531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYSMD4
Entrez Gene ID
145748
Gene Name
LysM domain containing 4
Gene Aliases
-
UniGene
Hs.562568
Gene Chromosome Location
Chr.15: 99715033-99733561 on Build GRCh38
Transcripts
NM_001284417.1 NM_001284418.1 NM_001284419.1 NM_001284420.1 NM_001284421.1 NM_001284422.1 NM_152449.3 XM_006720397.3 XM_011521241.1 XM_011521242.2 XM_011521243.2 XM_011521244.1 XM_011521245.2 XM_017021926.1 XM_017021927.1 XM_017021928.1 XM_017021929.1 XM_017021930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6353357 c.542C>G p.A181G Chr.15: 99729475-99729475 on Build GRCh38
6445125 c.829G>A p.V277I Chr.15: 99729188-99729188 on Build GRCh38
4851780 c.756G>A p.L252L Chr.15: 99729258-99729258 on Build GRCh38
1239373 c.682C>T p.L228L Chr.15: 99729335-99729335 on Build GRCh38
5654301 c.606G>A p.P202P Chr.15: 99729408-99729408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764725681 Chr.15: 99729327-99729327 on Build GRCh38
rs745701553 Chr.15: 99729190-99729190 on Build GRCh38
rs180886521 Chr.15: 99729482-99729482 on Build GRCh38
rs776842951 Chr.15: 99729303-99729303 on Build GRCh38
rs774956829 Chr.15: 99729400-99729400 on Build GRCh38
rs6598263 Chr.15: 99729104-99729104 on Build GRCh38
rs2061007 Chr.15: 99729475-99729475 on Build GRCh38
rs745688203 Chr.15: 99729360-99729360 on Build GRCh38
rs781547211 Chr.15: 99729504-99729507 on Build GRCh38
rs962685946 Chr.15: 99729399-99729399 on Build GRCh38
rs770940073 Chr.15: 99729287-99729287 on Build GRCh38
rs143085338 Chr.15: 99729160-99729160 on Build GRCh38
rs141407567 Chr.15: 99729496-99729496 on Build GRCh38
rs998294243 Chr.15: 99729321-99729321 on Build GRCh38
rs772056491 Chr.15: 99729200-99729200 on Build GRCh38
rs142195798 Chr.15: 99729215-99729215 on Build GRCh38
rs766627816 Chr.15: 99729212-99729212 on Build GRCh38
rs756836859 Chr.15: 99729331-99729331 on Build GRCh38
rs915548760 Chr.15: 99729153-99729153 on Build GRCh38
rs201793909 Chr.15: 99729478-99729478 on Build GRCh38
rs750125554 Chr.15: 99729344-99729344 on Build GRCh38
rs755305352 Chr.15: 99729148-99729148 on Build GRCh38
rs139777997 Chr.15: 99729238-99729238 on Build GRCh38
rs911922150 Chr.15: 99729154-99729154 on Build GRCh38
rs367989718 Chr.15: 99729380-99729380 on Build GRCh38
rs750508131 Chr.15: 99729198-99729198 on Build GRCh38
rs751992671 Chr.15: 99729330-99729330 on Build GRCh38
rs775790912 Chr.15: 99729507-99729507 on Build GRCh38
rs148348952 Chr.15: 99729196-99729196 on Build GRCh38
rs746634883 Chr.15: 99729088-99729088 on Build GRCh38
rs765547128 Chr.15: 99729217-99729217 on Build GRCh38
rs964571026 Chr.15: 99729357-99729357 on Build GRCh38
rs761817697 Chr.15: 99729115-99729115 on Build GRCh38
rs760867625 Chr.15: 99729271-99729271 on Build GRCh38
rs780990711 Chr.15: 99729176-99729176 on Build GRCh38
rs763193728 Chr.15: 99729323-99729323 on Build GRCh38
rs947007733 Chr.15: 99729498-99729498 on Build GRCh38
rs777654091 Chr.15: 99729484-99729484 on Build GRCh38
rs766394415 Chr.15: 99729406-99729407 on Build GRCh38
rs769745125 Chr.15: 99729248-99729248 on Build GRCh38
rs572309722 Chr.15: 99729309-99729309 on Build GRCh38
rs1018836200 Chr.15: 99729311-99729314 on Build GRCh38
rs755123508 Chr.15: 99729485-99729486 on Build GRCh38
rs149795703 Chr.15: 99729394-99729394 on Build GRCh38
rs762878279 Chr.15: 99729199-99729199 on Build GRCh38
rs371804717 Chr.15: 99729174-99729174 on Build GRCh38
rs770040306 Chr.15: 99729385-99729385 on Build GRCh38
rs754716399 Chr.15: 99729363-99729363 on Build GRCh38
rs753151527 Chr.15: 99729224-99729224 on Build GRCh38
rs745922798 Chr.15: 99729381-99729381 on Build GRCh38
rs775970675 Chr.15: 99729313-99729313 on Build GRCh38
rs149471133 Chr.15: 99729136-99729136 on Build GRCh38
rs868584626 Chr.15: 99729329-99729329 on Build GRCh38
rs754267618 Chr.15: 99729141-99729141 on Build GRCh38
rs999613228 Chr.15: 99729220-99729220 on Build GRCh38
rs746011521 Chr.15: 99729490-99729490 on Build GRCh38
rs748011186 Chr.15: 99729374-99729374 on Build GRCh38
rs951165597 Chr.15: 99729097-99729097 on Build GRCh38
rs148658487 Chr.15: 99729409-99729409 on Build GRCh38
rs542617254 Chr.15: 99729372-99729372 on Build GRCh38
rs760892155 Chr.15: 99729133-99729133 on Build GRCh38
rs200602575 Chr.15: 99729449-99729449 on Build GRCh38
rs773397157 Chr.15: 99729201-99729201 on Build GRCh38
rs72760587 Chr.15: 99729430-99729430 on Build GRCh38
rs775566815 Chr.15: 99729389-99729389 on Build GRCh38
rs924323404 Chr.15: 99729461-99729461 on Build GRCh38
rs760324722 Chr.15: 99729439-99729439 on Build GRCh38
rs549657944 Chr.15: 99729103-99729103 on Build GRCh38
rs143078595 Chr.15: 99729251-99729251 on Build GRCh38
rs572277329 Chr.15: 99729260-99729260 on Build GRCh38
rs749994991 Chr.15: 99729418-99729418 on Build GRCh38
rs147293942 Chr.15: 99729391-99729391 on Build GRCh38
rs779543388 Chr.15: 99729361-99729361 on Build GRCh38
rs769660029 Chr.15: 99729194-99729194 on Build GRCh38
rs760665517 Chr.15: 99729206-99729206 on Build GRCh38
rs745450515 Chr.15: 99729241-99729241 on Build GRCh38
rs939619529 Chr.15: 99729202-99729202 on Build GRCh38
rs146387788 Chr.15: 99729231-99729231 on Build GRCh38
rs774152631 Chr.15: 99729269-99729269 on Build GRCh38
rs757879405 Chr.15: 99729161-99729161 on Build GRCh38
rs779445756 Chr.15: 99729159-99729159 on Build GRCh38
rs755881721 Chr.15: 99729346-99729346 on Build GRCh38
rs768681252 Chr.15: 99729266-99729266 on Build GRCh38
rs770638338 Chr.15: 99729094-99729094 on Build GRCh38
rs772762971 Chr.15: 99729083-99729083 on Build GRCh38
rs150607972 Chr.15: 99729333-99729333 on Build GRCh38
rs759917643 Chr.15: 99729216-99729216 on Build GRCh38
rs771028227 Chr.15: 99729375-99729375 on Build GRCh38
rs964367480 Chr.15: 99729302-99729302 on Build GRCh38
rs751118210 Chr.15: 99729232-99729232 on Build GRCh38
rs753374402 Chr.15: 99729459-99729459 on Build GRCh38
rs759713410 Chr.15: 99729307-99729307 on Build GRCh38
rs751807461 Chr.15: 99729420-99729421 on Build GRCh38
rs11852387 Chr.15: 99729408-99729408 on Build GRCh38
rs777272117 Chr.15: 99729169-99729169 on Build GRCh38
rs202140736 Chr.15: 99729173-99729173 on Build GRCh38
rs748104985 Chr.15: 99729479-99729479 on Build GRCh38
rs375247885 Chr.15: 99729379-99729379 on Build GRCh38
rs78996435 Chr.15: 99729132-99729132 on Build GRCh38
rs763064220 Chr.15: 99729267-99729268 on Build GRCh38

View Full Product Details