Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101271298-101271794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIMP
Entrez Gene ID
55829
Gene Name
VCP interacting membrane selenoprotein
Gene Aliases
AD-015, ADO15, SBBI8, SELS, SEPS1
UniGene
Hs.32148
Gene Chromosome Location
Chr.15: 101270909-101277520 on Build GRCh38
Transcripts
NM_203472.2

SNP Detail

SNP ID Chromosome Location
rs902477205 Chr.15: 101271373-101271373 on Build GRCh38
rs903230873 Chr.15: 101271455-101271455 on Build GRCh38
rs893663252 Chr.15: 101271436-101271436 on Build GRCh38
rs946558394 Chr.15: 101271634-101271634 on Build GRCh38
rs937978984 Chr.15: 101271427-101271427 on Build GRCh38
rs942199732 Chr.15: 101271604-101271604 on Build GRCh38
rs1013821076 Chr.15: 101271356-101271356 on Build GRCh38
rs1042164586 Chr.15: 101271659-101271659 on Build GRCh38
rs141344105 Chr.15: 101271482-101271482 on Build GRCh38
rs928973090 Chr.15: 101271350-101271350 on Build GRCh38
rs10577938 Chr.15: 101271732-101271734 on Build GRCh38
rs935335570 Chr.15: 101271753-101271753 on Build GRCh38
rs553110926 Chr.15: 101271677-101271677 on Build GRCh38
rs528270764 Chr.15: 101271320-101271320 on Build GRCh38
rs1030415037 Chr.15: 101271520-101271520 on Build GRCh38
rs751684040 Chr.15: 101271441-101271441 on Build GRCh38
rs1032422160 Chr.15: 101271459-101271459 on Build GRCh38
rs546856418 Chr.15: 101271380-101271380 on Build GRCh38
rs762041100 Chr.15: 101271602-101271602 on Build GRCh38
rs563412896 Chr.15: 101271721-101271721 on Build GRCh38
rs964613885 Chr.15: 101271503-101271503 on Build GRCh38
rs184973878 Chr.15: 101271593-101271593 on Build GRCh38
rs919007078 Chr.15: 101271523-101271523 on Build GRCh38
rs775523675 Chr.15: 101271360-101271360 on Build GRCh38
rs1017878127 Chr.15: 101271708-101271708 on Build GRCh38
rs915128887 Chr.15: 101271633-101271633 on Build GRCh38
rs557838600 Chr.15: 101271570-101271572 on Build GRCh38
rs1054955254 Chr.15: 101271429-101271429 on Build GRCh38
rs575107708 Chr.15: 101271744-101271744 on Build GRCh38
rs561477808 Chr.15: 101271715-101271715 on Build GRCh38
rs373837137 Chr.15: 101271397-101271397 on Build GRCh38
rs115633005 Chr.15: 101271468-101271468 on Build GRCh38
rs902549744 Chr.15: 101271712-101271712 on Build GRCh38
rs1017369797 Chr.15: 101271473-101271473 on Build GRCh38
rs574695180 Chr.15: 101271694-101271694 on Build GRCh38
rs575871584 Chr.15: 101271361-101271361 on Build GRCh38
rs1007538301 Chr.15: 101271707-101271707 on Build GRCh38
rs929112271 Chr.15: 101271552-101271552 on Build GRCh38
rs995087294 Chr.15: 101271729-101271729 on Build GRCh38
rs765324889 Chr.15: 101271675-101271675 on Build GRCh38
rs1011349314 Chr.15: 101271472-101271472 on Build GRCh38
rs537490063 Chr.15: 101271511-101271511 on Build GRCh38
rs986406109 Chr.15: 101271493-101271493 on Build GRCh38
rs764448632 Chr.15: 101271364-101271364 on Build GRCh38
rs79912783 Chr.15: 101271553-101271553 on Build GRCh38
rs192592171 Chr.15: 101271483-101271483 on Build GRCh38
rs908948228 Chr.15: 101271582-101271582 on Build GRCh38
rs954864698 Chr.15: 101271486-101271486 on Build GRCh38
rs568267981 Chr.15: 101271443-101271443 on Build GRCh38
rs758303350 Chr.15: 101271737-101271737 on Build GRCh38
rs534882055 Chr.15: 101271557-101271557 on Build GRCh38
rs189705638 Chr.15: 101271709-101271709 on Build GRCh38
rs558967296 Chr.15: 101271514-101271514 on Build GRCh38
rs543162848 Chr.15: 101271700-101271700 on Build GRCh38

View Full Product Details