Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 192474-193021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUC7L
Entrez Gene ID
55692
Gene Name
LUC7 like
Gene Aliases
LUC7B1, Luc7, SR+89, hLuc7B1
UniGene
Hs.16803
Gene Chromosome Location
Chr.16: 188969-229482 on Build GRCh38
Transcripts
NM_001320226.1 NM_018032.4 NM_201412.2 XM_005255427.3 XM_005255429.3 XM_011522561.2 XM_017023437.1 XM_017023438.1 XM_017023439.1 XM_017023440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6335930 c.721C>A p.Q241K Chr.16: 192982-192982 on Build GRCh38
6335931 c.721C>A p.Q241K Chr.16: 192982-192982 on Build GRCh38
4652700 c.755G>A p.R252Q Chr.16: 192948-192948 on Build GRCh38
6705144 c.754C>T p.R252W Chr.16: 192949-192949 on Build GRCh38
968928 c.763C>T p.R255C Chr.16: 192940-192940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014089323 Chr.16: 192627-192627 on Build GRCh38
rs927514318 Chr.16: 192801-192801 on Build GRCh38
rs999582071 Chr.16: 192646-192646 on Build GRCh38
rs199615679 Chr.16: 192944-192944 on Build GRCh38
rs951600283 Chr.16: 192871-192871 on Build GRCh38
rs776966705 Chr.16: 192908-192908 on Build GRCh38
rs1024936978 Chr.16: 192635-192635 on Build GRCh38
rs950330732 Chr.16: 192504-192504 on Build GRCh38
rs113080227 Chr.16: 192579-192579 on Build GRCh38
rs966016454 Chr.16: 192671-192671 on Build GRCh38
rs991227854 Chr.16: 192913-192913 on Build GRCh38
rs188596559 Chr.16: 192557-192557 on Build GRCh38
rs745906154 Chr.16: 192661-192661 on Build GRCh38
rs937105840 Chr.16: 192728-192728 on Build GRCh38
rs111775488 Chr.16: 192899-192899 on Build GRCh38
rs999510068 Chr.16: 192814-192814 on Build GRCh38
rs770669679 Chr.16: 192870-192870 on Build GRCh38
rs749801639 Chr.16: 192704-192704 on Build GRCh38
rs184685361 Chr.16: 192886-192886 on Build GRCh38
rs998337115 Chr.16: 192855-192855 on Build GRCh38
rs944294375 Chr.16: 192503-192503 on Build GRCh38
rs1005996402 Chr.16: 192723-192723 on Build GRCh38
rs1053782921 Chr.16: 192818-192818 on Build GRCh38
rs200402563 Chr.16: 192882-192882 on Build GRCh38
rs929725021 Chr.16: 192541-192541 on Build GRCh38
rs914721291 Chr.16: 192822-192822 on Build GRCh38
rs917596247 Chr.16: 192789-192789 on Build GRCh38
rs565115845 Chr.16: 192794-192794 on Build GRCh38
rs983775989 Chr.16: 192722-192722 on Build GRCh38
rs558287207 Chr.16: 192798-192798 on Build GRCh38
rs764788928 Chr.16: 192958-192958 on Build GRCh38
rs192953158 Chr.16: 192835-192835 on Build GRCh38
rs540799703 Chr.16: 192782-192782 on Build GRCh38
rs763299823 Chr.16: 192940-192940 on Build GRCh38
rs1032862422 Chr.16: 192649-192649 on Build GRCh38
rs893214135 Chr.16: 192653-192653 on Build GRCh38
rs532813200 Chr.16: 192555-192555 on Build GRCh38
rs767811000 Chr.16: 192976-192976 on Build GRCh38
rs567169264 Chr.16: 192502-192503 on Build GRCh38
rs554363979 Chr.16: 192829-192829 on Build GRCh38
rs756058279 Chr.16: 192992-192994 on Build GRCh38
rs143543557 Chr.16: 192969-192969 on Build GRCh38
rs576736452 Chr.16: 192827-192827 on Build GRCh38
rs536625279 Chr.16: 192659-192659 on Build GRCh38
rs775775803 Chr.16: 192939-192939 on Build GRCh38
rs971768381 Chr.16: 192503-192507 on Build GRCh38
rs1054570584 Chr.16: 192623-192623 on Build GRCh38
rs534641061 Chr.16: 192834-192834 on Build GRCh38
rs751782813 Chr.16: 192597-192597 on Build GRCh38
rs918275685 Chr.16: 192689-192689 on Build GRCh38
rs137968943 Chr.16: 192938-192938 on Build GRCh38
rs937202164 Chr.16: 192586-192586 on Build GRCh38
rs1055625461 Chr.16: 192588-192588 on Build GRCh38
rs746683397 Chr.16: 192853-192853 on Build GRCh38
rs918223324 Chr.16: 192538-192538 on Build GRCh38
rs759167157 Chr.16: 192949-192949 on Build GRCh38
rs893922106 Chr.16: 192823-192823 on Build GRCh38
rs986065766 Chr.16: 192660-192660 on Build GRCh38
rs1056040371 Chr.16: 192750-192750 on Build GRCh38
rs746143522 Chr.16: 192930-192930 on Build GRCh38
rs917057842 Chr.16: 192767-192767 on Build GRCh38
rs562118225 Chr.16: 192783-192783 on Build GRCh38
rs1041519941 Chr.16: 192968-192968 on Build GRCh38
rs757411867 Chr.16: 192657-192657 on Build GRCh38
rs7185345 Chr.16: 192771-192771 on Build GRCh38
rs909659719 Chr.16: 192727-192727 on Build GRCh38
rs747998270 Chr.16: 192890-192890 on Build GRCh38
rs976453979 Chr.16: 192536-192536 on Build GRCh38
rs780471737 Chr.16: 192682-192682 on Build GRCh38
rs373418558 Chr.16: 192885-192885 on Build GRCh38
rs1018871188 Chr.16: 192852-192852 on Build GRCh38
rs972182682 Chr.16: 192778-192778 on Build GRCh38
rs575252955 Chr.16: 192948-192948 on Build GRCh38
rs763945120 Chr.16: 192589-192589 on Build GRCh38
rs895763028 Chr.16: 192611-192611 on Build GRCh38
rs74322194 Chr.16: 192503-192504 on Build GRCh38
rs146839044 Chr.16: 192533-192533 on Build GRCh38
rs1006068493 Chr.16: 192901-192901 on Build GRCh38
rs902491688 Chr.16: 192645-192645 on Build GRCh38

View Full Product Details