Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 229072-229569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUC7L
Entrez Gene ID
55692
Gene Name
LUC7 like
Gene Aliases
LUC7B1, Luc7, SR+89, hLuc7B1
UniGene
Hs.16803
Gene Chromosome Location
Chr.16: 188969-229482 on Build GRCh38
Transcripts
NM_001320226.1 NM_018032.4 NM_201412.2 XM_005255427.3 XM_005255429.3 XM_011522561.2 XM_017023437.1 XM_017023438.1 XM_017023439.1 XM_017023440.1

SNP Detail

SNP ID Chromosome Location
rs966144355 Chr.16: 229120-229120 on Build GRCh38
rs868785848 Chr.16: 229290-229290 on Build GRCh38
rs865817515 Chr.16: 229284-229284 on Build GRCh38
rs570118809 Chr.16: 229155-229155 on Build GRCh38
rs11863920 Chr.16: 229265-229265 on Build GRCh38
rs763056180 Chr.16: 229254-229254 on Build GRCh38
rs764152034 Chr.16: 229374-229374 on Build GRCh38
rs760316777 Chr.16: 229221-229221 on Build GRCh38
rs759199307 Chr.16: 229263-229263 on Build GRCh38
rs905899218 Chr.16: 229153-229153 on Build GRCh38
rs986577824 Chr.16: 229112-229112 on Build GRCh38
rs766542563 Chr.16: 229273-229273 on Build GRCh38
rs1020089629 Chr.16: 229099-229099 on Build GRCh38
rs1042122830 Chr.16: 229244-229244 on Build GRCh38
rs765976342 Chr.16: 229199-229222 on Build GRCh38
rs749080714 Chr.16: 229361-229361 on Build GRCh38
rs74527341 Chr.16: 229146-229146 on Build GRCh38
rs547428290 Chr.16: 229105-229105 on Build GRCh38
rs1028669231 Chr.16: 229096-229096 on Build GRCh38
rs751327937 Chr.16: 229203-229203 on Build GRCh38
rs1045805737 Chr.16: 229150-229150 on Build GRCh38
rs748703812 Chr.16: 229169-229169 on Build GRCh38
rs565128358 Chr.16: 229450-229450 on Build GRCh38
rs926902087 Chr.16: 229154-229154 on Build GRCh38
rs779022595 Chr.16: 229331-229331 on Build GRCh38
rs1037623906 Chr.16: 229135-229135 on Build GRCh38
rs767265194 Chr.16: 229359-229359 on Build GRCh38
rs563276688 Chr.16: 229436-229436 on Build GRCh38
rs148122312 Chr.16: 229399-229399 on Build GRCh38
rs531744740 Chr.16: 229366-229366 on Build GRCh38
rs1056737283 Chr.16: 229375-229375 on Build GRCh38
rs536754053 Chr.16: 229140-229140 on Build GRCh38
rs762930156 Chr.16: 229373-229373 on Build GRCh38
rs938273712 Chr.16: 229157-229157 on Build GRCh38
rs768811026 Chr.16: 229391-229391 on Build GRCh38
rs184516407 Chr.16: 229417-229417 on Build GRCh38
rs972994693 Chr.16: 229283-229283 on Build GRCh38
rs1056556012 Chr.16: 229201-229201 on Build GRCh38
rs959570864 Chr.16: 229462-229462 on Build GRCh38
rs918743666 Chr.16: 229139-229139 on Build GRCh38
rs896169530 Chr.16: 229220-229220 on Build GRCh38
rs957237282 Chr.16: 229097-229097 on Build GRCh38
rs145503016 Chr.16: 229541-229541 on Build GRCh38
rs1000616026 Chr.16: 229092-229092 on Build GRCh38
rs926792064 Chr.16: 229369-229369 on Build GRCh38
rs910142501 Chr.16: 229128-229128 on Build GRCh38
rs536875547 Chr.16: 229162-229162 on Build GRCh38
rs774158254 Chr.16: 229379-229379 on Build GRCh38
rs773569434 Chr.16: 229199-229199 on Build GRCh38
rs953098800 Chr.16: 229257-229257 on Build GRCh38
rs1000691185 Chr.16: 229259-229259 on Build GRCh38
rs754131259 Chr.16: 229279-229279 on Build GRCh38
rs547638950 Chr.16: 229489-229489 on Build GRCh38
rs751327549 Chr.16: 229263-229263 on Build GRCh38
rs778061788 Chr.16: 229348-229348 on Build GRCh38
rs770511203 Chr.16: 229230-229230 on Build GRCh38
rs11863890 Chr.16: 229118-229118 on Build GRCh38
rs1010132230 Chr.16: 229196-229196 on Build GRCh38
rs116776296 Chr.16: 229453-229453 on Build GRCh38
rs941587010 Chr.16: 229130-229131 on Build GRCh38
rs866981245 Chr.16: 229478-229478 on Build GRCh38
rs377574098 Chr.16: 229351-229351 on Build GRCh38
rs952101191 Chr.16: 229363-229363 on Build GRCh38
rs558712666 Chr.16: 229198-229198 on Build GRCh38
rs541927086 Chr.16: 229415-229415 on Build GRCh38
rs775149546 Chr.16: 229371-229371 on Build GRCh38
rs953041614 Chr.16: 229115-229115 on Build GRCh38
rs779362465 Chr.16: 229091-229091 on Build GRCh38
rs772421480 Chr.16: 229197-229197 on Build GRCh38
rs758918456 Chr.16: 229339-229339 on Build GRCh38
rs762568560 Chr.16: 229174-229175 on Build GRCh38
rs1008135428 Chr.16: 229276-229276 on Build GRCh38
rs1019919869 Chr.16: 229321-229321 on Build GRCh38
rs560934230 Chr.16: 229378-229379 on Build GRCh38
rs1005958826 Chr.16: 229538-229538 on Build GRCh38
rs748587466 Chr.16: 229334-229334 on Build GRCh38
rs992701919 Chr.16: 229485-229485 on Build GRCh38
rs761482771 Chr.16: 229386-229386 on Build GRCh38
rs1034157037 Chr.16: 229457-229457 on Build GRCh38
rs758935602 Chr.16: 229241-229241 on Build GRCh38
rs754991358 Chr.16: 229292-229292 on Build GRCh38
rs763918949 Chr.16: 229256-229256 on Build GRCh38
rs993530600 Chr.16: 229443-229443 on Build GRCh38
rs532666333 Chr.16: 229100-229100 on Build GRCh38
rs140285632 Chr.16: 229442-229442 on Build GRCh38
rs370802608 Chr.16: 229418-229418 on Build GRCh38
rs903653821 Chr.16: 229407-229407 on Build GRCh38
rs764885755 Chr.16: 229249-229249 on Build GRCh38
rs1042030623 Chr.16: 229404-229404 on Build GRCh38
rs34193597 Chr.16: 229484-229485 on Build GRCh38
rs1050244882 Chr.16: 229508-229508 on Build GRCh38
rs1010082901 Chr.16: 229098-229098 on Build GRCh38
rs1025895550 Chr.16: 229435-229435 on Build GRCh38
rs919315908 Chr.16: 229132-229132 on Build GRCh38
rs192508401 Chr.16: 229356-229356 on Build GRCh38
rs769420366 Chr.16: 229370-229370 on Build GRCh38
rs965894054 Chr.16: 229200-229200 on Build GRCh38
rs962954770 Chr.16: 229266-229266 on Build GRCh38
rs867869855 Chr.16: 229304-229304 on Build GRCh38
rs961727214 Chr.16: 229423-229423 on Build GRCh38
rs776165021 Chr.16: 229237-229237 on Build GRCh38
rs893073669 Chr.16: 229181-229181 on Build GRCh38
rs918782919 Chr.16: 229341-229341 on Build GRCh38
rs775550952 Chr.16: 229252-229252 on Build GRCh38
rs1026316019 Chr.16: 229444-229444 on Build GRCh38
rs930799754 Chr.16: 229144-229144 on Build GRCh38
rs966206946 Chr.16: 229104-229104 on Build GRCh38
rs1054479917 Chr.16: 229368-229368 on Build GRCh38
rs901772320 Chr.16: 229498-229498 on Build GRCh38
rs889638494 Chr.16: 229431-229431 on Build GRCh38
rs933208954 Chr.16: 229500-229500 on Build GRCh38
rs953982249 Chr.16: 229108-229108 on Build GRCh38
rs1009166050 Chr.16: 229243-229243 on Build GRCh38
rs1028753779 Chr.16: 229414-229414 on Build GRCh38
rs972153922 Chr.16: 229124-229124 on Build GRCh38
rs866883515 Chr.16: 229288-229288 on Build GRCh38
rs1008453153 Chr.16: 229434-229434 on Build GRCh38
rs865937981 Chr.16: 229303-229303 on Build GRCh38
rs888934763 Chr.16: 229518-229518 on Build GRCh38
rs575278592 Chr.16: 229441-229441 on Build GRCh38

View Full Product Details