Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 589483-589972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB40C
Entrez Gene ID
57799
Gene Name
RAB40C, member RAS oncogene family
Gene Aliases
RARL, RASL8C
UniGene
Hs.459630
Gene Chromosome Location
Chr.16: 589357-629273 on Build GRCh38
Transcripts
NM_001172663.1 NM_001172664.1

SNP Detail

SNP ID Chromosome Location
rs933940698 Chr.16: 589616-589616 on Build GRCh38
rs933291880 Chr.16: 589803-589803 on Build GRCh38
rs555752311 Chr.16: 589886-589886 on Build GRCh38
rs1053240330 Chr.16: 589625-589625 on Build GRCh38
rs537817166 Chr.16: 589883-589883 on Build GRCh38
rs569131531 Chr.16: 589590-589590 on Build GRCh38
rs981781572 Chr.16: 589751-589751 on Build GRCh38
rs914762850 Chr.16: 589774-589774 on Build GRCh38
rs1017137037 Chr.16: 589518-589518 on Build GRCh38
rs977912472 Chr.16: 589950-589950 on Build GRCh38
rs970336367 Chr.16: 589548-589548 on Build GRCh38
rs879940538 Chr.16: 589630-589630 on Build GRCh38
rs1011842396 Chr.16: 589627-589627 on Build GRCh38
rs959534986 Chr.16: 589695-589695 on Build GRCh38
rs937979152 Chr.16: 589576-589576 on Build GRCh38
rs554977045 Chr.16: 589581-589581 on Build GRCh38
rs897406897 Chr.16: 589783-589783 on Build GRCh38
rs977922489 Chr.16: 589732-589732 on Build GRCh38
rs1030419665 Chr.16: 589642-589642 on Build GRCh38
rs994255214 Chr.16: 589793-589793 on Build GRCh38
rs947585960 Chr.16: 589779-589779 on Build GRCh38
rs909113025 Chr.16: 589513-589513 on Build GRCh38
rs941375820 Chr.16: 589767-589767 on Build GRCh38
rs570180155 Chr.16: 589617-589617 on Build GRCh38
rs919024564 Chr.16: 589537-589537 on Build GRCh38
rs533633339 Chr.16: 589744-589744 on Build GRCh38
rs150384847 Chr.16: 589717-589717 on Build GRCh38
rs1038587391 Chr.16: 589771-589771 on Build GRCh38
rs989391190 Chr.16: 589714-589714 on Build GRCh38
rs934827225 Chr.16: 589608-589608 on Build GRCh38
rs112713499 Chr.16: 589798-589798 on Build GRCh38
rs116606022 Chr.16: 589898-589898 on Build GRCh38
rs994307719 Chr.16: 589635-589635 on Build GRCh38
rs915104868 Chr.16: 589588-589588 on Build GRCh38
rs988908400 Chr.16: 589773-589773 on Build GRCh38
rs990863714 Chr.16: 589522-589522 on Build GRCh38
rs111490416 Chr.16: 589819-589819 on Build GRCh38
rs1033337589 Chr.16: 589679-589679 on Build GRCh38
rs954281176 Chr.16: 589514-589514 on Build GRCh38
rs934097959 Chr.16: 589754-589754 on Build GRCh38
rs997961248 Chr.16: 589632-589632 on Build GRCh38
rs935108882 Chr.16: 589759-589759 on Build GRCh38
rs923674865 Chr.16: 589755-589755 on Build GRCh38
rs1033867366 Chr.16: 589885-589885 on Build GRCh38
rs893362910 Chr.16: 589626-589626 on Build GRCh38
rs989708486 Chr.16: 589580-589580 on Build GRCh38
rs945196964 Chr.16: 589720-589720 on Build GRCh38
rs981748868 Chr.16: 589570-589570 on Build GRCh38
rs906282583 Chr.16: 589825-589825 on Build GRCh38
rs1003272721 Chr.16: 589835-589835 on Build GRCh38
rs1039421420 Chr.16: 589629-589629 on Build GRCh38
rs551595510 Chr.16: 589634-589634 on Build GRCh38
rs889816477 Chr.16: 589756-589756 on Build GRCh38
rs886549315 Chr.16: 589649-589649 on Build GRCh38
rs1016118062 Chr.16: 589659-589659 on Build GRCh38
rs970410335 Chr.16: 589747-589747 on Build GRCh38
rs967069536 Chr.16: 589913-589913 on Build GRCh38
rs963354314 Chr.16: 589521-589521 on Build GRCh38
rs1039281488 Chr.16: 589792-589792 on Build GRCh38
rs989561149 Chr.16: 589888-589888 on Build GRCh38
rs949117685 Chr.16: 589550-589550 on Build GRCh38
rs1049797113 Chr.16: 589621-589621 on Build GRCh38
rs900791656 Chr.16: 589795-589795 on Build GRCh38
rs34353619 Chr.16: 589502-589502 on Build GRCh38
rs926500065 Chr.16: 589575-589575 on Build GRCh38
rs1006131722 Chr.16: 589515-589515 on Build GRCh38
rs539746497 Chr.16: 589607-589607 on Build GRCh38
rs990756562 Chr.16: 589670-589670 on Build GRCh38
rs1004706313 Chr.16: 589656-589656 on Build GRCh38
rs550713690 Chr.16: 589530-589530 on Build GRCh38
rs573678983 Chr.16: 589824-589824 on Build GRCh38
rs963603322 Chr.16: 589669-589669 on Build GRCh38

View Full Product Details