Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 629120-629619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB40C
Entrez Gene ID
57799
Gene Name
RAB40C, member RAS oncogene family
Gene Aliases
RARL, RASL8C
UniGene
Hs.459630
Gene Chromosome Location
Chr.16: 589357-629273 on Build GRCh38
Transcripts
NM_001172663.1 NM_001172664.1 NM_001172665.1 NM_001172666.1 NM_021168.4

SNP Detail

SNP ID Chromosome Location
rs1025239509 Chr.16: 629164-629164 on Build GRCh38
rs537886389 Chr.16: 629410-629410 on Build GRCh38
rs989906713 Chr.16: 629201-629201 on Build GRCh38
rs1028618750 Chr.16: 629526-629526 on Build GRCh38
rs979744397 Chr.16: 629328-629328 on Build GRCh38
rs896583877 Chr.16: 629311-629311 on Build GRCh38
rs573361567 Chr.16: 629543-629543 on Build GRCh38
rs909060597 Chr.16: 629270-629270 on Build GRCh38
rs913786109 Chr.16: 629173-629173 on Build GRCh38
rs987937782 Chr.16: 629157-629157 on Build GRCh38
rs930863130 Chr.16: 629397-629397 on Build GRCh38
rs985963686 Chr.16: 629252-629252 on Build GRCh38
rs879307419 Chr.16: 629318-629318 on Build GRCh38
rs1019307413 Chr.16: 629544-629544 on Build GRCh38
rs960528613 Chr.16: 629153-629153 on Build GRCh38
rs558300700 Chr.16: 629507-629507 on Build GRCh38
rs562490665 Chr.16: 629240-629240 on Build GRCh38
rs963195902 Chr.16: 629426-629426 on Build GRCh38
rs986540199 Chr.16: 629210-629210 on Build GRCh38
rs551588218 Chr.16: 629258-629258 on Build GRCh38
rs1009765600 Chr.16: 629546-629546 on Build GRCh38
rs909027787 Chr.16: 629413-629413 on Build GRCh38
rs967086296 Chr.16: 629209-629209 on Build GRCh38
rs759766134 Chr.16: 629473-629473 on Build GRCh38
rs972233129 Chr.16: 629374-629374 on Build GRCh38
rs952274026 Chr.16: 629577-629577 on Build GRCh38
rs757258143 Chr.16: 629436-629436 on Build GRCh38
rs878863837 Chr.16: 629346-629346 on Build GRCh38
rs373749522 Chr.16: 629398-629398 on Build GRCh38
rs920909066 Chr.16: 629186-629186 on Build GRCh38
rs35609555 Chr.16: 629157-629157 on Build GRCh38
rs979098204 Chr.16: 629179-629179 on Build GRCh38
rs1036943921 Chr.16: 629237-629237 on Build GRCh38
rs932334255 Chr.16: 629194-629194 on Build GRCh38
rs544093931 Chr.16: 629213-629213 on Build GRCh38
rs938204047 Chr.16: 629515-629515 on Build GRCh38
rs919408588 Chr.16: 629376-629376 on Build GRCh38
rs756090083 Chr.16: 629190-629190 on Build GRCh38
rs1028274596 Chr.16: 629331-629331 on Build GRCh38
rs949443603 Chr.16: 629294-629294 on Build GRCh38
rs35784676 Chr.16: 629400-629400 on Build GRCh38
rs1010049506 Chr.16: 629414-629414 on Build GRCh38
rs1034096191 Chr.16: 629189-629189 on Build GRCh38
rs752775167 Chr.16: 629531-629531 on Build GRCh38
rs17555255 Chr.16: 629241-629241 on Build GRCh38
rs750577095 Chr.16: 629314-629315 on Build GRCh38
rs568707517 Chr.16: 629228-629228 on Build GRCh38
rs369325187 Chr.16: 629479-629479 on Build GRCh38
rs968383273 Chr.16: 629174-629174 on Build GRCh38
rs185520196 Chr.16: 629147-629147 on Build GRCh38
rs3180836 Chr.16: 629268-629268 on Build GRCh38
rs919223886 Chr.16: 629291-629291 on Build GRCh38
rs953689668 Chr.16: 629332-629332 on Build GRCh38
rs1047067401 Chr.16: 629301-629301 on Build GRCh38
rs569710001 Chr.16: 629456-629456 on Build GRCh38
rs903676080 Chr.16: 629491-629491 on Build GRCh38
rs1035031012 Chr.16: 629288-629288 on Build GRCh38
rs926624008 Chr.16: 629495-629495 on Build GRCh38
rs949411275 Chr.16: 629455-629455 on Build GRCh38
rs529261613 Chr.16: 629180-629180 on Build GRCh38
rs944814969 Chr.16: 629231-629231 on Build GRCh38
rs527332033 Chr.16: 629348-629348 on Build GRCh38
rs1056405182 Chr.16: 629411-629411 on Build GRCh38
rs967932778 Chr.16: 629319-629319 on Build GRCh38
rs762834719 Chr.16: 629347-629347 on Build GRCh38
rs202063822 Chr.16: 629335-629335 on Build GRCh38
rs9929621 Chr.16: 629412-629412 on Build GRCh38
rs35915772 Chr.16: 629274-629274 on Build GRCh38
rs144198679 Chr.16: 629557-629557 on Build GRCh38
rs1020058228 Chr.16: 629380-629380 on Build GRCh38
rs953949826 Chr.16: 629527-629527 on Build GRCh38
rs73485459 Chr.16: 629563-629563 on Build GRCh38
rs1019920752 Chr.16: 629207-629207 on Build GRCh38
rs764701111 Chr.16: 629553-629553 on Build GRCh38
rs986433985 Chr.16: 629530-629530 on Build GRCh38
rs766897107 Chr.16: 629493-629493 on Build GRCh38
rs569602082 Chr.16: 629427-629427 on Build GRCh38
rs953333235 Chr.16: 629251-629251 on Build GRCh38
rs1009326675 Chr.16: 629315-629315 on Build GRCh38
rs905186791 Chr.16: 629273-629273 on Build GRCh38
rs939763742 Chr.16: 629358-629358 on Build GRCh38
rs1041857896 Chr.16: 629550-629550 on Build GRCh38

View Full Product Details