Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1096013-1096506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF8
Entrez Gene ID
390664
Gene Name
C1q and tumor necrosis factor related protein 8
Gene Aliases
CTRP8, UNQ5829
UniGene
Hs.527853
Gene Chromosome Location
Chr.16: 1088226-1096306 on Build GRCh38
Transcripts
NM_207419.3 XM_011522483.2

SNP Detail

SNP ID Chromosome Location
rs900156437 Chr.16: 1096399-1096399 on Build GRCh38
rs916583378 Chr.16: 1096337-1096337 on Build GRCh38
rs950351286 Chr.16: 1096161-1096161 on Build GRCh38
rs117851085 Chr.16: 1096191-1096191 on Build GRCh38
rs4113901 Chr.16: 1096070-1096070 on Build GRCh38
rs968220348 Chr.16: 1096422-1096422 on Build GRCh38
rs977866208 Chr.16: 1096425-1096425 on Build GRCh38
rs13330580 Chr.16: 1096363-1096363 on Build GRCh38
rs1004702956 Chr.16: 1096471-1096471 on Build GRCh38
rs566684067 Chr.16: 1096050-1096050 on Build GRCh38
rs2076422 Chr.16: 1096228-1096228 on Build GRCh38
rs948063236 Chr.16: 1096469-1096469 on Build GRCh38
rs146748469 Chr.16: 1096098-1096098 on Build GRCh38
rs1027006256 Chr.16: 1096313-1096313 on Build GRCh38
rs963249360 Chr.16: 1096129-1096129 on Build GRCh38
rs974514846 Chr.16: 1096281-1096281 on Build GRCh38
rs950403284 Chr.16: 1096055-1096055 on Build GRCh38
rs534249317 Chr.16: 1096323-1096323 on Build GRCh38
rs191881467 Chr.16: 1096249-1096249 on Build GRCh38
rs546304625 Chr.16: 1096204-1096204 on Build GRCh38
rs542840449 Chr.16: 1096477-1096477 on Build GRCh38
rs963007129 Chr.16: 1096045-1096045 on Build GRCh38
rs936408364 Chr.16: 1096391-1096391 on Build GRCh38
rs7187586 Chr.16: 1096109-1096109 on Build GRCh38
rs13336008 Chr.16: 1096432-1096432 on Build GRCh38
rs892084988 Chr.16: 1096411-1096411 on Build GRCh38
rs1047276436 Chr.16: 1096374-1096374 on Build GRCh38
rs535354058 Chr.16: 1096062-1096062 on Build GRCh38
rs780102690 Chr.16: 1096264-1096264 on Build GRCh38
rs866846920 Chr.16: 1096481-1096481 on Build GRCh38
rs766751712 Chr.16: 1096466-1096466 on Build GRCh38
rs903618895 Chr.16: 1096270-1096270 on Build GRCh38
rs896349405 Chr.16: 1096126-1096127 on Build GRCh38
rs757082502 Chr.16: 1096348-1096348 on Build GRCh38
rs760309820 Chr.16: 1096461-1096461 on Build GRCh38
rs1056683850 Chr.16: 1096113-1096113 on Build GRCh38
rs1056408644 Chr.16: 1096222-1096222 on Build GRCh38
rs576359694 Chr.16: 1096375-1096375 on Build GRCh38
rs60992236 Chr.16: 1096334-1096334 on Build GRCh38
rs562749059 Chr.16: 1096480-1096480 on Build GRCh38
rs573332000 Chr.16: 1096171-1096171 on Build GRCh38
rs1038888197 Chr.16: 1096075-1096075 on Build GRCh38
rs756330794 Chr.16: 1096263-1096263 on Build GRCh38
rs557299658 Chr.16: 1096403-1096403 on Build GRCh38
rs895257486 Chr.16: 1096114-1096114 on Build GRCh38
rs549270144 Chr.16: 1096065-1096065 on Build GRCh38
rs992030126 Chr.16: 1096449-1096449 on Build GRCh38
rs967893232 Chr.16: 1096288-1096288 on Build GRCh38
rs761356264 Chr.16: 1096258-1096258 on Build GRCh38
rs560242218 Chr.16: 1096445-1096445 on Build GRCh38
rs775220531 Chr.16: 1096166-1096166 on Build GRCh38
rs532838611 Chr.16: 1096193-1096193 on Build GRCh38
rs12719787 Chr.16: 1096126-1096126 on Build GRCh38
rs139504276 Chr.16: 1096072-1096072 on Build GRCh38
rs919922412 Chr.16: 1096086-1096086 on Build GRCh38
rs1000643131 Chr.16: 1096272-1096272 on Build GRCh38
rs144235084 Chr.16: 1096318-1096318 on Build GRCh38
rs558837129 Chr.16: 1096116-1096116 on Build GRCh38
rs946579430 Chr.16: 1096181-1096181 on Build GRCh38
rs1042328834 Chr.16: 1096089-1096089 on Build GRCh38
rs574888793 Chr.16: 1096132-1096132 on Build GRCh38
rs1027556059 Chr.16: 1096468-1096468 on Build GRCh38
rs753356452 Chr.16: 1096129-1096129 on Build GRCh38
rs558272589 Chr.16: 1096081-1096081 on Build GRCh38
rs764683044 Chr.16: 1096136-1096136 on Build GRCh38
rs1039941812 Chr.16: 1096484-1096484 on Build GRCh38
rs930055429 Chr.16: 1096478-1096478 on Build GRCh38
rs2076423 Chr.16: 1096254-1096254 on Build GRCh38
rs930107705 Chr.16: 1096369-1096369 on Build GRCh38
rs957158867 Chr.16: 1096144-1096144 on Build GRCh38
rs376363084 Chr.16: 1096277-1096277 on Build GRCh38
rs192323963 Chr.16: 1096085-1096085 on Build GRCh38
rs918929168 Chr.16: 1096051-1096051 on Build GRCh38
rs948085614 Chr.16: 1096338-1096338 on Build GRCh38
rs1022496293 Chr.16: 1096283-1096283 on Build GRCh38
rs752055383 Chr.16: 1096162-1096162 on Build GRCh38
rs569172160 Chr.16: 1096252-1096252 on Build GRCh38
rs150976348 Chr.16: 1096473-1096473 on Build GRCh38
rs1012336205 Chr.16: 1096229-1096229 on Build GRCh38
rs1017585486 Chr.16: 1096041-1096041 on Build GRCh38
rs1047220072 Chr.16: 1096479-1096479 on Build GRCh38
rs13337047 Chr.16: 1096415-1096415 on Build GRCh38
rs545318624 Chr.16: 1096387-1096387 on Build GRCh38
rs911622934 Chr.16: 1096165-1096165 on Build GRCh38
rs915111931 Chr.16: 1096076-1096076 on Build GRCh38
rs939336895 Chr.16: 1096214-1096214 on Build GRCh38
rs977916771 Chr.16: 1096312-1096312 on Build GRCh38
rs544520539 Chr.16: 1096268-1096268 on Build GRCh38
rs915080287 Chr.16: 1096178-1096178 on Build GRCh38
rs960568062 Chr.16: 1096446-1096446 on Build GRCh38
rs987213186 Chr.16: 1096064-1096064 on Build GRCh38
rs567705549 Chr.16: 1096335-1096335 on Build GRCh38
rs1012805486 Chr.16: 1096118-1096118 on Build GRCh38
rs184113409 Chr.16: 1096429-1096429 on Build GRCh38
rs117406712 Chr.16: 1096130-1096130 on Build GRCh38
rs891346982 Chr.16: 1096376-1096376 on Build GRCh38
rs569111744 Chr.16: 1096071-1096071 on Build GRCh38
rs542666605 Chr.16: 1096390-1096390 on Build GRCh38
rs751257905 Chr.16: 1096132-1096133 on Build GRCh38
rs562917089 Chr.16: 1096278-1096278 on Build GRCh38
rs386787668 Chr.16: 1096126-1096128 on Build GRCh38
rs943712695 Chr.16: 1096412-1096412 on Build GRCh38
rs975427440 Chr.16: 1096194-1096194 on Build GRCh38
rs560049491 Chr.16: 1096217-1096217 on Build GRCh38
rs57793087 Chr.16: 1096094-1096094 on Build GRCh38
rs1044876454 Chr.16: 1096128-1096128 on Build GRCh38
rs964774681 Chr.16: 1096280-1096280 on Build GRCh38
rs767232937 Chr.16: 1096077-1096077 on Build GRCh38
rs996273143 Chr.16: 1096400-1096400 on Build GRCh38
rs939284604 Chr.16: 1096322-1096322 on Build GRCh38
rs370446472 Chr.16: 1096362-1096362 on Build GRCh38
rs60628434 Chr.16: 1096098-1096098 on Build GRCh38
rs543840571 Chr.16: 1096143-1096143 on Build GRCh38
rs920020901 Chr.16: 1096195-1096195 on Build GRCh38

View Full Product Details