Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1438718-1439213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC154
Entrez Gene ID
645811
Gene Name
coiled-coil domain containing 154
Gene Aliases
C16orf29
UniGene
Hs.355232
Gene Chromosome Location
Chr.16: 1434388-1444489 on Build GRCh38
Transcripts
NM_001143980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559126 c.792G>A p.S264S Chr.16: 1438929-1438929 on Build GRCh38
3957146 c.777+5G>C p.? Chr.16: 1439020-1439020 on Build GRCh38
4646508 c.728A>G p.E243G Chr.16: 1439074-1439074 on Build GRCh38
5659960 c.762C>T p.F254F Chr.16: 1439040-1439040 on Build GRCh38
6318888 c.767C>A p.A256D Chr.16: 1439035-1439035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748984287 Chr.16: 1439133-1439133 on Build GRCh38
rs113168089 Chr.16: 1439034-1439034 on Build GRCh38
rs765755549 Chr.16: 1439037-1439051 on Build GRCh38
rs777908652 Chr.16: 1438807-1438807 on Build GRCh38
rs754264923 Chr.16: 1438918-1438918 on Build GRCh38
rs375024782 Chr.16: 1439061-1439061 on Build GRCh38
rs549704561 Chr.16: 1439060-1439060 on Build GRCh38
rs1006217770 Chr.16: 1439036-1439036 on Build GRCh38
rs751545891 Chr.16: 1438939-1438939 on Build GRCh38
rs980427537 Chr.16: 1439107-1439107 on Build GRCh38
rs928542660 Chr.16: 1439175-1439175 on Build GRCh38
rs917114130 Chr.16: 1439088-1439089 on Build GRCh38
rs1030527485 Chr.16: 1438894-1438894 on Build GRCh38
rs1032392294 Chr.16: 1438800-1438800 on Build GRCh38
rs775870024 Chr.16: 1438762-1438762 on Build GRCh38
rs776593368 Chr.16: 1438748-1438748 on Build GRCh38
rs1016490294 Chr.16: 1438876-1438876 on Build GRCh38
rs773930484 Chr.16: 1439008-1439008 on Build GRCh38
rs543638828 Chr.16: 1438844-1438844 on Build GRCh38
rs964177036 Chr.16: 1439115-1439115 on Build GRCh38
rs945878269 Chr.16: 1438812-1438812 on Build GRCh38
rs757483668 Chr.16: 1438948-1438948 on Build GRCh38
rs759504955 Chr.16: 1438756-1438756 on Build GRCh38
rs1036928502 Chr.16: 1438772-1438772 on Build GRCh38
rs991765108 Chr.16: 1439178-1439178 on Build GRCh38
rs867487247 Chr.16: 1438889-1438889 on Build GRCh38
rs762109285 Chr.16: 1438864-1438908 on Build GRCh38
rs756482598 Chr.16: 1438795-1438795 on Build GRCh38
rs767229385 Chr.16: 1438897-1438897 on Build GRCh38
rs534794382 Chr.16: 1438806-1438806 on Build GRCh38
rs752965730 Chr.16: 1439054-1439054 on Build GRCh38
rs779927486 Chr.16: 1439125-1439125 on Build GRCh38
rs11860973 Chr.16: 1438767-1438767 on Build GRCh38
rs564038542 Chr.16: 1438919-1438919 on Build GRCh38
rs993722067 Chr.16: 1439089-1439089 on Build GRCh38
rs529941512 Chr.16: 1439035-1439035 on Build GRCh38
rs989811489 Chr.16: 1438931-1438931 on Build GRCh38
rs747725604 Chr.16: 1439150-1439150 on Build GRCh38
rs768648577 Chr.16: 1439134-1439134 on Build GRCh38
rs773913725 Chr.16: 1438890-1438890 on Build GRCh38
rs190211752 Chr.16: 1438930-1438930 on Build GRCh38
rs116378034 Chr.16: 1438834-1438834 on Build GRCh38
rs763635543 Chr.16: 1438866-1438866 on Build GRCh38
rs914489184 Chr.16: 1438955-1438955 on Build GRCh38
rs949880058 Chr.16: 1439090-1439090 on Build GRCh38
rs893865730 Chr.16: 1438951-1438951 on Build GRCh38
rs554613502 Chr.16: 1438809-1438809 on Build GRCh38
rs1056293865 Chr.16: 1438836-1438836 on Build GRCh38
rs1053890261 Chr.16: 1438929-1438929 on Build GRCh38
rs370275710 Chr.16: 1438870-1438870 on Build GRCh38
rs750625067 Chr.16: 1439110-1439110 on Build GRCh38
rs761169957 Chr.16: 1439009-1439009 on Build GRCh38
rs920965099 Chr.16: 1438852-1438852 on Build GRCh38
rs1031019687 Chr.16: 1439108-1439108 on Build GRCh38
rs554676814 Chr.16: 1439177-1439177 on Build GRCh38
rs953706076 Chr.16: 1438879-1438879 on Build GRCh38
rs539067141 Chr.16: 1438782-1438782 on Build GRCh38
rs1041573077 Chr.16: 1438815-1438815 on Build GRCh38
rs149644956 Chr.16: 1439149-1439149 on Build GRCh38
rs771105661 Chr.16: 1438819-1438819 on Build GRCh38
rs751668688 Chr.16: 1439094-1439094 on Build GRCh38
rs907265352 Chr.16: 1438832-1438832 on Build GRCh38
rs1044311915 Chr.16: 1438921-1438921 on Build GRCh38
rs184952683 Chr.16: 1439159-1439159 on Build GRCh38
rs984282113 Chr.16: 1438885-1438885 on Build GRCh38
rs917062002 Chr.16: 1438914-1438914 on Build GRCh38
rs748177333 Chr.16: 1438803-1438803 on Build GRCh38
rs953864117 Chr.16: 1439049-1439049 on Build GRCh38
rs764451368 Chr.16: 1439067-1439067 on Build GRCh38
rs1036855023 Chr.16: 1438891-1438891 on Build GRCh38
rs757328195 Chr.16: 1439106-1439106 on Build GRCh38
rs769728163 Chr.16: 1438837-1438837 on Build GRCh38
rs760033583 Chr.16: 1439011-1439011 on Build GRCh38
rs1018017897 Chr.16: 1439113-1439113 on Build GRCh38
rs910966098 Chr.16: 1438926-1438926 on Build GRCh38
rs552401459 Chr.16: 1439154-1439154 on Build GRCh38
rs3751896 Chr.16: 1438989-1438989 on Build GRCh38
rs959198939 Chr.16: 1438775-1438775 on Build GRCh38
rs776436544 Chr.16: 1438833-1438833 on Build GRCh38
rs560934205 Chr.16: 1439010-1439010 on Build GRCh38
rs77488687 Chr.16: 1438990-1438990 on Build GRCh38
rs745732661 Chr.16: 1438979-1438979 on Build GRCh38
rs983724149 Chr.16: 1439055-1439055 on Build GRCh38
rs779501245 Chr.16: 1438799-1438799 on Build GRCh38
rs998343101 Chr.16: 1439182-1439182 on Build GRCh38
rs777942614 Chr.16: 1438938-1438938 on Build GRCh38
rs755941093 Chr.16: 1439111-1439111 on Build GRCh38
rs765115110 Chr.16: 1438757-1438757 on Build GRCh38
rs754214235 Chr.16: 1438798-1438798 on Build GRCh38
rs751208512 Chr.16: 1438789-1438789 on Build GRCh38
rs972453984 Chr.16: 1439088-1439088 on Build GRCh38
rs867648011 Chr.16: 1439017-1439017 on Build GRCh38
rs768434732 Chr.16: 1438998-1438998 on Build GRCh38
rs781453689 Chr.16: 1439109-1439109 on Build GRCh38
rs986544876 Chr.16: 1438925-1438925 on Build GRCh38
rs961135099 Chr.16: 1439070-1439070 on Build GRCh38
rs773046038 Chr.16: 1439166-1439166 on Build GRCh38
rs753232019 Chr.16: 1438932-1438932 on Build GRCh38
rs545030062 Chr.16: 1438936-1438936 on Build GRCh38
rs938530162 Chr.16: 1438993-1438993 on Build GRCh38
rs572066006 Chr.16: 1438802-1438802 on Build GRCh38
rs58293579 Chr.16: 1438796-1438796 on Build GRCh38
rs762460376 Chr.16: 1438781-1438781 on Build GRCh38
rs774994961 Chr.16: 1438776-1438776 on Build GRCh38
rs534664971 Chr.16: 1439172-1439172 on Build GRCh38
rs180995283 Chr.16: 1438994-1438994 on Build GRCh38
rs781328839 Chr.16: 1438978-1438978 on Build GRCh38

View Full Product Details