Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1997611-1998112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF598
Entrez Gene ID
90850
Gene Name
zinc finger protein 598
Gene Aliases
-
UniGene
Hs.343828
Gene Chromosome Location
Chr.16: 1997652-2009821 on Build GRCh38
Transcripts
NM_178167.3

SNP Detail

SNP ID Chromosome Location
rs566273496 Chr.16: 1997895-1997895 on Build GRCh38
rs745667479 Chr.16: 1998017-1998017 on Build GRCh38
rs745924392 Chr.16: 1997923-1997923 on Build GRCh38
rs530065537 Chr.16: 1997734-1997734 on Build GRCh38
rs1036772011 Chr.16: 1997787-1997787 on Build GRCh38
rs866340967 Chr.16: 1997938-1997938 on Build GRCh38
rs558562407 Chr.16: 1998064-1998064 on Build GRCh38
rs376196191 Chr.16: 1997909-1997909 on Build GRCh38
rs988176724 Chr.16: 1997648-1997648 on Build GRCh38
rs866200618 Chr.16: 1997868-1997868 on Build GRCh38
rs183296380 Chr.16: 1997917-1997917 on Build GRCh38
rs3889 Chr.16: 1997862-1997862 on Build GRCh38
rs1020971864 Chr.16: 1997886-1997886 on Build GRCh38
rs984437860 Chr.16: 1997921-1997921 on Build GRCh38
rs755642311 Chr.16: 1997807-1997807 on Build GRCh38
rs370279541 Chr.16: 1997912-1997912 on Build GRCh38
rs529845257 Chr.16: 1997656-1997657 on Build GRCh38
rs935778725 Chr.16: 1997652-1997652 on Build GRCh38
rs898022133 Chr.16: 1997804-1997804 on Build GRCh38
rs138776921 Chr.16: 1997828-1997832 on Build GRCh38
rs921160258 Chr.16: 1997690-1997690 on Build GRCh38
rs3890 Chr.16: 1997915-1997915 on Build GRCh38
rs576060422 Chr.16: 1997980-1997980 on Build GRCh38
rs774148515 Chr.16: 1998051-1998051 on Build GRCh38
rs532440651 Chr.16: 1997870-1997870 on Build GRCh38
rs888112024 Chr.16: 1997727-1997727 on Build GRCh38
rs895541195 Chr.16: 1998040-1998040 on Build GRCh38
rs11865915 Chr.16: 1997920-1997920 on Build GRCh38
rs1048018751 Chr.16: 1997705-1997705 on Build GRCh38
rs1002619783 Chr.16: 1997827-1997827 on Build GRCh38
rs1052709575 Chr.16: 1997655-1997655 on Build GRCh38
rs778649539 Chr.16: 1998042-1998044 on Build GRCh38
rs572250922 Chr.16: 1998085-1998085 on Build GRCh38
rs761864228 Chr.16: 1998069-1998069 on Build GRCh38
rs932315091 Chr.16: 1997700-1997700 on Build GRCh38
rs781202945 Chr.16: 1997896-1997896 on Build GRCh38
rs538934139 Chr.16: 1998013-1998013 on Build GRCh38
rs939788225 Chr.16: 1997752-1997752 on Build GRCh38
rs769899591 Chr.16: 1998048-1998048 on Build GRCh38
rs1013730823 Chr.16: 1997808-1997808 on Build GRCh38
rs534898997 Chr.16: 1997901-1997901 on Build GRCh38
rs905411759 Chr.16: 1997821-1997821 on Build GRCh38
rs9081 Chr.16: 1997890-1997890 on Build GRCh38
rs199912012 Chr.16: 1998051-1998051 on Build GRCh38
rs921181053 Chr.16: 1997918-1997918 on Build GRCh38
rs1036801454 Chr.16: 1998033-1998033 on Build GRCh38
rs771706934 Chr.16: 1998073-1998073 on Build GRCh38
rs1058502 Chr.16: 1997810-1997810 on Build GRCh38

View Full Product Details