Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3058681-3059174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP25
Entrez Gene ID
64386
Gene Name
matrix metallopeptidase 25
Gene Aliases
MMP-25, MMP20, MMP20A, MMPL1, MT-MMP 6, MT-MMP6, MT6-MMP, MT6MMP, MTMMP6
UniGene
Hs.654979
Gene Chromosome Location
Chr.16: 3045963-3060729 on Build GRCh38
Transcripts
NM_022468.4 XM_011522602.2 XM_011522604.2 XM_011522605.2 NR_123723.1

Gene Symbol
MMP25-AS1
Entrez Gene ID
100507419
Gene Name
MMP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.312579
Gene Chromosome Location
Chr.16: 3052125-3059370 on Build GRCh38
Transcripts
NM_022468.4 XM_011522602.2 XM_011522604.2 XM_011522605.2 NR_123723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4702717 c.1558delC p.A523fs*37 Chr.16: 3058967-3058967 on Build GRCh38
5658748 c.1654T>A p.L552M Chr.16: 3059063-3059063 on Build GRCh38
7276384 c.1557G>A p.R519R Chr.16: 3058966-3058966 on Build GRCh38
4388134 c.1674T>G p.G558G Chr.16: 3059083-3059083 on Build GRCh38
970090 c.1573C>A p.P525T Chr.16: 3058982-3058982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753378201 Chr.16: 3058792-3058792 on Build GRCh38
rs993068660 Chr.16: 3058889-3058889 on Build GRCh38
rs569853317 Chr.16: 3058971-3058971 on Build GRCh38
rs575147898 Chr.16: 3059066-3059066 on Build GRCh38
rs201420558 Chr.16: 3058952-3058952 on Build GRCh38
rs779147422 Chr.16: 3058969-3058969 on Build GRCh38
rs888747567 Chr.16: 3059147-3059147 on Build GRCh38
rs370097888 Chr.16: 3058944-3058944 on Build GRCh38
rs555289844 Chr.16: 3059040-3059040 on Build GRCh38
rs757304887 Chr.16: 3059068-3059068 on Build GRCh38
rs1045751560 Chr.16: 3058874-3058874 on Build GRCh38
rs532757551 Chr.16: 3059100-3059100 on Build GRCh38
rs145529221 Chr.16: 3058864-3058864 on Build GRCh38
rs199549053 Chr.16: 3058956-3058956 on Build GRCh38
rs761320147 Chr.16: 3058902-3058902 on Build GRCh38
rs541071753 Chr.16: 3059033-3059033 on Build GRCh38
rs759906251 Chr.16: 3058898-3058898 on Build GRCh38
rs980072126 Chr.16: 3059087-3059087 on Build GRCh38
rs201570902 Chr.16: 3058893-3058893 on Build GRCh38
rs770167110 Chr.16: 3058706-3058706 on Build GRCh38
rs762482944 Chr.16: 3059023-3059023 on Build GRCh38
rs762488230 Chr.16: 3058908-3058908 on Build GRCh38
rs759939900 Chr.16: 3058783-3058783 on Build GRCh38
rs747535469 Chr.16: 3058704-3058705 on Build GRCh38
rs200461438 Chr.16: 3058903-3058903 on Build GRCh38
rs774987299 Chr.16: 3058714-3058714 on Build GRCh38
rs558651639 Chr.16: 3059019-3059019 on Build GRCh38
rs141638950 Chr.16: 3058812-3058812 on Build GRCh38
rs764887334 Chr.16: 3058796-3058796 on Build GRCh38
rs978285162 Chr.16: 3059007-3059007 on Build GRCh38
rs567271071 Chr.16: 3058955-3058955 on Build GRCh38
rs748416917 Chr.16: 3058704-3058704 on Build GRCh38
rs995856113 Chr.16: 3058985-3058985 on Build GRCh38
rs945528100 Chr.16: 3059117-3059117 on Build GRCh38
rs1018616486 Chr.16: 3058753-3058753 on Build GRCh38
rs763388100 Chr.16: 3058709-3058709 on Build GRCh38
rs193188076 Chr.16: 3059122-3059122 on Build GRCh38
rs758822516 Chr.16: 3059081-3059081 on Build GRCh38
rs78688869 Chr.16: 3058939-3058939 on Build GRCh38
rs753887651 Chr.16: 3059067-3059067 on Build GRCh38
rs928747045 Chr.16: 3058800-3058800 on Build GRCh38
rs1009266928 Chr.16: 3058748-3058748 on Build GRCh38
rs1026614026 Chr.16: 3058716-3058716 on Build GRCh38
rs771382761 Chr.16: 3058876-3058876 on Build GRCh38
rs771091807 Chr.16: 3058960-3058960 on Build GRCh38
rs1000310751 Chr.16: 3059135-3059135 on Build GRCh38
rs978664020 Chr.16: 3058743-3058743 on Build GRCh38
rs867374421 Chr.16: 3058991-3058991 on Build GRCh38
rs754837029 Chr.16: 3058838-3058838 on Build GRCh38
rs930735629 Chr.16: 3059108-3059108 on Build GRCh38
rs886359096 Chr.16: 3058726-3058726 on Build GRCh38
rs930664099 Chr.16: 3059047-3059047 on Build GRCh38
rs528328005 Chr.16: 3059082-3059082 on Build GRCh38
rs1022074926 Chr.16: 3058804-3058804 on Build GRCh38
rs1028662444 Chr.16: 3059144-3059144 on Build GRCh38
rs769053375 Chr.16: 3058837-3058839 on Build GRCh38
rs993016513 Chr.16: 3059074-3059074 on Build GRCh38
rs980793559 Chr.16: 3059063-3059063 on Build GRCh38
rs766234133 Chr.16: 3058910-3058910 on Build GRCh38
rs772562593 Chr.16: 3059000-3059000 on Build GRCh38
rs1031825297 Chr.16: 3059020-3059020 on Build GRCh38
rs749980117 Chr.16: 3058803-3058803 on Build GRCh38
rs750150544 Chr.16: 3058904-3058904 on Build GRCh38
rs773571920 Chr.16: 3058871-3058871 on Build GRCh38
rs1048308132 Chr.16: 3059114-3059114 on Build GRCh38
rs761160421 Chr.16: 3058978-3058978 on Build GRCh38
rs372753938 Chr.16: 3059038-3059038 on Build GRCh38
rs781232240 Chr.16: 3058851-3058851 on Build GRCh38
rs747774238 Chr.16: 3058980-3058980 on Build GRCh38
rs779011016 Chr.16: 3059071-3059071 on Build GRCh38
rs752738148 Chr.16: 3059042-3059042 on Build GRCh38
rs953364502 Chr.16: 3058761-3058761 on Build GRCh38
rs948746593 Chr.16: 3058791-3058791 on Build GRCh38
rs748257213 Chr.16: 3058853-3058853 on Build GRCh38
rs948740319 Chr.16: 3058922-3058922 on Build GRCh38
rs544102381 Chr.16: 3059079-3059079 on Build GRCh38
rs1022019577 Chr.16: 3058988-3058988 on Build GRCh38
rs377201438 Chr.16: 3059101-3059101 on Build GRCh38
rs370064844 Chr.16: 3058790-3058790 on Build GRCh38
rs200340446 Chr.16: 3058899-3058899 on Build GRCh38
rs367827689 Chr.16: 3058873-3058873 on Build GRCh38
rs1040288285 Chr.16: 3059142-3059142 on Build GRCh38
rs939625737 Chr.16: 3059076-3059076 on Build GRCh38
rs751337883 Chr.16: 3058936-3058936 on Build GRCh38
rs930432426 Chr.16: 3058942-3058942 on Build GRCh38
rs765965181 Chr.16: 3059026-3059026 on Build GRCh38
rs1057087925 Chr.16: 3059080-3059080 on Build GRCh38
rs201557156 Chr.16: 3058983-3058983 on Build GRCh38
rs964405153 Chr.16: 3058762-3058762 on Build GRCh38
rs79530442 Chr.16: 3058712-3058712 on Build GRCh38
rs528502666 Chr.16: 3058949-3058949 on Build GRCh38
rs868327401 Chr.16: 3058713-3058713 on Build GRCh38
rs141880798 Chr.16: 3058825-3058825 on Build GRCh38
rs369473134 Chr.16: 3059119-3059119 on Build GRCh38
rs372012951 Chr.16: 3058879-3058879 on Build GRCh38
rs746233138 Chr.16: 3058970-3058970 on Build GRCh38
rs917512177 Chr.16: 3059075-3059075 on Build GRCh38
rs556037619 Chr.16: 3058959-3058959 on Build GRCh38
rs541727512 Chr.16: 3058773-3058773 on Build GRCh38
rs781724086 Chr.16: 3058864-3058865 on Build GRCh38
rs545966266 Chr.16: 3059116-3059116 on Build GRCh38
rs968376419 Chr.16: 3058945-3058945 on Build GRCh38
rs773518604 Chr.16: 3058708-3058708 on Build GRCh38
rs776827299 Chr.16: 3058702-3058702 on Build GRCh38
rs561593513 Chr.16: 3058795-3058795 on Build GRCh38
rs1031976307 Chr.16: 3058732-3058742 on Build GRCh38
rs756097083 Chr.16: 3059058-3059058 on Build GRCh38
rs7188234 Chr.16: 3059031-3059031 on Build GRCh38

View Full Product Details