Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4444275-4444775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
UniGene
Hs.459779
Gene Chromosome Location
Chr.16: 4425805-4456775 on Build GRCh38
Transcripts
NM_001135110.2 NM_001286516.1 NM_005147.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144585 c.953T>A p.V318E Chr.16: 4444685-4444685 on Build GRCh38
1708703 c.956_957GG>AA p.R319K Chr.16: 4444688-4444689 on Build GRCh38
4678640 c.943G>A p.G315S Chr.16: 4444675-4444675 on Build GRCh38
5828385 c.280T>G p.F94V Chr.16: 4444710-4444710 on Build GRCh38
6895068 c.974G>T p.R325M Chr.16: 4444706-4444706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149048405 Chr.16: 4444319-4444319 on Build GRCh38
rs984087589 Chr.16: 4444566-4444566 on Build GRCh38
rs758278552 Chr.16: 4444633-4444633 on Build GRCh38
rs776238026 Chr.16: 4444624-4444624 on Build GRCh38
rs141164294 Chr.16: 4444586-4444586 on Build GRCh38
rs571452954 Chr.16: 4444547-4444547 on Build GRCh38
rs1053237286 Chr.16: 4444334-4444334 on Build GRCh38
rs200540538 Chr.16: 4444722-4444722 on Build GRCh38
rs541077743 Chr.16: 4444410-4444410 on Build GRCh38
rs907874774 Chr.16: 4444573-4444573 on Build GRCh38
rs775282100 Chr.16: 4444614-4444614 on Build GRCh38
rs894992573 Chr.16: 4444335-4444335 on Build GRCh38
rs369168810 Chr.16: 4444626-4444626 on Build GRCh38
rs768425516 Chr.16: 4444750-4444750 on Build GRCh38
rs201740340 Chr.16: 4444668-4444668 on Build GRCh38
rs988821790 Chr.16: 4444461-4444461 on Build GRCh38
rs746706262 Chr.16: 4444746-4444746 on Build GRCh38
rs569774934 Chr.16: 4444462-4444462 on Build GRCh38
rs1033297610 Chr.16: 4444706-4444706 on Build GRCh38
rs751316297 Chr.16: 4444700-4444700 on Build GRCh38
rs925947434 Chr.16: 4444422-4444422 on Build GRCh38
rs12446640 Chr.16: 4444548-4444548 on Build GRCh38
rs573980937 Chr.16: 4444655-4444655 on Build GRCh38
rs769181008 Chr.16: 4444683-4444683 on Build GRCh38
rs1046675387 Chr.16: 4444484-4444484 on Build GRCh38
rs1026619626 Chr.16: 4444312-4444312 on Build GRCh38
rs767670444 Chr.16: 4444709-4444709 on Build GRCh38
rs189567430 Chr.16: 4444350-4444350 on Build GRCh38
rs536564148 Chr.16: 4444625-4444625 on Build GRCh38
rs532142213 Chr.16: 4444488-4444488 on Build GRCh38
rs1003967903 Chr.16: 4444315-4444315 on Build GRCh38
rs933602458 Chr.16: 4444551-4444551 on Build GRCh38
rs766073992 Chr.16: 4444636-4444636 on Build GRCh38
rs776291442 Chr.16: 4444676-4444676 on Build GRCh38
rs545138927 Chr.16: 4444368-4444368 on Build GRCh38
rs777703827 Chr.16: 4444727-4444727 on Build GRCh38
rs781280911 Chr.16: 4444640-4444640 on Build GRCh38
rs757677342 Chr.16: 4444735-4444735 on Build GRCh38
rs950147177 Chr.16: 4444463-4444463 on Build GRCh38
rs934699071 Chr.16: 4444451-4444451 on Build GRCh38
rs755162483 Chr.16: 4444637-4444637 on Build GRCh38
rs779522920 Chr.16: 4444741-4444741 on Build GRCh38
rs879601570 Chr.16: 4444455-4444455 on Build GRCh38
rs766236286 Chr.16: 4444696-4444696 on Build GRCh38
rs527289421 Chr.16: 4444395-4444395 on Build GRCh38
rs954291072 Chr.16: 4444393-4444393 on Build GRCh38
rs749756331 Chr.16: 4444650-4444650 on Build GRCh38
rs774560623 Chr.16: 4444656-4444656 on Build GRCh38
rs750188043 Chr.16: 4444632-4444632 on Build GRCh38
rs905003406 Chr.16: 4444485-4444485 on Build GRCh38
rs773425726 Chr.16: 4444322-4444322 on Build GRCh38
rs529978933 Chr.16: 4444421-4444421 on Build GRCh38
rs560932862 Chr.16: 4444413-4444413 on Build GRCh38
rs749610345 Chr.16: 4444613-4444613 on Build GRCh38
rs551463095 Chr.16: 4444489-4444489 on Build GRCh38
rs201339747 Chr.16: 4444345-4444345 on Build GRCh38
rs759776888 Chr.16: 4444702-4444702 on Build GRCh38
rs888078005 Chr.16: 4444314-4444314 on Build GRCh38
rs963784084 Chr.16: 4444509-4444509 on Build GRCh38
rs761337737 Chr.16: 4444679-4444679 on Build GRCh38
rs888360020 Chr.16: 4444500-4444500 on Build GRCh38
rs567626550 Chr.16: 4444623-4444623 on Build GRCh38
rs762777837 Chr.16: 4444694-4444694 on Build GRCh38
rs752780622 Chr.16: 4444721-4444721 on Build GRCh38
rs893714889 Chr.16: 4444745-4444745 on Build GRCh38
rs779438306 Chr.16: 4444743-4444743 on Build GRCh38
rs906484325 Chr.16: 4444376-4444376 on Build GRCh38
rs143100360 Chr.16: 4444324-4444324 on Build GRCh38
rs868514996 Chr.16: 4444730-4444730 on Build GRCh38
rs773257595 Chr.16: 4444686-4444686 on Build GRCh38
rs1028504483 Chr.16: 4444600-4444600 on Build GRCh38
rs181295539 Chr.16: 4444375-4444375 on Build GRCh38
rs771388805 Chr.16: 4444653-4444653 on Build GRCh38
rs1004318521 Chr.16: 4444528-4444528 on Build GRCh38
rs746687380 Chr.16: 4444657-4444657 on Build GRCh38
rs550027746 Chr.16: 4444446-4444446 on Build GRCh38
rs1038153385 Chr.16: 4444333-4444334 on Build GRCh38
rs748178730 Chr.16: 4444642-4444642 on Build GRCh38
rs765161342 Chr.16: 4444628-4444628 on Build GRCh38
rs999836658 Chr.16: 4444390-4444390 on Build GRCh38
rs746173834 Chr.16: 4444744-4444744 on Build GRCh38
rs941993693 Chr.16: 4444581-4444581 on Build GRCh38
rs750600215 Chr.16: 4444532-4444532 on Build GRCh38
rs756594607 Chr.16: 4444644-4444644 on Build GRCh38

View Full Product Details