Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4811535-4812047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYR1
Entrez Gene ID
84656
Gene Name
glyoxylate reductase 1 homolog
Gene Aliases
BM045, HIBDL, N-PAC, NP60
UniGene
Hs.387255
Gene Chromosome Location
Chr.16: 4803203-4847388 on Build GRCh38
Transcripts
NM_001308096.1 NM_001324096.1 NM_001324097.1 NM_001324098.1 NM_032569.3 NR_136695.1 NR_136696.1 NR_136697.1 NR_136698.1 NR_136699.1 NR_136700.1 XM_005255639.4 XM_011522716.2 XM_011522717.1 XM_017023786.1 XM_017023787.1 XM_017023788.1 XM_017023789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749629 c.1351A>G p.I451V Chr.16: 4811734-4811734 on Build GRCh38
5531752 c.1387C>T p.Q463* Chr.16: 4811698-4811698 on Build GRCh38
5028066 c.1305G>C p.K435N Chr.16: 4811780-4811780 on Build GRCh38
2833296 c.1433C>T p.A478V Chr.16: 4811652-4811652 on Build GRCh38
435317 c.1357G>A p.E453K Chr.16: 4811728-4811728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371241430 Chr.16: 4811792-4811792 on Build GRCh38
rs774376541 Chr.16: 4811840-4811840 on Build GRCh38
rs752856025 Chr.16: 4811904-4811904 on Build GRCh38
rs965143931 Chr.16: 4811875-4811875 on Build GRCh38
rs113260683 Chr.16: 4811604-4811604 on Build GRCh38
rs368646886 Chr.16: 4811657-4811657 on Build GRCh38
rs535460107 Chr.16: 4811877-4811877 on Build GRCh38
rs771017570 Chr.16: 4811885-4811894 on Build GRCh38
rs780924000 Chr.16: 4811694-4811694 on Build GRCh38
rs774328231 Chr.16: 4811718-4811718 on Build GRCh38
rs746207953 Chr.16: 4811667-4811667 on Build GRCh38
rs757796810 Chr.16: 4811765-4811765 on Build GRCh38
rs902466214 Chr.16: 4811771-4811771 on Build GRCh38
rs766291160 Chr.16: 4811583-4811583 on Build GRCh38
rs528959001 Chr.16: 4811913-4811913 on Build GRCh38
rs777762239 Chr.16: 4811633-4811633 on Build GRCh38
rs542406917 Chr.16: 4811996-4811996 on Build GRCh38
rs200073565 Chr.16: 4811849-4811849 on Build GRCh38
rs752966274 Chr.16: 4811732-4811732 on Build GRCh38
rs770988536 Chr.16: 4811716-4811716 on Build GRCh38
rs756248855 Chr.16: 4811607-4811607 on Build GRCh38
rs761563328 Chr.16: 4811636-4811636 on Build GRCh38
rs934203132 Chr.16: 4811976-4811976 on Build GRCh38
rs147198447 Chr.16: 4811576-4811576 on Build GRCh38
rs1004446506 Chr.16: 4811639-4811639 on Build GRCh38
rs566988951 Chr.16: 4811564-4811564 on Build GRCh38
rs1000274835 Chr.16: 4811841-4811841 on Build GRCh38
rs919531392 Chr.16: 4811565-4811565 on Build GRCh38
rs377422986 Chr.16: 4811790-4811790 on Build GRCh38
rs866661965 Chr.16: 4811755-4811755 on Build GRCh38
rs769436673 Chr.16: 4811702-4811702 on Build GRCh38
rs376242476 Chr.16: 4811868-4811868 on Build GRCh38
rs559749728 Chr.16: 4811905-4811905 on Build GRCh38
rs763044128 Chr.16: 4811715-4811715 on Build GRCh38
rs758825584 Chr.16: 4811784-4811784 on Build GRCh38
rs575388850 Chr.16: 4811681-4811681 on Build GRCh38
rs754355505 Chr.16: 4811645-4811645 on Build GRCh38
rs144342530 Chr.16: 4811824-4811824 on Build GRCh38
rs777494249 Chr.16: 4811823-4811823 on Build GRCh38
rs750850526 Chr.16: 4811778-4811778 on Build GRCh38
rs1000304261 Chr.16: 4812023-4812023 on Build GRCh38
rs2085329 Chr.16: 4811708-4811708 on Build GRCh38
rs981967404 Chr.16: 4811888-4811888 on Build GRCh38
rs953882624 Chr.16: 4811842-4811842 on Build GRCh38
rs759324611 Chr.16: 4811843-4811843 on Build GRCh38
rs774201993 Chr.16: 4811569-4811569 on Build GRCh38
rs1032185084 Chr.16: 4811827-4811827 on Build GRCh38
rs368485326 Chr.16: 4811962-4811962 on Build GRCh38
rs901428695 Chr.16: 4811971-4811971 on Build GRCh38
rs760748474 Chr.16: 4811734-4811734 on Build GRCh38
rs918015749 Chr.16: 4812016-4812016 on Build GRCh38
rs1014031149 Chr.16: 4811638-4811638 on Build GRCh38
rs753907173 Chr.16: 4811764-4811764 on Build GRCh38
rs374896672 Chr.16: 4811592-4811592 on Build GRCh38
rs796839531 Chr.16: 4811832-4811832 on Build GRCh38
rs769558013 Chr.16: 4811798-4811798 on Build GRCh38
rs909427008 Chr.16: 4811959-4811959 on Build GRCh38
rs752812369 Chr.16: 4811606-4811606 on Build GRCh38
rs778653028 Chr.16: 4811580-4811580 on Build GRCh38
rs985050694 Chr.16: 4811923-4811923 on Build GRCh38
rs949492466 Chr.16: 4812017-4812017 on Build GRCh38
rs909581852 Chr.16: 4811945-4811945 on Build GRCh38
rs562301356 Chr.16: 4812004-4812004 on Build GRCh38
rs889957304 Chr.16: 4812021-4812021 on Build GRCh38
rs937794802 Chr.16: 4811922-4811922 on Build GRCh38
rs1037083090 Chr.16: 4811970-4811970 on Build GRCh38
rs896547684 Chr.16: 4811614-4811614 on Build GRCh38
rs773316228 Chr.16: 4811572-4811572 on Build GRCh38
rs757745527 Chr.16: 4811649-4811649 on Build GRCh38
rs764212930 Chr.16: 4811753-4811753 on Build GRCh38
rs1049960687 Chr.16: 4811991-4811991 on Build GRCh38
rs779366601 Chr.16: 4811767-4811767 on Build GRCh38
rs754043101 Chr.16: 4811775-4811775 on Build GRCh38
rs538290929 Chr.16: 4811597-4811599 on Build GRCh38
rs755201543 Chr.16: 4811852-4811852 on Build GRCh38
rs1037275262 Chr.16: 4811676-4811676 on Build GRCh38
rs1020883294 Chr.16: 4811860-4811860 on Build GRCh38
rs1033049328 Chr.16: 4811846-4811846 on Build GRCh38
rs751088649 Chr.16: 4811829-4811830 on Build GRCh38
rs983818860 Chr.16: 4811925-4811925 on Build GRCh38
rs759397509 Chr.16: 4811729-4811729 on Build GRCh38

View Full Product Details