Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9759935-9760445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2A
Entrez Gene ID
2903
Gene Name
glutamate ionotropic receptor NMDA type subunit 2A
Gene Aliases
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A
UniGene
Hs.411472
Gene Chromosome Location
Chr.16: 9753404-10183364 on Build GRCh38
Transcripts
NM_000833.4 NM_001134407.2 NM_001134408.2 XM_011522458.2 XM_011522461.2 XM_017023172.1 XM_017023173.1

SNP Detail

SNP ID Chromosome Location
rs983508695 Chr.16: 9760400-9760400 on Build GRCh38
rs1046756810 Chr.16: 9759995-9759995 on Build GRCh38
rs374191190 Chr.16: 9760082-9760082 on Build GRCh38
rs200271631 Chr.16: 9760374-9760374 on Build GRCh38
rs904430933 Chr.16: 9760314-9760314 on Build GRCh38
rs1047428029 Chr.16: 9760115-9760115 on Build GRCh38
rs35189803 Chr.16: 9760398-9760399 on Build GRCh38
rs1021922809 Chr.16: 9760307-9760307 on Build GRCh38
rs930271606 Chr.16: 9760106-9760106 on Build GRCh38
rs930275915 Chr.16: 9760394-9760394 on Build GRCh38
rs1024981523 Chr.16: 9760366-9760366 on Build GRCh38
rs16966263 Chr.16: 9760346-9760346 on Build GRCh38
rs1031125231 Chr.16: 9760155-9760155 on Build GRCh38
rs886052537 Chr.16: 9759983-9759983 on Build GRCh38
rs773324954 Chr.16: 9760032-9760032 on Build GRCh38
rs772997228 Chr.16: 9760374-9760375 on Build GRCh38
rs955520778 Chr.16: 9760011-9760011 on Build GRCh38
rs746299808 Chr.16: 9760358-9760358 on Build GRCh38
rs78329263 Chr.16: 9760252-9760252 on Build GRCh38
rs997145935 Chr.16: 9760127-9760127 on Build GRCh38
rs899772683 Chr.16: 9760091-9760091 on Build GRCh38
rs1040734664 Chr.16: 9760276-9760276 on Build GRCh38
rs899469404 Chr.16: 9760282-9760282 on Build GRCh38
rs769874284 Chr.16: 9759963-9759963 on Build GRCh38
rs886179846 Chr.16: 9760128-9760128 on Build GRCh38
rs200830330 Chr.16: 9760399-9760399 on Build GRCh38
rs905670138 Chr.16: 9760169-9760169 on Build GRCh38
rs988831643 Chr.16: 9759999-9759999 on Build GRCh38
rs996783443 Chr.16: 9760286-9760286 on Build GRCh38
rs949858213 Chr.16: 9760130-9760130 on Build GRCh38
rs550965301 Chr.16: 9759982-9759982 on Build GRCh38
rs956762590 Chr.16: 9760172-9760172 on Build GRCh38
rs955626916 Chr.16: 9760264-9760264 on Build GRCh38
rs184843694 Chr.16: 9760175-9760175 on Build GRCh38
rs1047213109 Chr.16: 9760247-9760247 on Build GRCh38
rs907557216 Chr.16: 9760413-9760413 on Build GRCh38
rs956793806 Chr.16: 9760345-9760345 on Build GRCh38
rs147134369 Chr.16: 9760215-9760215 on Build GRCh38
rs886052539 Chr.16: 9760373-9760374 on Build GRCh38
rs763997430 Chr.16: 9760271-9760271 on Build GRCh38
rs913305012 Chr.16: 9760001-9760001 on Build GRCh38
rs77580194 Chr.16: 9760225-9760225 on Build GRCh38
rs115979840 Chr.16: 9760174-9760174 on Build GRCh38
rs1008542780 Chr.16: 9759983-9759983 on Build GRCh38
rs188578360 Chr.16: 9760263-9760263 on Build GRCh38
rs886052538 Chr.16: 9760314-9760314 on Build GRCh38
rs974213162 Chr.16: 9760356-9760356 on Build GRCh38
rs992183281 Chr.16: 9760173-9760173 on Build GRCh38
rs34869819 Chr.16: 9760399-9760399 on Build GRCh38
rs545566999 Chr.16: 9760373-9760373 on Build GRCh38
rs1045252896 Chr.16: 9759968-9759968 on Build GRCh38
rs898629077 Chr.16: 9759981-9759981 on Build GRCh38
rs977249693 Chr.16: 9760142-9760142 on Build GRCh38
rs372201763 Chr.16: 9760231-9760232 on Build GRCh38
rs987362569 Chr.16: 9760033-9760033 on Build GRCh38
rs559525035 Chr.16: 9760333-9760333 on Build GRCh38
rs920183121 Chr.16: 9760090-9760090 on Build GRCh38
rs749467245 Chr.16: 9760070-9760070 on Build GRCh38
rs886888877 Chr.16: 9760255-9760255 on Build GRCh38
rs969752321 Chr.16: 9760394-9760395 on Build GRCh38
rs957324338 Chr.16: 9759993-9759993 on Build GRCh38
rs562770874 Chr.16: 9760131-9760131 on Build GRCh38
rs1031489441 Chr.16: 9760338-9760338 on Build GRCh38
rs942892294 Chr.16: 9760076-9760076 on Build GRCh38

View Full Product Details