Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10471970-10472503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_011522666.2 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023708.1 XM_017023709.1 XM_017023710.1 XM_017023711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266055 c.1377T>A p.I459I Chr.16: 10472134-10472134 on Build GRCh38
4390389 c.1398G>A p.L466L Chr.16: 10472155-10472155 on Build GRCh38
966910 c.1379C>T p.S460F Chr.16: 10472136-10472136 on Build GRCh38
4888517 c.1404T>C p.T468T Chr.16: 10472161-10472161 on Build GRCh38
4824447 c.1384G>T p.E462* Chr.16: 10472141-10472141 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766546555 Chr.16: 10472371-10472371 on Build GRCh38
rs1000179065 Chr.16: 10472157-10472157 on Build GRCh38
rs144771489 Chr.16: 10472160-10472160 on Build GRCh38
rs367708962 Chr.16: 10472154-10472154 on Build GRCh38
rs767570210 Chr.16: 10472114-10472116 on Build GRCh38
rs901400781 Chr.16: 10472443-10472443 on Build GRCh38
rs566280429 Chr.16: 10472122-10472124 on Build GRCh38
rs966743968 Chr.16: 10472390-10472390 on Build GRCh38
rs546281888 Chr.16: 10472094-10472094 on Build GRCh38
rs763389327 Chr.16: 10472165-10472165 on Build GRCh38
rs572369241 Chr.16: 10472474-10472474 on Build GRCh38
rs957971079 Chr.16: 10472321-10472321 on Build GRCh38
rs1004464946 Chr.16: 10472019-10472019 on Build GRCh38
rs1024770207 Chr.16: 10472290-10472290 on Build GRCh38
rs752653223 Chr.16: 10472211-10472211 on Build GRCh38
rs926578609 Chr.16: 10472012-10472012 on Build GRCh38
rs945162793 Chr.16: 10472066-10472066 on Build GRCh38
rs73497902 Chr.16: 10472278-10472278 on Build GRCh38
rs563151134 Chr.16: 10472237-10472237 on Build GRCh38
rs768082632 Chr.16: 10472203-10472203 on Build GRCh38
rs4453490 Chr.16: 10472289-10472289 on Build GRCh38
rs752163858 Chr.16: 10472089-10472089 on Build GRCh38
rs901344222 Chr.16: 10472079-10472079 on Build GRCh38
rs770154400 Chr.16: 10472119-10472119 on Build GRCh38
rs548765832 Chr.16: 10472266-10472266 on Build GRCh38
rs566384462 Chr.16: 10472304-10472304 on Build GRCh38
rs778166623 Chr.16: 10472184-10472185 on Build GRCh38
rs756250843 Chr.16: 10472213-10472213 on Build GRCh38
rs138524305 Chr.16: 10472145-10472145 on Build GRCh38
rs943092381 Chr.16: 10472430-10472430 on Build GRCh38
rs781493149 Chr.16: 10472090-10472090 on Build GRCh38
rs745612326 Chr.16: 10472113-10472113 on Build GRCh38
rs376890755 Chr.16: 10472180-10472180 on Build GRCh38
rs1037235766 Chr.16: 10472030-10472030 on Build GRCh38
rs750683218 Chr.16: 10472064-10472064 on Build GRCh38
rs766893851 Chr.16: 10472169-10472169 on Build GRCh38
rs930929923 Chr.16: 10472463-10472463 on Build GRCh38
rs989144801 Chr.16: 10472243-10472243 on Build GRCh38
rs778062793 Chr.16: 10472095-10472095 on Build GRCh38
rs771900989 Chr.16: 10472115-10472115 on Build GRCh38
rs1039250493 Chr.16: 10472442-10472442 on Build GRCh38
rs542623480 Chr.16: 10472142-10472142 on Build GRCh38
rs538353066 Chr.16: 10472210-10472210 on Build GRCh38
rs758861771 Chr.16: 10472065-10472065 on Build GRCh38
rs1033458818 Chr.16: 10472316-10472316 on Build GRCh38
rs756604565 Chr.16: 10472163-10472168 on Build GRCh38
rs754097979 Chr.16: 10472231-10472231 on Build GRCh38
rs777811901 Chr.16: 10472218-10472218 on Build GRCh38
rs1051922271 Chr.16: 10472129-10472129 on Build GRCh38
rs954097702 Chr.16: 10472153-10472153 on Build GRCh38
rs180738991 Chr.16: 10472048-10472048 on Build GRCh38
rs755258610 Chr.16: 10472084-10472084 on Build GRCh38
rs754089942 Chr.16: 10472084-10472084 on Build GRCh38
rs768140924 Chr.16: 10472151-10472151 on Build GRCh38
rs752110961 Chr.16: 10472171-10472171 on Build GRCh38
rs1048418166 Chr.16: 10472469-10472469 on Build GRCh38
rs911683610 Chr.16: 10472412-10472412 on Build GRCh38
rs971657513 Chr.16: 10472259-10472259 on Build GRCh38
rs571907452 Chr.16: 10472118-10472118 on Build GRCh38
rs538623135 Chr.16: 10472417-10472417 on Build GRCh38
rs899450627 Chr.16: 10472215-10472215 on Build GRCh38
rs373609538 Chr.16: 10472148-10472148 on Build GRCh38
rs987242043 Chr.16: 10472401-10472401 on Build GRCh38
rs1026340241 Chr.16: 10472249-10472249 on Build GRCh38
rs936582872 Chr.16: 10472035-10472035 on Build GRCh38
rs989779742 Chr.16: 10472360-10472360 on Build GRCh38
rs546701454 Chr.16: 10472301-10472301 on Build GRCh38
rs1048745617 Chr.16: 10472000-10472000 on Build GRCh38
rs1048798692 Chr.16: 10472326-10472326 on Build GRCh38
rs199605421 Chr.16: 10472124-10472124 on Build GRCh38
rs531821400 Chr.16: 10472263-10472263 on Build GRCh38
rs989189539 Chr.16: 10472045-10472045 on Build GRCh38
rs766941125 Chr.16: 10472068-10472068 on Build GRCh38
rs970111993 Chr.16: 10472293-10472293 on Build GRCh38
rs1014709330 Chr.16: 10472475-10472475 on Build GRCh38
rs371612637 Chr.16: 10472155-10472155 on Build GRCh38
rs920745333 Chr.16: 10472456-10472456 on Build GRCh38
rs888724994 Chr.16: 10472016-10472016 on Build GRCh38

View Full Product Details