Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29806802-29807342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAZ
Entrez Gene ID
4150
Gene Name
MYC associated zinc finger protein
Gene Aliases
PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87
UniGene
Hs.23650
Gene Chromosome Location
Chr.16: 29806096-29811183 on Build GRCh38
Transcripts
NM_001042539.2 NM_001276275.1 NM_001276276.1 NM_002383.3 NR_074080.1 XM_006721047.3 XM_006721048.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442453 c.106G>A p.G36S Chr.16: 29806807-29806807 on Build GRCh38
4059939 c.122C>G p.P41R Chr.16: 29806823-29806823 on Build GRCh38
5726244 c.170G>A p.G57D Chr.16: 29806871-29806871 on Build GRCh38
4059941 c.165C>T p.S55S Chr.16: 29806866-29806866 on Build GRCh38
1743762 c.125T>A p.L42Q Chr.16: 29806826-29806826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201551085 Chr.16: 29807281-29807281 on Build GRCh38
rs988520613 Chr.16: 29807023-29807023 on Build GRCh38
rs891131907 Chr.16: 29807100-29807101 on Build GRCh38
rs532233987 Chr.16: 29806932-29806932 on Build GRCh38
rs763758729 Chr.16: 29806923-29806924 on Build GRCh38
rs571410983 Chr.16: 29806931-29806931 on Build GRCh38
rs776747054 Chr.16: 29806924-29806924 on Build GRCh38
rs544305382 Chr.16: 29807306-29807306 on Build GRCh38
rs576787515 Chr.16: 29807305-29807305 on Build GRCh38
rs750343342 Chr.16: 29806902-29806902 on Build GRCh38
rs760281541 Chr.16: 29806897-29806899 on Build GRCh38
rs779781775 Chr.16: 29806904-29806904 on Build GRCh38
rs762011955 Chr.16: 29806944-29806944 on Build GRCh38
rs755780142 Chr.16: 29807240-29807240 on Build GRCh38
rs893145375 Chr.16: 29807109-29807110 on Build GRCh38
rs550654524 Chr.16: 29806939-29806939 on Build GRCh38
rs752936281 Chr.16: 29806822-29806822 on Build GRCh38
rs772612921 Chr.16: 29806933-29806933 on Build GRCh38
rs1054528416 Chr.16: 29807102-29807103 on Build GRCh38
rs762705565 Chr.16: 29807180-29807188 on Build GRCh38
rs780073980 Chr.16: 29806936-29806936 on Build GRCh38
rs749627105 Chr.16: 29806906-29806906 on Build GRCh38
rs775622770 Chr.16: 29806869-29806869 on Build GRCh38
rs957086362 Chr.16: 29807124-29807124 on Build GRCh38
rs1047748685 Chr.16: 29806945-29806946 on Build GRCh38
rs199539324 Chr.16: 29806910-29806910 on Build GRCh38
rs767598451 Chr.16: 29806865-29806865 on Build GRCh38
rs758356460 Chr.16: 29807080-29807088 on Build GRCh38
rs758791535 Chr.16: 29807269-29807269 on Build GRCh38
rs760950274 Chr.16: 29806870-29806870 on Build GRCh38
rs772930063 Chr.16: 29806954-29806954 on Build GRCh38
rs200519536 Chr.16: 29807316-29807316 on Build GRCh38
rs749735908 Chr.16: 29806851-29806851 on Build GRCh38
rs971474353 Chr.16: 29807150-29807150 on Build GRCh38
rs767684774 Chr.16: 29806821-29806821 on Build GRCh38
rs1019914856 Chr.16: 29807142-29807142 on Build GRCh38
rs996693832 Chr.16: 29807101-29807112 on Build GRCh38
rs757209598 Chr.16: 29807209-29807214 on Build GRCh38
rs745421122 Chr.16: 29806833-29806833 on Build GRCh38
rs200279675 Chr.16: 29807313-29807313 on Build GRCh38
rs536032245 Chr.16: 29807214-29807214 on Build GRCh38
rs981378159 Chr.16: 29807176-29807176 on Build GRCh38
rs758472099 Chr.16: 29806926-29806926 on Build GRCh38
rs558521196 Chr.16: 29807294-29807294 on Build GRCh38
rs761614851 Chr.16: 29806925-29806925 on Build GRCh38
rs759056981 Chr.16: 29807184-29807184 on Build GRCh38
rs750822992 Chr.16: 29807267-29807267 on Build GRCh38
rs746646924 Chr.16: 29806905-29806905 on Build GRCh38
rs767039661 Chr.16: 29807300-29807300 on Build GRCh38
rs1010416119 Chr.16: 29807129-29807129 on Build GRCh38
rs779951400 Chr.16: 29807089-29807100 on Build GRCh38
rs772133943 Chr.16: 29806919-29806919 on Build GRCh38
rs777547895 Chr.16: 29806823-29806823 on Build GRCh38
rs199581929 Chr.16: 29807264-29807264 on Build GRCh38
rs759611940 Chr.16: 29807295-29807295 on Build GRCh38
rs762952781 Chr.16: 29806965-29806965 on Build GRCh38
rs746668933 Chr.16: 29806843-29806843 on Build GRCh38
rs780466757 Chr.16: 29807309-29807309 on Build GRCh38
rs780487188 Chr.16: 29807271-29807271 on Build GRCh38
rs1008912031 Chr.16: 29806967-29806967 on Build GRCh38
rs768779626 Chr.16: 29807274-29807274 on Build GRCh38
rs971310516 Chr.16: 29807058-29807058 on Build GRCh38
rs1048726245 Chr.16: 29807193-29807193 on Build GRCh38
rs1019090206 Chr.16: 29806847-29806847 on Build GRCh38
rs778085863 Chr.16: 29807315-29807315 on Build GRCh38
rs756759816 Chr.16: 29806925-29806925 on Build GRCh38
rs747767840 Chr.16: 29806928-29806928 on Build GRCh38
rs747047927 Chr.16: 29806938-29806938 on Build GRCh38
rs989128490 Chr.16: 29806920-29806920 on Build GRCh38
rs746822421 Chr.16: 29807101-29807109 on Build GRCh38
rs368747764 Chr.16: 29806853-29806853 on Build GRCh38
rs757107607 Chr.16: 29806885-29806885 on Build GRCh38
rs774752780 Chr.16: 29806857-29806857 on Build GRCh38
rs1047596096 Chr.16: 29807223-29807223 on Build GRCh38
rs937214255 Chr.16: 29807076-29807076 on Build GRCh38
rs747522546 Chr.16: 29807272-29807272 on Build GRCh38
rs753767784 Chr.16: 29806824-29806824 on Build GRCh38
rs763858893 Chr.16: 29806876-29806876 on Build GRCh38
rs753714134 Chr.16: 29806878-29806878 on Build GRCh38
rs753325046 Chr.16: 29806924-29806925 on Build GRCh38
rs765206077 Chr.16: 29806933-29806934 on Build GRCh38
rs981774567 Chr.16: 29807071-29807079 on Build GRCh38
rs755491921 Chr.16: 29807310-29807310 on Build GRCh38
rs973899554 Chr.16: 29807196-29807196 on Build GRCh38
rs774862411 Chr.16: 29806909-29806909 on Build GRCh38
rs752193277 Chr.16: 29807302-29807302 on Build GRCh38
rs930046769 Chr.16: 29807221-29807221 on Build GRCh38
rs770864200 Chr.16: 29807291-29807291 on Build GRCh38
rs773771891 Chr.16: 29806845-29806845 on Build GRCh38
rs995926567 Chr.16: 29807010-29807010 on Build GRCh38
rs766381022 Chr.16: 29807091-29807091 on Build GRCh38
rs546595080 Chr.16: 29806929-29806929 on Build GRCh38
rs773515431 Chr.16: 29807283-29807283 on Build GRCh38
rs569075875 Chr.16: 29807106-29807106 on Build GRCh38
rs929946715 Chr.16: 29807095-29807103 on Build GRCh38
rs778262536 Chr.16: 29806830-29806830 on Build GRCh38
rs771220812 Chr.16: 29806908-29806908 on Build GRCh38
rs928948004 Chr.16: 29807019-29807019 on Build GRCh38
rs776848757 Chr.16: 29806941-29806941 on Build GRCh38
rs756381325 Chr.16: 29807194-29807230 on Build GRCh38
rs750267503 Chr.16: 29807074-29807082 on Build GRCh38
rs760860266 Chr.16: 29806921-29806921 on Build GRCh38
rs754567940 Chr.16: 29806927-29806927 on Build GRCh38
rs964212318 Chr.16: 29806999-29806999 on Build GRCh38
rs766939790 Chr.16: 29807226-29807226 on Build GRCh38
rs900672278 Chr.16: 29806976-29806977 on Build GRCh38
rs779798066 Chr.16: 29806841-29806841 on Build GRCh38
rs764337589 Chr.16: 29806923-29806923 on Build GRCh38
rs1032745575 Chr.16: 29807111-29807111 on Build GRCh38
rs771508618 Chr.16: 29806834-29806834 on Build GRCh38
rs757272498 Chr.16: 29806825-29806825 on Build GRCh38
rs758697723 Chr.16: 29807307-29807307 on Build GRCh38
rs748138200 Chr.16: 29807311-29807311 on Build GRCh38
rs554863365 Chr.16: 29807238-29807238 on Build GRCh38
rs779310779 Chr.16: 29807246-29807246 on Build GRCh38
rs1009949502 Chr.16: 29807308-29807308 on Build GRCh38
rs528121327 Chr.16: 29806901-29806901 on Build GRCh38
rs534246223 Chr.16: 29807289-29807289 on Build GRCh38
rs748384422 Chr.16: 29807282-29807282 on Build GRCh38
rs1047033609 Chr.16: 29807100-29807100 on Build GRCh38
rs769621729 Chr.16: 29806953-29806953 on Build GRCh38
rs768740117 Chr.16: 29806940-29806940 on Build GRCh38

View Full Product Details