Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29995795-29996289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4 NM_001304562.1 NM_001304563.1 NM_173618.2 NR_130786.1 NR_130787.1 NR_134855.1 XM_011545809.2 XM_011545811.2 XM_011545812.2 XM_017023169.1

Gene Symbol
INO80E
Entrez Gene ID
283899
Gene Name
INO80 complex subunit E
Gene Aliases
CCDC95
UniGene
Hs.434864
Gene Chromosome Location
Chr.16: 29996209-30005794 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4 NM_001304562.1 NM_001304563.1 NM_173618.2 NR_130786.1 NR_130787.1 NR_134855.1 XM_011545809.2 XM_011545811.2 XM_011545812.2 XM_017023169.1

SNP Detail

SNP ID Chromosome Location
rs991428547 Chr.16: 29996088-29996088 on Build GRCh38
rs959026371 Chr.16: 29995930-29995930 on Build GRCh38
rs141937893 Chr.16: 29996193-29996193 on Build GRCh38
rs565296002 Chr.16: 29996104-29996104 on Build GRCh38
rs981999081 Chr.16: 29995871-29995871 on Build GRCh38
rs766761279 Chr.16: 29996228-29996228 on Build GRCh38
rs778694067 Chr.16: 29996263-29996263 on Build GRCh38
rs542404365 Chr.16: 29996143-29996143 on Build GRCh38
rs192213643 Chr.16: 29996037-29996037 on Build GRCh38
rs60185882 Chr.16: 29995942-29995942 on Build GRCh38
rs771761356 Chr.16: 29996200-29996200 on Build GRCh38
rs544926594 Chr.16: 29995888-29995888 on Build GRCh38
rs1036124674 Chr.16: 29996157-29996157 on Build GRCh38
rs760474749 Chr.16: 29996206-29996206 on Build GRCh38
rs940167293 Chr.16: 29996156-29996156 on Build GRCh38
rs1032424908 Chr.16: 29995841-29995841 on Build GRCh38
rs931587572 Chr.16: 29996113-29996113 on Build GRCh38
rs551304122 Chr.16: 29996127-29996127 on Build GRCh38
rs1041311112 Chr.16: 29996014-29996014 on Build GRCh38
rs983613134 Chr.16: 29996167-29996167 on Build GRCh38
rs534728857 Chr.16: 29996202-29996202 on Build GRCh38
rs985783802 Chr.16: 29996119-29996119 on Build GRCh38
rs997153434 Chr.16: 29995936-29995936 on Build GRCh38
rs973012049 Chr.16: 29996160-29996160 on Build GRCh38
rs1010085659 Chr.16: 29996110-29996110 on Build GRCh38
rs932620388 Chr.16: 29996038-29996038 on Build GRCh38
rs928679576 Chr.16: 29995823-29995823 on Build GRCh38
rs990069600 Chr.16: 29995996-29995996 on Build GRCh38
rs570914942 Chr.16: 29996243-29996243 on Build GRCh38
rs74460967 Chr.16: 29996161-29996161 on Build GRCh38
rs944201305 Chr.16: 29996185-29996185 on Build GRCh38
rs893906145 Chr.16: 29995861-29995861 on Build GRCh38
rs748998623 Chr.16: 29996251-29996251 on Build GRCh38
rs763404319 Chr.16: 29996222-29996222 on Build GRCh38
rs1011948899 Chr.16: 29996184-29996184 on Build GRCh38
rs534298902 Chr.16: 29995840-29995840 on Build GRCh38
rs887271836 Chr.16: 29995901-29995901 on Build GRCh38
rs538507892 Chr.16: 29996245-29996245 on Build GRCh38
rs1033556998 Chr.16: 29996148-29996148 on Build GRCh38
rs528614717 Chr.16: 29996026-29996026 on Build GRCh38
rs982946364 Chr.16: 29995993-29995993 on Build GRCh38
rs970648914 Chr.16: 29995868-29995868 on Build GRCh38
rs920330109 Chr.16: 29995974-29995974 on Build GRCh38
rs959308384 Chr.16: 29996159-29996159 on Build GRCh38
rs1021221379 Chr.16: 29995863-29995863 on Build GRCh38
rs994713985 Chr.16: 29996266-29996266 on Build GRCh38
rs762125688 Chr.16: 29995958-29995958 on Build GRCh38
rs567231861 Chr.16: 29996201-29996201 on Build GRCh38
rs1054895607 Chr.16: 29996261-29996261 on Build GRCh38
rs929059463 Chr.16: 29995976-29995976 on Build GRCh38
rs911679378 Chr.16: 29996170-29996170 on Build GRCh38
rs752106106 Chr.16: 29996230-29996230 on Build GRCh38
rs778081216 Chr.16: 29996249-29996249 on Build GRCh38
rs115906553 Chr.16: 29996124-29996124 on Build GRCh38
rs569610299 Chr.16: 29996145-29996145 on Build GRCh38
rs552965372 Chr.16: 29995860-29995860 on Build GRCh38
rs915948389 Chr.16: 29996252-29996252 on Build GRCh38
rs952703589 Chr.16: 29996264-29996264 on Build GRCh38
rs912364930 Chr.16: 29996134-29996134 on Build GRCh38
rs1049065230 Chr.16: 29995894-29995894 on Build GRCh38
rs756270119 Chr.16: 29996248-29996248 on Build GRCh38
rs758380981 Chr.16: 29996166-29996166 on Build GRCh38
rs750584471 Chr.16: 29996215-29996215 on Build GRCh38
rs77199037 Chr.16: 29995879-29995879 on Build GRCh38
rs552929565 Chr.16: 29996218-29996218 on Build GRCh38
rs1014641377 Chr.16: 29995920-29995920 on Build GRCh38
rs915231306 Chr.16: 29996214-29996214 on Build GRCh38
rs1005078146 Chr.16: 29995915-29995915 on Build GRCh38
rs145052397 Chr.16: 29996013-29996013 on Build GRCh38
rs556692775 Chr.16: 29995939-29995939 on Build GRCh38
rs1003367469 Chr.16: 29996125-29996125 on Build GRCh38
rs866064237 Chr.16: 29996151-29996151 on Build GRCh38
rs1055979942 Chr.16: 29995854-29995854 on Build GRCh38
rs56183743 Chr.16: 29996004-29996004 on Build GRCh38
rs757116886 Chr.16: 29996254-29996254 on Build GRCh38
rs755037713 Chr.16: 29996241-29996241 on Build GRCh38
rs953297794 Chr.16: 29995971-29995971 on Build GRCh38
rs752100036 Chr.16: 29995862-29995862 on Build GRCh38
rs138833603 Chr.16: 29996042-29996042 on Build GRCh38
rs1020748936 Chr.16: 29996121-29996121 on Build GRCh38
rs879182965 Chr.16: 29996152-29996152 on Build GRCh38
rs893862375 Chr.16: 29996069-29996069 on Build GRCh38

View Full Product Details