Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29996455-29996954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80E
Entrez Gene ID
283899
Gene Name
INO80 complex subunit E
Gene Aliases
CCDC95
UniGene
Hs.434864
Gene Chromosome Location
Chr.16: 29996209-30005794 on Build GRCh38
Transcripts
NM_001304562.1 NM_001304563.1 NM_173618.2 NR_130786.1 NR_130787.1 NR_134855.1 XM_011545809.2 XM_011545811.2 XM_011545812.2 XM_017023169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3711917 c.82-1G>A p.? Chr.16: 29996546-29996546 on Build GRCh38
7079112 c.193G>A p.E65K Chr.16: 29996848-29996848 on Build GRCh38
3111584 c.163G>T p.D55Y Chr.16: 29996818-29996818 on Build GRCh38
3508707 c.185A>G p.N62S Chr.16: 29996840-29996840 on Build GRCh38
7412337 c.173T>A p.L58Q Chr.16: 29996828-29996828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748998325 Chr.16: 29996561-29996561 on Build GRCh38
rs755135463 Chr.16: 29996636-29996636 on Build GRCh38
rs954179752 Chr.16: 29996752-29996752 on Build GRCh38
rs148592234 Chr.16: 29996703-29996703 on Build GRCh38
rs994662914 Chr.16: 29996546-29996546 on Build GRCh38
rs775494495 Chr.16: 29996803-29996803 on Build GRCh38
rs1056975875 Chr.16: 29996725-29996725 on Build GRCh38
rs776391289 Chr.16: 29996872-29996872 on Build GRCh38
rs771821825 Chr.16: 29996801-29996801 on Build GRCh38
rs369415677 Chr.16: 29996924-29996924 on Build GRCh38
rs753300078 Chr.16: 29996835-29996835 on Build GRCh38
rs750098054 Chr.16: 29996841-29996841 on Build GRCh38
rs752209937 Chr.16: 29996538-29996538 on Build GRCh38
rs569042356 Chr.16: 29996851-29996851 on Build GRCh38
rs746863593 Chr.16: 29996797-29996797 on Build GRCh38
rs766721370 Chr.16: 29996635-29996635 on Build GRCh38
rs780402845 Chr.16: 29996595-29996595 on Build GRCh38
rs189412116 Chr.16: 29996684-29996684 on Build GRCh38
rs376608569 Chr.16: 29996787-29996787 on Build GRCh38
rs563150678 Chr.16: 29996527-29996527 on Build GRCh38
rs761824058 Chr.16: 29996505-29996505 on Build GRCh38
rs763199745 Chr.16: 29996634-29996634 on Build GRCh38
rs952834157 Chr.16: 29996536-29996536 on Build GRCh38
rs573001840 Chr.16: 29996920-29996920 on Build GRCh38
rs761295725 Chr.16: 29996909-29996909 on Build GRCh38
rs376449564 Chr.16: 29996681-29996681 on Build GRCh38
rs563768061 Chr.16: 29996694-29996694 on Build GRCh38
rs756312938 Chr.16: 29996800-29996801 on Build GRCh38
rs773559761 Chr.16: 29996632-29996632 on Build GRCh38
rs115295336 Chr.16: 29996767-29996767 on Build GRCh38
rs1049447798 Chr.16: 29996616-29996616 on Build GRCh38
rs112386063 Chr.16: 29996796-29996796 on Build GRCh38
rs763662275 Chr.16: 29996664-29996664 on Build GRCh38
rs976994166 Chr.16: 29996605-29996605 on Build GRCh38
rs753353801 Chr.16: 29996665-29996665 on Build GRCh38
rs754886932 Chr.16: 29996531-29996531 on Build GRCh38
rs778455003 Chr.16: 29996760-29996760 on Build GRCh38
rs908520168 Chr.16: 29996563-29996563 on Build GRCh38
rs755344744 Chr.16: 29996765-29996765 on Build GRCh38
rs779184151 Chr.16: 29996868-29996868 on Build GRCh38
rs927989456 Chr.16: 29996818-29996818 on Build GRCh38
rs544664924 Chr.16: 29996513-29996513 on Build GRCh38
rs769159875 Chr.16: 29996608-29996608 on Build GRCh38
rs774144348 Chr.16: 29996637-29996637 on Build GRCh38
rs748233433 Chr.16: 29996619-29996619 on Build GRCh38
rs748132424 Chr.16: 29996686-29996686 on Build GRCh38
rs928007874 Chr.16: 29996518-29996518 on Build GRCh38
rs770426374 Chr.16: 29996776-29996776 on Build GRCh38
rs777727242 Chr.16: 29996785-29996785 on Build GRCh38
rs55963099 Chr.16: 29996528-29996528 on Build GRCh38
rs770966981 Chr.16: 29996858-29996858 on Build GRCh38
rs770474513 Chr.16: 29996822-29996822 on Build GRCh38
rs764036416 Chr.16: 29996824-29996824 on Build GRCh38
rs777038099 Chr.16: 29996614-29996614 on Build GRCh38
rs998712665 Chr.16: 29996507-29996507 on Build GRCh38
rs538247025 Chr.16: 29996779-29996779 on Build GRCh38
rs368841674 Chr.16: 29996856-29996856 on Build GRCh38
rs747362864 Chr.16: 29996762-29996762 on Build GRCh38
rs368205302 Chr.16: 29996781-29996781 on Build GRCh38
rs769052020 Chr.16: 29996497-29996497 on Build GRCh38
rs184542030 Chr.16: 29996625-29996625 on Build GRCh38
rs781341435 Chr.16: 29996842-29996842 on Build GRCh38
rs548456485 Chr.16: 29996602-29996602 on Build GRCh38
rs752628348 Chr.16: 29996648-29996648 on Build GRCh38
rs760489989 Chr.16: 29996893-29996893 on Build GRCh38
rs916687788 Chr.16: 29996919-29996919 on Build GRCh38
rs755609366 Chr.16: 29996650-29996650 on Build GRCh38
rs774476134 Chr.16: 29996790-29996790 on Build GRCh38
rs753124614 Chr.16: 29996846-29996846 on Build GRCh38
rs746143717 Chr.16: 29996884-29996884 on Build GRCh38
rs759279678 Chr.16: 29996639-29996639 on Build GRCh38
rs745338824 Chr.16: 29996899-29996899 on Build GRCh38
rs372023618 Chr.16: 29996642-29996642 on Build GRCh38
rs762824372 Chr.16: 29996520-29996520 on Build GRCh38
rs184528092 Chr.16: 29996925-29996925 on Build GRCh38
rs755662691 Chr.16: 29996544-29996544 on Build GRCh38
rs947189084 Chr.16: 29996928-29996928 on Build GRCh38
rs781713457 Chr.16: 29996766-29996766 on Build GRCh38
rs764193575 Chr.16: 29996805-29996806 on Build GRCh38
rs761346635 Chr.16: 29996836-29996836 on Build GRCh38
rs772343501 Chr.16: 29996885-29996885 on Build GRCh38
rs776717553 Chr.16: 29996500-29996500 on Build GRCh38
rs191800755 Chr.16: 29996890-29996890 on Build GRCh38
rs1042417449 Chr.16: 29996475-29996475 on Build GRCh38
rs777384606 Chr.16: 29996552-29996552 on Build GRCh38
rs536565115 Chr.16: 29996854-29996854 on Build GRCh38
rs758758778 Chr.16: 29996584-29996584 on Build GRCh38
rs773614841 Chr.16: 29996515-29996515 on Build GRCh38
rs756850190 Chr.16: 29996758-29996758 on Build GRCh38

View Full Product Details